Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 72207723-72208264 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ERCC6L
Entrez Gene ID
54821
Gene Name
ERCC excision repair 6 like, spindle assembly checkpoint helicase
Gene Aliases
PICH, RAD26L
UniGene
Hs.47558
Gene Chromosome Location
Chr.X: 72204657-72239008 on Build GRCh38
Transcripts
NM_017669.2 NM_001170747.1

Gene Symbol
PIN4
Entrez Gene ID
5303
Gene Name
peptidylprolyl cis/trans isomerase, NIMA-interacting 4
Gene Aliases
EPVH, PAR14, PAR17
UniGene
Hs.655623
Gene Chromosome Location
Chr.X: 72181676-72263964 on Build GRCh38
Transcripts
NM_017669.2 NM_001170747.1

SNP Detail

SNP ID Chromosome Location
rs760640928 Chr.X: 72208220-72208220 on Build GRCh38
rs768473946 Chr.X: 72208185-72208185 on Build GRCh38
rs765699023 Chr.X: 72208236-72208236 on Build GRCh38
rs766505796 Chr.X: 72208023-72208023 on Build GRCh38
rs749073988 Chr.X: 72207897-72207897 on Build GRCh38
rs771327951 Chr.X: 72208224-72208224 on Build GRCh38
rs773897881 Chr.X: 72208210-72208210 on Build GRCh38
rs780525195 Chr.X: 72207771-72207771 on Build GRCh38
rs751239411 Chr.X: 72208161-72208161 on Build GRCh38
rs775318402 Chr.X: 72207862-72207862 on Build GRCh38
rs774298340 Chr.X: 72207965-72207965 on Build GRCh38
rs748131332 Chr.X: 72208199-72208199 on Build GRCh38
rs1035123324 Chr.X: 72207858-72207858 on Build GRCh38
rs151269287 Chr.X: 72208033-72208033 on Build GRCh38
rs753051049 Chr.X: 72208242-72208242 on Build GRCh38
rs745535504 Chr.X: 72207780-72207780 on Build GRCh38
rs768633896 Chr.X: 72207945-72207945 on Build GRCh38
rs374124933 Chr.X: 72207989-72207989 on Build GRCh38
rs1004045390 Chr.X: 72207788-72207788 on Build GRCh38
rs753286014 Chr.X: 72208062-72208062 on Build GRCh38
rs777014115 Chr.X: 72208225-72208225 on Build GRCh38
rs755858395 Chr.X: 72208095-72208095 on Build GRCh38
rs138446302 Chr.X: 72208206-72208206 on Build GRCh38
rs769270474 Chr.X: 72207796-72207796 on Build GRCh38
rs759122393 Chr.X: 72208231-72208231 on Build GRCh38
rs758762458 Chr.X: 72208063-72208063 on Build GRCh38
rs765647521 Chr.X: 72208044-72208044 on Build GRCh38
rs200143714 Chr.X: 72208083-72208083 on Build GRCh38
rs746565139 Chr.X: 72207770-72207770 on Build GRCh38
rs182482991 Chr.X: 72208038-72208038 on Build GRCh38
rs778615639 Chr.X: 72208198-72208198 on Build GRCh38
rs779662839 Chr.X: 72208134-72208134 on Build GRCh38
rs142293032 Chr.X: 72207966-72207966 on Build GRCh38
rs1056896422 Chr.X: 72207777-72207777 on Build GRCh38
rs201800132 Chr.X: 72208012-72208012 on Build GRCh38
rs918253192 Chr.X: 72208217-72208217 on Build GRCh38
rs773109937 Chr.X: 72207999-72207999 on Build GRCh38
rs142549919 Chr.X: 72208201-72208201 on Build GRCh38

View Full Product Details