Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 73563070-73563567 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHIC1
Entrez Gene ID
53344
Gene Name
cysteine rich hydrophobic domain 1
Gene Aliases
BRX
UniGene
Hs.496323
Gene Chromosome Location
Chr.X: 73563045-73687109 on Build GRCh38
Transcripts
NM_001039840.2 NM_001300884.1 XM_017029582.1 XM_017029584.1 NR_029423.1

Gene Symbol
MAP2K4P1
Entrez Gene ID
139201
Gene Name
mitogen-activated protein kinase kinase 4 pseudogene 1
Gene Aliases
-
UniGene
Hs.586477
Gene Chromosome Location
Chr.X: 73524275-73563085 on Build GRCh38
Transcripts
NM_001039840.2 NM_001300884.1 XM_017029582.1 XM_017029584.1 NR_029423.1

SNP Detail

SNP ID Chromosome Location
rs761055802 Chr.X: 73563348-73563350 on Build GRCh38
rs771438204 Chr.X: 73563363-73563364 on Build GRCh38
rs762302121 Chr.X: 73563471-73563476 on Build GRCh38
rs147848664 Chr.X: 73563521-73563521 on Build GRCh38
rs975154572 Chr.X: 73563254-73563254 on Build GRCh38
rs747845503 Chr.X: 73563326-73563326 on Build GRCh38
rs751226621 Chr.X: 73563426-73563434 on Build GRCh38
rs901554604 Chr.X: 73563233-73563233 on Build GRCh38
rs1041664399 Chr.X: 73563538-73563538 on Build GRCh38
rs765880949 Chr.X: 73563474-73563474 on Build GRCh38
rs771379222 Chr.X: 73563459-73563473 on Build GRCh38
rs200762232 Chr.X: 73563339-73563339 on Build GRCh38
rs765811970 Chr.X: 73563378-73563379 on Build GRCh38
rs1040524770 Chr.X: 73563139-73563139 on Build GRCh38
rs1013572963 Chr.X: 73563439-73563439 on Build GRCh38
rs754644575 Chr.X: 73563491-73563491 on Build GRCh38
rs112505584 Chr.X: 73563504-73563504 on Build GRCh38
rs892731239 Chr.X: 73563502-73563502 on Build GRCh38
rs772200466 Chr.X: 73563447-73563461 on Build GRCh38
rs1010993295 Chr.X: 73563539-73563539 on Build GRCh38
rs895327658 Chr.X: 73563364-73563381 on Build GRCh38
rs943018851 Chr.X: 73563404-73563404 on Build GRCh38
rs777824683 Chr.X: 73563540-73563540 on Build GRCh38
rs956972124 Chr.X: 73563268-73563268 on Build GRCh38
rs760335915 Chr.X: 73563441-73563458 on Build GRCh38
rs765509610 Chr.X: 73563393-73563393 on Build GRCh38
rs760023223 Chr.X: 73563461-73563462 on Build GRCh38
rs774809877 Chr.X: 73563132-73563132 on Build GRCh38
rs773550444 Chr.X: 73563335-73563337 on Build GRCh38
rs770923373 Chr.X: 73563545-73563545 on Build GRCh38
rs866971272 Chr.X: 73563381-73563381 on Build GRCh38
rs772987376 Chr.X: 73563345-73563345 on Build GRCh38
rs776130498 Chr.X: 73563465-73563476 on Build GRCh38
rs762253564 Chr.X: 73563347-73563347 on Build GRCh38
rs951356930 Chr.X: 73563143-73563143 on Build GRCh38
rs748955512 Chr.X: 73563247-73563247 on Build GRCh38
rs771785330 Chr.X: 73563131-73563131 on Build GRCh38
rs750259989 Chr.X: 73563435-73563437 on Build GRCh38
rs896458368 Chr.X: 73563390-73563390 on Build GRCh38
rs754463259 Chr.X: 73563262-73563262 on Build GRCh38
rs778709635 Chr.X: 73563306-73563306 on Build GRCh38
rs760149927 Chr.X: 73563379-73563381 on Build GRCh38
rs748176889 Chr.X: 73563444-73563446 on Build GRCh38
rs1022283457 Chr.X: 73563335-73563340 on Build GRCh38
rs760241560 Chr.X: 73563133-73563133 on Build GRCh38
rs998769511 Chr.X: 73563236-73563236 on Build GRCh38
rs753308044 Chr.X: 73563480-73563480 on Build GRCh38
rs752000885 Chr.X: 73563448-73563448 on Build GRCh38
rs982706133 Chr.X: 73563173-73563173 on Build GRCh38
rs1052640498 Chr.X: 73563352-73563352 on Build GRCh38
rs757706818 Chr.X: 73563469-73563469 on Build GRCh38
rs932126867 Chr.X: 73563275-73563275 on Build GRCh38
rs912206392 Chr.X: 73563197-73563197 on Build GRCh38
rs200325370 Chr.X: 73563433-73563433 on Build GRCh38
rs142151591 Chr.X: 73563464-73563464 on Build GRCh38
rs748012410 Chr.X: 73563505-73563505 on Build GRCh38
rs1043931574 Chr.X: 73563320-73563320 on Build GRCh38
rs775973130 Chr.X: 73563407-73563407 on Build GRCh38
rs772092016 Chr.X: 73563329-73563329 on Build GRCh38
rs768233042 Chr.X: 73563256-73563256 on Build GRCh38
rs759841020 Chr.X: 73563388-73563388 on Build GRCh38
rs759148357 Chr.X: 73563125-73563125 on Build GRCh38
rs764628296 Chr.X: 73563133-73563133 on Build GRCh38
rs763645313 Chr.X: 73563379-73563387 on Build GRCh38
rs1027298433 Chr.X: 73563095-73563095 on Build GRCh38
rs762577341 Chr.X: 73563379-73563384 on Build GRCh38
rs1055194208 Chr.X: 73563364-73563378 on Build GRCh38
rs777226608 Chr.X: 73563364-73563384 on Build GRCh38
rs750642868 Chr.X: 73563462-73563467 on Build GRCh38
rs764069325 Chr.X: 73563367-73563378 on Build GRCh38
rs780098080 Chr.X: 73563440-73563441 on Build GRCh38
rs1049000602 Chr.X: 73563384-73563384 on Build GRCh38
rs760875879 Chr.X: 73563355-73563355 on Build GRCh38
rs780998301 Chr.X: 73563432-73563458 on Build GRCh38
rs1040006463 Chr.X: 73563443-73563443 on Build GRCh38
rs746955427 Chr.X: 73563542-73563542 on Build GRCh38
rs776792696 Chr.X: 73563380-73563380 on Build GRCh38
rs921056177 Chr.X: 73563274-73563274 on Build GRCh38
rs747259732 Chr.X: 73563456-73563458 on Build GRCh38
rs756985409 Chr.X: 73563425-73563426 on Build GRCh38
rs777803740 Chr.X: 73563450-73563461 on Build GRCh38
rs770188430 Chr.X: 73563462-73563464 on Build GRCh38
rs763197820 Chr.X: 73563417-73563417 on Build GRCh38
rs771121335 Chr.X: 73563374-73563374 on Build GRCh38
rs780347596 Chr.X: 73563490-73563490 on Build GRCh38
rs1011097830 Chr.X: 73563328-73563328 on Build GRCh38

View Full Product Details