Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 73683210-73683720 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHIC1
Entrez Gene ID
53344
Gene Name
cysteine rich hydrophobic domain 1
Gene Aliases
BRX
UniGene
Hs.496323
Gene Chromosome Location
Chr.X: 73563045-73687109 on Build GRCh38
Transcripts
NM_001039840.2 NM_001300884.1 XM_017029583.1

SNP Detail

SNP ID Chromosome Location
rs982682107 Chr.X: 73683576-73683576 on Build GRCh38
rs1019461255 Chr.X: 73683591-73683591 on Build GRCh38
rs965017827 Chr.X: 73683604-73683604 on Build GRCh38
rs761908598 Chr.X: 73683335-73683335 on Build GRCh38
rs112987259 Chr.X: 73683338-73683338 on Build GRCh38
rs1047290988 Chr.X: 73683595-73683595 on Build GRCh38
rs1021319443 Chr.X: 73683336-73683336 on Build GRCh38
rs776656468 Chr.X: 73683284-73683284 on Build GRCh38
rs140720032 Chr.X: 73683411-73683411 on Build GRCh38
rs995595424 Chr.X: 73683469-73683469 on Build GRCh38
rs935594643 Chr.X: 73683290-73683290 on Build GRCh38
rs991761017 Chr.X: 73683536-73683536 on Build GRCh38
rs926339926 Chr.X: 73683450-73683450 on Build GRCh38
rs979706981 Chr.X: 73683424-73683424 on Build GRCh38
rs41303727 Chr.X: 73683679-73683679 on Build GRCh38
rs766284032 Chr.X: 73683349-73683349 on Build GRCh38
rs368014112 Chr.X: 73683599-73683599 on Build GRCh38
rs937763977 Chr.X: 73683475-73683475 on Build GRCh38
rs150099393 Chr.X: 73683530-73683530 on Build GRCh38
rs941532366 Chr.X: 73683639-73683639 on Build GRCh38
rs1044319385 Chr.X: 73683262-73683262 on Build GRCh38
rs947155056 Chr.X: 73683234-73683234 on Build GRCh38
rs755762391 Chr.X: 73683636-73683636 on Build GRCh38
rs184748977 Chr.X: 73683397-73683397 on Build GRCh38
rs1052666732 Chr.X: 73683298-73683298 on Build GRCh38
rs917593863 Chr.X: 73683554-73683554 on Build GRCh38
rs572987330 Chr.X: 73683532-73683532 on Build GRCh38
rs1029674243 Chr.X: 73683315-73683315 on Build GRCh38
rs904009701 Chr.X: 73683263-73683263 on Build GRCh38
rs896504562 Chr.X: 73683329-73683329 on Build GRCh38
rs1027260864 Chr.X: 73683509-73683509 on Build GRCh38
rs996818459 Chr.X: 73683283-73683283 on Build GRCh38

View Full Product Details