Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 73683504-73684025 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHIC1
Entrez Gene ID
53344
Gene Name
cysteine rich hydrophobic domain 1
Gene Aliases
BRX
UniGene
Hs.496323
Gene Chromosome Location
Chr.X: 73563045-73687109 on Build GRCh38
Transcripts
NM_001039840.2 NM_001300884.1 XM_017029583.1

SNP Detail

SNP ID Chromosome Location
rs920932692 Chr.X: 73683749-73683749 on Build GRCh38
rs982682107 Chr.X: 73683576-73683576 on Build GRCh38
rs1019461255 Chr.X: 73683591-73683591 on Build GRCh38
rs965017827 Chr.X: 73683604-73683604 on Build GRCh38
rs745369292 Chr.X: 73683830-73683830 on Build GRCh38
rs1047290988 Chr.X: 73683595-73683595 on Build GRCh38
rs968376711 Chr.X: 73683968-73683968 on Build GRCh38
rs949612468 Chr.X: 73683903-73683903 on Build GRCh38
rs753375725 Chr.X: 73683703-73683703 on Build GRCh38
rs1033491596 Chr.X: 73683994-73683994 on Build GRCh38
rs991761017 Chr.X: 73683536-73683536 on Build GRCh38
rs772645131 Chr.X: 73683850-73683851 on Build GRCh38
rs41303727 Chr.X: 73683679-73683679 on Build GRCh38
rs780873324 Chr.X: 73683765-73683765 on Build GRCh38
rs528769512 Chr.X: 73683933-73683933 on Build GRCh38
rs368014112 Chr.X: 73683599-73683599 on Build GRCh38
rs996019368 Chr.X: 73683975-73683975 on Build GRCh38
rs780212286 Chr.X: 73683885-73683885 on Build GRCh38
rs979248828 Chr.X: 73683983-73683983 on Build GRCh38
rs905179448 Chr.X: 73683961-73683961 on Build GRCh38
rs150099393 Chr.X: 73683530-73683530 on Build GRCh38
rs899881198 Chr.X: 73683698-73683698 on Build GRCh38
rs941532366 Chr.X: 73683639-73683639 on Build GRCh38
rs978518909 Chr.X: 73683766-73683766 on Build GRCh38
rs769417302 Chr.X: 73683894-73683894 on Build GRCh38
rs755762391 Chr.X: 73683636-73683636 on Build GRCh38
rs202243058 Chr.X: 73683763-73683764 on Build GRCh38
rs540856639 Chr.X: 73683752-73683752 on Build GRCh38
rs747401611 Chr.X: 73683897-73683897 on Build GRCh38
rs896697924 Chr.X: 73683900-73683900 on Build GRCh38
rs746417230 Chr.X: 73683807-73683807 on Build GRCh38
rs917593863 Chr.X: 73683554-73683554 on Build GRCh38
rs572987330 Chr.X: 73683532-73683532 on Build GRCh38
rs1045505092 Chr.X: 73683941-73683941 on Build GRCh38
rs925463814 Chr.X: 73683795-73683795 on Build GRCh38

View Full Product Details