Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 77505570-77506116 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATRX
Entrez Gene ID
546
Gene Name
ATRX, chromatin remodeler
Gene Aliases
ATR2, JMS, MRX52, MRXHF1, RAD54, RAD54L, SFM1, SHS, XH2, XNP, ZNF-HX
UniGene
Hs.533526
Gene Chromosome Location
Chr.X: 77504878-77786269 on Build GRCh38
Transcripts
NM_000489.4 NM_138270.3 XM_005262153.4 XM_005262154.4 XM_005262155.4 XM_005262156.4 XM_005262157.4 XM_006724666.3 XM_006724667.3 XM_017029601.1 XM_017029602.1 XM_017029603.1 XM_017029604.1 XM_017029605.1 XM_017029606.1 XM_017029607.1 XM_017029608.1 XM_017029609.1 XM_017029610.1 XM_017029611.1

SNP Detail

SNP ID Chromosome Location
rs960539705 Chr.X: 77505649-77505649 on Build GRCh38
rs781957872 Chr.X: 77505828-77505828 on Build GRCh38
rs3135467 Chr.X: 77506044-77506044 on Build GRCh38
rs1035062301 Chr.X: 77505611-77505611 on Build GRCh38
rs569397881 Chr.X: 77505702-77505702 on Build GRCh38
rs782373142 Chr.X: 77505708-77505708 on Build GRCh38
rs916505770 Chr.X: 77505676-77505676 on Build GRCh38
rs962641767 Chr.X: 77505707-77505707 on Build GRCh38
rs990642097 Chr.X: 77505675-77505675 on Build GRCh38
rs782100927 Chr.X: 77505896-77505896 on Build GRCh38
rs782392612 Chr.X: 77505599-77505599 on Build GRCh38

View Full Product Details