Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 78272931-78273435 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYSLTR1
Entrez Gene ID
10800
Gene Name
cysteinyl leukotriene receptor 1
Gene Aliases
CYSLT1, CYSLT1R, CYSLTR, HMTMF81
UniGene
Hs.733809
Gene Chromosome Location
Chr.X: 78271468-78327691 on Build GRCh38
Transcripts
NM_001282186.1 NM_001282187.1 NM_001282188.1 NM_006639.3

SNP Detail

SNP ID Chromosome Location
rs762344427 Chr.X: 78273152-78273152 on Build GRCh38
rs781123232 Chr.X: 78273197-78273197 on Build GRCh38
rs1027420210 Chr.X: 78273269-78273269 on Build GRCh38
rs144735906 Chr.X: 78273123-78273123 on Build GRCh38
rs144279487 Chr.X: 78273270-78273270 on Build GRCh38
rs760527258 Chr.X: 78273021-78273021 on Build GRCh38
rs777215159 Chr.X: 78273257-78273257 on Build GRCh38
rs35745545 Chr.X: 78273096-78273096 on Build GRCh38
rs989101160 Chr.X: 78273029-78273029 on Build GRCh38
rs773734601 Chr.X: 78273386-78273386 on Build GRCh38
rs139639671 Chr.X: 78273379-78273379 on Build GRCh38
rs770115123 Chr.X: 78273111-78273111 on Build GRCh38
rs201344297 Chr.X: 78273002-78273002 on Build GRCh38
rs34170395 Chr.X: 78273062-78273062 on Build GRCh38
rs746912468 Chr.X: 78273185-78273185 on Build GRCh38
rs147386972 Chr.X: 78273272-78273272 on Build GRCh38
rs201233807 Chr.X: 78273351-78273351 on Build GRCh38
rs201536627 Chr.X: 78273393-78273393 on Build GRCh38
rs201447425 Chr.X: 78273014-78273014 on Build GRCh38
rs369005693 Chr.X: 78273053-78273053 on Build GRCh38
rs985624483 Chr.X: 78273004-78273004 on Build GRCh38
rs1027101532 Chr.X: 78272978-78272978 on Build GRCh38
rs761039907 Chr.X: 78272985-78272985 on Build GRCh38
rs372597227 Chr.X: 78273068-78273068 on Build GRCh38
rs752872123 Chr.X: 78273035-78273035 on Build GRCh38
rs770880534 Chr.X: 78273092-78273092 on Build GRCh38
rs768749198 Chr.X: 78273254-78273254 on Build GRCh38
rs752093115 Chr.X: 78273175-78273175 on Build GRCh38
rs760911761 Chr.X: 78273390-78273390 on Build GRCh38
rs767842487 Chr.X: 78273154-78273154 on Build GRCh38
rs746755262 Chr.X: 78273090-78273090 on Build GRCh38
rs1043476808 Chr.X: 78273116-78273116 on Build GRCh38
rs199546618 Chr.X: 78273008-78273008 on Build GRCh38
rs757257673 Chr.X: 78273320-78273320 on Build GRCh38
rs747484187 Chr.X: 78273241-78273241 on Build GRCh38
rs954835366 Chr.X: 78273003-78273003 on Build GRCh38
rs866204187 Chr.X: 78273303-78273303 on Build GRCh38
rs748746989 Chr.X: 78272972-78272972 on Build GRCh38
rs762901050 Chr.X: 78273222-78273222 on Build GRCh38
rs202230050 Chr.X: 78272960-78272960 on Build GRCh38
rs201161779 Chr.X: 78273156-78273156 on Build GRCh38
rs28365142 Chr.X: 78273130-78273130 on Build GRCh38
rs200181664 Chr.X: 78273038-78273038 on Build GRCh38
rs751174492 Chr.X: 78273318-78273318 on Build GRCh38
rs528622685 Chr.X: 78272956-78272956 on Build GRCh38
rs954337414 Chr.X: 78272987-78272987 on Build GRCh38
rs768814904 Chr.X: 78273382-78273382 on Build GRCh38
rs774459855 Chr.X: 78273259-78273259 on Build GRCh38
rs199638285 Chr.X: 78273018-78273018 on Build GRCh38
rs186267921 Chr.X: 78273327-78273327 on Build GRCh38
rs143852533 Chr.X: 78273183-78273183 on Build GRCh38
rs768181799 Chr.X: 78272979-78272979 on Build GRCh38
rs759997255 Chr.X: 78273204-78273207 on Build GRCh38
rs764665474 Chr.X: 78273044-78273044 on Build GRCh38
rs773948845 Chr.X: 78272989-78272989 on Build GRCh38
rs374673923 Chr.X: 78273040-78273040 on Build GRCh38
rs201634768 Chr.X: 78273030-78273030 on Build GRCh38
rs200173665 Chr.X: 78273292-78273292 on Build GRCh38
rs764778799 Chr.X: 78273216-78273217 on Build GRCh38
rs765378217 Chr.X: 78273033-78273033 on Build GRCh38
rs200624361 Chr.X: 78273056-78273056 on Build GRCh38
rs763639239 Chr.X: 78273293-78273293 on Build GRCh38
rs945051585 Chr.X: 78273072-78273072 on Build GRCh38
rs770423432 Chr.X: 78273359-78273359 on Build GRCh38
rs747471147 Chr.X: 78272958-78272958 on Build GRCh38
rs779693746 Chr.X: 78272966-78272966 on Build GRCh38
rs919071650 Chr.X: 78273055-78273055 on Build GRCh38
rs752138528 Chr.X: 78273150-78273150 on Build GRCh38
rs755695510 Chr.X: 78272962-78272962 on Build GRCh38
rs138981290 Chr.X: 78273057-78273057 on Build GRCh38
rs200446566 Chr.X: 78272996-78272996 on Build GRCh38
rs201022495 Chr.X: 78273084-78273084 on Build GRCh38
rs757639521 Chr.X: 78273054-78273054 on Build GRCh38
rs779328785 Chr.X: 78273073-78273073 on Build GRCh38
rs181815373 Chr.X: 78273191-78273191 on Build GRCh38
rs745607492 Chr.X: 78273347-78273347 on Build GRCh38
rs777225112 Chr.X: 78273089-78273090 on Build GRCh38
rs200236484 Chr.X: 78273344-78273344 on Build GRCh38
rs200785164 Chr.X: 78273028-78273028 on Build GRCh38
rs199931252 Chr.X: 78273058-78273058 on Build GRCh38
rs201954015 Chr.X: 78273167-78273167 on Build GRCh38
rs140302261 Chr.X: 78273220-78273220 on Build GRCh38
rs965569427 Chr.X: 78273354-78273354 on Build GRCh38
rs768075700 Chr.X: 78273266-78273266 on Build GRCh38
rs759811173 Chr.X: 78273105-78273105 on Build GRCh38
rs893499511 Chr.X: 78273194-78273194 on Build GRCh38
rs148732852 Chr.X: 78273013-78273013 on Build GRCh38
rs749500035 Chr.X: 78273381-78273381 on Build GRCh38
rs746676205 Chr.X: 78273267-78273267 on Build GRCh38
rs776609177 Chr.X: 78273103-78273103 on Build GRCh38
rs759648705 Chr.X: 78273031-78273031 on Build GRCh38
rs745726108 Chr.X: 78273201-78273201 on Build GRCh38
rs202227631 Chr.X: 78273087-78273087 on Build GRCh38
rs200459301 Chr.X: 78273385-78273385 on Build GRCh38
rs897151198 Chr.X: 78273257-78273257 on Build GRCh38
rs775364230 Chr.X: 78273112-78273112 on Build GRCh38
rs774406808 Chr.X: 78273384-78273384 on Build GRCh38
rs766746913 Chr.X: 78273075-78273076 on Build GRCh38
rs200008187 Chr.X: 78273145-78273145 on Build GRCh38
rs1015501712 Chr.X: 78273226-78273226 on Build GRCh38
rs776909263 Chr.X: 78273397-78273397 on Build GRCh38
rs747762878 Chr.X: 78273077-78273077 on Build GRCh38
rs762654756 Chr.X: 78273410-78273410 on Build GRCh38

View Full Product Details