Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 80671633-80672149 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BRWD3
Entrez Gene ID
254065
Gene Name
bromodomain and WD repeat domain containing 3
Gene Aliases
BRODL, MRX93
UniGene
Hs.147027
Gene Chromosome Location
Chr.X: 80669488-80809736 on Build GRCh38
Transcripts
NM_153252.4 XM_005262113.3

SNP Detail

SNP ID Chromosome Location
rs767676792 Chr.X: 80671691-80671691 on Build GRCh38
rs962196336 Chr.X: 80671737-80671737 on Build GRCh38
rs942043521 Chr.X: 80671736-80671736 on Build GRCh38
rs999310349 Chr.X: 80672037-80672037 on Build GRCh38
rs41311569 Chr.X: 80671800-80671800 on Build GRCh38
rs1006371756 Chr.X: 80671676-80671676 on Build GRCh38
rs1030671459 Chr.X: 80672075-80672075 on Build GRCh38
rs888054548 Chr.X: 80671670-80671670 on Build GRCh38
rs996724905 Chr.X: 80671903-80671903 on Build GRCh38
rs992168819 Chr.X: 80671797-80671798 on Build GRCh38
rs890194850 Chr.X: 80672035-80672035 on Build GRCh38
rs750520733 Chr.X: 80671750-80671750 on Build GRCh38
rs913036163 Chr.X: 80672044-80672045 on Build GRCh38
rs990097317 Chr.X: 80672001-80672001 on Build GRCh38
rs1057515996 Chr.X: 80672028-80672028 on Build GRCh38
rs900766422 Chr.X: 80671829-80671829 on Build GRCh38
rs935895595 Chr.X: 80671939-80671939 on Build GRCh38
rs553501091 Chr.X: 80672107-80672107 on Build GRCh38
rs981210783 Chr.X: 80671849-80671849 on Build GRCh38
rs886477527 Chr.X: 80671726-80671726 on Build GRCh38
rs778718534 Chr.X: 80671933-80671933 on Build GRCh38
rs969353813 Chr.X: 80671841-80671841 on Build GRCh38
rs1030527377 Chr.X: 80671999-80671999 on Build GRCh38
rs957902881 Chr.X: 80672077-80672077 on Build GRCh38
rs761052536 Chr.X: 80671943-80671943 on Build GRCh38
rs1047708947 Chr.X: 80671698-80671698 on Build GRCh38

View Full Product Details