Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 80672947-80673493 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BRWD3
Entrez Gene ID
254065
Gene Name
bromodomain and WD repeat domain containing 3
Gene Aliases
BRODL, MRX93
UniGene
Hs.147027
Gene Chromosome Location
Chr.X: 80669488-80809736 on Build GRCh38
Transcripts
NM_153252.4 XM_005262113.3

SNP Detail

SNP ID Chromosome Location
rs1032431930 Chr.X: 80673137-80673137 on Build GRCh38
rs1017176551 Chr.X: 80673398-80673398 on Build GRCh38
rs912125921 Chr.X: 80673190-80673190 on Build GRCh38
rs1056078595 Chr.X: 80673309-80673309 on Build GRCh38
rs749020327 Chr.X: 80673453-80673453 on Build GRCh38
rs946458343 Chr.X: 80673234-80673234 on Build GRCh38
rs938995022 Chr.X: 80673289-80673289 on Build GRCh38
rs949609783 Chr.X: 80673372-80673372 on Build GRCh38
rs1034870540 Chr.X: 80672995-80672995 on Build GRCh38
rs953325451 Chr.X: 80673163-80673163 on Build GRCh38
rs767696195 Chr.X: 80673162-80673162 on Build GRCh38
rs951316524 Chr.X: 80673011-80673011 on Build GRCh38
rs958278693 Chr.X: 80673145-80673145 on Build GRCh38
rs923071958 Chr.X: 80673263-80673263 on Build GRCh38
rs879201626 Chr.X: 80673015-80673015 on Build GRCh38
rs1016795681 Chr.X: 80673086-80673086 on Build GRCh38
rs762108287 Chr.X: 80673147-80673147 on Build GRCh38
rs369334946 Chr.X: 80673231-80673231 on Build GRCh38
rs941680050 Chr.X: 80673297-80673297 on Build GRCh38
rs918942793 Chr.X: 80673319-80673319 on Build GRCh38
rs756159408 Chr.X: 80673296-80673296 on Build GRCh38
rs764587615 Chr.X: 80673077-80673077 on Build GRCh38
rs765169545 Chr.X: 80673316-80673316 on Build GRCh38
rs1044784684 Chr.X: 80673269-80673269 on Build GRCh38
rs781124974 Chr.X: 80673122-80673122 on Build GRCh38
rs750446858 Chr.X: 80673209-80673209 on Build GRCh38
rs987483264 Chr.X: 80673173-80673173 on Build GRCh38
rs1046641346 Chr.X: 80673441-80673441 on Build GRCh38
rs116559809 Chr.X: 80673126-80673126 on Build GRCh38
rs967310623 Chr.X: 80673198-80673198 on Build GRCh38
rs904962342 Chr.X: 80673287-80673287 on Build GRCh38
rs935594585 Chr.X: 80673207-80673207 on Build GRCh38
rs769363805 Chr.X: 80673232-80673232 on Build GRCh38
rs898289689 Chr.X: 80673408-80673408 on Build GRCh38
rs887012917 Chr.X: 80673312-80673312 on Build GRCh38
rs963786326 Chr.X: 80673123-80673123 on Build GRCh38

View Full Product Details