Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 93711432-93711979 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM133A
Entrez Gene ID
286499
Gene Name
family with sequence similarity 133 member A
Gene Aliases
CT115
UniGene
Hs.110069
Gene Chromosome Location
Chr.X: 93673730-93712274 on Build GRCh38
Transcripts
NM_001171109.1 NM_001171110.1 NM_001171111.1 NM_173698.2 XM_006724641.3 XM_011530925.2 XM_011530926.2 XM_011530927.2 XM_011530928.1 XM_011530929.1 XM_011530931.2 XM_017029454.1 XM_017029455.1 XM_017029456.1

SNP Detail

SNP ID Chromosome Location
rs751455534 Chr.X: 93711826-93711826 on Build GRCh38
rs772429397 Chr.X: 93711519-93711519 on Build GRCh38
rs1026050532 Chr.X: 93711766-93711766 on Build GRCh38
rs1036026816 Chr.X: 93711463-93711463 on Build GRCh38
rs977525269 Chr.X: 93711504-93711504 on Build GRCh38
rs1005882467 Chr.X: 93711893-93711893 on Build GRCh38
rs185504839 Chr.X: 93711518-93711518 on Build GRCh38
rs938371168 Chr.X: 93711910-93711910 on Build GRCh38
rs984730925 Chr.X: 93711830-93711830 on Build GRCh38
rs764844368 Chr.X: 93711578-93711578 on Build GRCh38
rs904643663 Chr.X: 93711597-93711597 on Build GRCh38
rs998929138 Chr.X: 93711600-93711600 on Build GRCh38
rs965383611 Chr.X: 93711665-93711665 on Build GRCh38
rs12012248 Chr.X: 93711639-93711639 on Build GRCh38
rs943310752 Chr.X: 93711646-93711646 on Build GRCh38
rs936019758 Chr.X: 93711545-93711545 on Build GRCh38
rs1056992599 Chr.X: 93711531-93711531 on Build GRCh38
rs763597106 Chr.X: 93711787-93711787 on Build GRCh38
rs959183809 Chr.X: 93711916-93711916 on Build GRCh38
rs998162278 Chr.X: 93711706-93711706 on Build GRCh38
rs963482023 Chr.X: 93711856-93711856 on Build GRCh38
rs901696948 Chr.X: 93711737-93711737 on Build GRCh38
rs918905278 Chr.X: 93711498-93711498 on Build GRCh38
rs41435748 Chr.X: 93711952-93711952 on Build GRCh38
rs971774777 Chr.X: 93711887-93711887 on Build GRCh38
rs762444977 Chr.X: 93711699-93711699 on Build GRCh38
rs1028581480 Chr.X: 93711761-93711761 on Build GRCh38
rs372189526 Chr.X: 93711574-93711574 on Build GRCh38
rs918980457 Chr.X: 93711901-93711901 on Build GRCh38
rs888993609 Chr.X: 93711643-93711643 on Build GRCh38
rs993636290 Chr.X: 93711747-93711747 on Build GRCh38
rs1013169798 Chr.X: 93711571-93711571 on Build GRCh38
rs188929220 Chr.X: 93711568-93711568 on Build GRCh38
rs910049306 Chr.X: 93711853-93711853 on Build GRCh38

View Full Product Details