Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 101224516-101225028 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DRP2
Entrez Gene ID
1821
Gene Name
dystrophin related protein 2
Gene Aliases
DRP-2
UniGene
Hs.159291
Gene Chromosome Location
Chr.X: 101219786-101264497 on Build GRCh38
Transcripts
NM_001171184.1 NM_001939.2 XM_017029333.1 XM_017029334.1

SNP Detail

SNP ID Chromosome Location
rs1052948103 Chr.X: 101224911-101224911 on Build GRCh38
rs777510274 Chr.X: 101224568-101224568 on Build GRCh38
rs755330749 Chr.X: 101224728-101224728 on Build GRCh38
rs867883277 Chr.X: 101224735-101224735 on Build GRCh38
rs180739170 Chr.X: 101224909-101224909 on Build GRCh38
rs149049298 Chr.X: 101224787-101224787 on Build GRCh38
rs757062474 Chr.X: 101224936-101224936 on Build GRCh38
rs998168336 Chr.X: 101224907-101224907 on Build GRCh38
rs774346284 Chr.X: 101224833-101224833 on Build GRCh38
rs755558116 Chr.X: 101224555-101224555 on Build GRCh38
rs73555045 Chr.X: 101224999-101224999 on Build GRCh38
rs923489392 Chr.X: 101224617-101224617 on Build GRCh38
rs946223499 Chr.X: 101224635-101224635 on Build GRCh38
rs1039760539 Chr.X: 101224843-101224843 on Build GRCh38
rs188712825 Chr.X: 101224890-101224890 on Build GRCh38
rs779378039 Chr.X: 101224744-101224744 on Build GRCh38
rs1057507314 Chr.X: 101224616-101224616 on Build GRCh38
rs186067868 Chr.X: 101224969-101224969 on Build GRCh38
rs201254498 Chr.X: 101224730-101224735 on Build GRCh38
rs1040587265 Chr.X: 101224601-101224601 on Build GRCh38
rs1050410378 Chr.X: 101224829-101224829 on Build GRCh38
rs917475994 Chr.X: 101224627-101224627 on Build GRCh38
rs940835601 Chr.X: 101224581-101224581 on Build GRCh38
rs1030012561 Chr.X: 101224913-101224913 on Build GRCh38
rs903860575 Chr.X: 101224841-101224841 on Build GRCh38
rs772172309 Chr.X: 101224926-101224926 on Build GRCh38

View Full Product Details