Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 101403734-101404191 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLA
Entrez Gene ID
2717
Gene Name
galactosidase alpha
Gene Aliases
GALA
UniGene
Hs.69089
Gene Chromosome Location
Chr.X: 101397791-101408013 on Build GRCh38
Transcripts
NM_000169.2 NM_001199973.1 NM_001199974.1

Gene Symbol
RPL36A-HNRNPH2
Entrez Gene ID
100529097
Gene Name
RPL36A-HNRNPH2 readthrough
Gene Aliases
-
UniGene
Hs.432485
Gene Chromosome Location
Chr.X: 101390890-101414140 on Build GRCh38
Transcripts
NM_000169.2 NM_001199973.1 NM_001199974.1

SNP Detail

SNP ID Chromosome Location
rs869312141 Chr.X: 101403908-101403908 on Build GRCh38
rs782507007 Chr.X: 101404008-101404008 on Build GRCh38
rs782455398 Chr.X: 101404026-101404026 on Build GRCh38
rs869312271 Chr.X: 101403898-101403898 on Build GRCh38
rs869312273 Chr.X: 101403881-101403881 on Build GRCh38
rs1057516429 Chr.X: 101403936-101403936 on Build GRCh38
rs869312252 Chr.X: 101403861-101403861 on Build GRCh38
rs869312279 Chr.X: 101403816-101403816 on Build GRCh38
rs104894834 Chr.X: 101403846-101403846 on Build GRCh38
rs782260908 Chr.X: 101403835-101403835 on Build GRCh38
rs372966991 Chr.X: 101403845-101403845 on Build GRCh38
rs869312276 Chr.X: 101403853-101403863 on Build GRCh38
rs12392549 Chr.X: 101403830-101403830 on Build GRCh38
rs781899607 Chr.X: 101403783-101403783 on Build GRCh38
rs869312142 Chr.X: 101403843-101403843 on Build GRCh38
rs947894100 Chr.X: 101404115-101404115 on Build GRCh38
rs782696004 Chr.X: 101403769-101403769 on Build GRCh38
rs782092398 Chr.X: 101403867-101403867 on Build GRCh38
rs909651693 Chr.X: 101404091-101404091 on Build GRCh38
rs986730205 Chr.X: 101403921-101403921 on Build GRCh38
rs886044900 Chr.X: 101403895-101403895 on Build GRCh38
rs398123208 Chr.X: 101403938-101403938 on Build GRCh38
rs918257809 Chr.X: 101404059-101404059 on Build GRCh38
rs781838005 Chr.X: 101403941-101403941 on Build GRCh38
rs869312275 Chr.X: 101403868-101403868 on Build GRCh38
rs797044670 Chr.X: 101403816-101403819 on Build GRCh38
rs886041315 Chr.X: 101403906-101403906 on Build GRCh38
rs567771967 Chr.X: 101404165-101404165 on Build GRCh38
rs113173389 Chr.X: 101403899-101403899 on Build GRCh38
rs797044669 Chr.X: 101403810-101403810 on Build GRCh38
rs398123209 Chr.X: 101403806-101403806 on Build GRCh38
rs782471208 Chr.X: 101403760-101403760 on Build GRCh38
rs782621192 Chr.X: 101403989-101403990 on Build GRCh38
rs104894833 Chr.X: 101403984-101403984 on Build GRCh38
rs730880443 Chr.X: 101403945-101403945 on Build GRCh38
rs782738098 Chr.X: 101404013-101404013 on Build GRCh38
rs869312277 Chr.X: 101403855-101403855 on Build GRCh38
rs398123206 Chr.X: 101403986-101403986 on Build GRCh38
rs782722577 Chr.X: 101403933-101403933 on Build GRCh38
rs188819250 Chr.X: 101404150-101404150 on Build GRCh38
rs869312268 Chr.X: 101403947-101403947 on Build GRCh38
rs387906483 Chr.X: 101403809-101403809 on Build GRCh38
rs782644591 Chr.X: 101403794-101403794 on Build GRCh38
rs782803696 Chr.X: 101403985-101403985 on Build GRCh38
rs782580023 Chr.X: 101404137-101404137 on Build GRCh38
rs74795363 Chr.X: 101403844-101403844 on Build GRCh38
rs886039140 Chr.X: 101403967-101403967 on Build GRCh38
rs869312267 Chr.X: 101403955-101403955 on Build GRCh38
rs782600529 Chr.X: 101403807-101403807 on Build GRCh38
rs929665863 Chr.X: 101404071-101404071 on Build GRCh38
rs869312264 Chr.X: 101403983-101403983 on Build GRCh38
rs782488441 Chr.X: 101403796-101403796 on Build GRCh38
rs782538843 Chr.X: 101404167-101404167 on Build GRCh38
rs953560993 Chr.X: 101403883-101403883 on Build GRCh38
rs781822743 Chr.X: 101404015-101404015 on Build GRCh38
rs375749402 Chr.X: 101403870-101403870 on Build GRCh38
rs869312270 Chr.X: 101403903-101403903 on Build GRCh38
rs376723747 Chr.X: 101404028-101404028 on Build GRCh38
rs143768851 Chr.X: 101403827-101403827 on Build GRCh38
rs148158093 Chr.X: 101403828-101403828 on Build GRCh38
rs782197638 Chr.X: 101403819-101403819 on Build GRCh38
rs782523891 Chr.X: 101403788-101403788 on Build GRCh38
rs869312140 Chr.X: 101403924-101403924 on Build GRCh38
rs869312269 Chr.X: 101403920-101403920 on Build GRCh38
rs869312266 Chr.X: 101403958-101403958 on Build GRCh38
rs727505060 Chr.X: 101403860-101403860 on Build GRCh38
rs983878520 Chr.X: 101404087-101404087 on Build GRCh38
rs781967285 Chr.X: 101403850-101403850 on Build GRCh38
rs886039136 Chr.X: 101403884-101403885 on Build GRCh38
rs869312274 Chr.X: 101403869-101403869 on Build GRCh38
rs869312272 Chr.X: 101403882-101403882 on Build GRCh38
rs886044767 Chr.X: 101403912-101403912 on Build GRCh38
rs781927744 Chr.X: 101403968-101403968 on Build GRCh38
rs781865046 Chr.X: 101403996-101403996 on Build GRCh38
rs113993944 Chr.X: 101403893-101403894 on Build GRCh38
rs869312282 Chr.X: 101403811-101403811 on Build GRCh38

View Full Product Details