Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 15481048-15481484 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CELA2B
Entrez Gene ID
51032
Gene Name
chymotrypsin like elastase family member 2B
Gene Aliases
ELA2B
UniGene
Hs.631871
Gene Chromosome Location
Chr.1: 15476101-15491400 on Build GRCh38
Transcripts
NM_015849.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4944526 c.134C>T p.S45F Chr.1: 15481150-15481150 on Build GRCh38
4944525 c.182C>T p.S61F Chr.1: 15481150-15481150 on Build GRCh38
6380341 c.151A>T p.N51Y Chr.1: 15481119-15481119 on Build GRCh38
4750340 c.136C>A p.L46M Chr.1: 15481152-15481152 on Build GRCh38
4750339 c.184C>A p.L62M Chr.1: 15481152-15481152 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs965176999 Chr.1: 15481367-15481367 on Build GRCh38
rs200158570 Chr.1: 15481125-15481125 on Build GRCh38
rs760471053 Chr.1: 15481236-15481236 on Build GRCh38
rs1046229699 Chr.1: 15481449-15481449 on Build GRCh38
rs556757250 Chr.1: 15481425-15481425 on Build GRCh38
rs202005938 Chr.1: 15481178-15481178 on Build GRCh38
rs149140983 Chr.1: 15481123-15481123 on Build GRCh38
rs770038491 Chr.1: 15481099-15481099 on Build GRCh38
rs989104970 Chr.1: 15481415-15481415 on Build GRCh38
rs764899818 Chr.1: 15481202-15481202 on Build GRCh38
rs756652164 Chr.1: 15481219-15481219 on Build GRCh38
rs377665483 Chr.1: 15481226-15481226 on Build GRCh38
rs975541215 Chr.1: 15481379-15481379 on Build GRCh38
rs1026588982 Chr.1: 15481340-15481340 on Build GRCh38
rs542066335 Chr.1: 15481222-15481222 on Build GRCh38
rs749696510 Chr.1: 15481225-15481225 on Build GRCh38
rs753419723 Chr.1: 15481074-15481074 on Build GRCh38
rs185343477 Chr.1: 15481366-15481366 on Build GRCh38
rs779296747 Chr.1: 15481362-15481362 on Build GRCh38
rs777397198 Chr.1: 15481215-15481215 on Build GRCh38
rs945270245 Chr.1: 15481157-15481157 on Build GRCh38
rs781517319 Chr.1: 15481088-15481088 on Build GRCh38
rs202130301 Chr.1: 15481143-15481143 on Build GRCh38
rs3737699 Chr.1: 15481285-15481285 on Build GRCh38
rs754628236 Chr.1: 15481079-15481079 on Build GRCh38
rs921001883 Chr.1: 15481388-15481388 on Build GRCh38
rs747677881 Chr.1: 15481083-15481083 on Build GRCh38
rs756679268 Chr.1: 15481177-15481177 on Build GRCh38
rs574365343 Chr.1: 15481360-15481360 on Build GRCh38
rs757878349 Chr.1: 15481211-15481211 on Build GRCh38
rs570562745 Chr.1: 15481128-15481128 on Build GRCh38
rs369728005 Chr.1: 15481111-15481111 on Build GRCh38
rs748430964 Chr.1: 15481229-15481229 on Build GRCh38
rs370530919 Chr.1: 15481228-15481228 on Build GRCh38
rs778335998 Chr.1: 15481081-15481081 on Build GRCh38
rs375989268 Chr.1: 15481095-15481095 on Build GRCh38
rs760724872 Chr.1: 15481172-15481172 on Build GRCh38
rs115476135 Chr.1: 15481318-15481318 on Build GRCh38
rs754574613 Chr.1: 15481196-15481196 on Build GRCh38
rs529933634 Chr.1: 15481142-15481142 on Build GRCh38
rs766783109 Chr.1: 15481211-15481218 on Build GRCh38
rs913853956 Chr.1: 15481419-15481419 on Build GRCh38
rs750887470 Chr.1: 15481216-15481216 on Build GRCh38
rs754200534 Chr.1: 15481233-15481238 on Build GRCh38
rs762018574 Chr.1: 15481141-15481141 on Build GRCh38
rs145448066 Chr.1: 15481100-15481100 on Build GRCh38
rs752386188 Chr.1: 15481205-15481205 on Build GRCh38
rs772268016 Chr.1: 15481234-15481234 on Build GRCh38
rs994825093 Chr.1: 15481324-15481324 on Build GRCh38
rs957725535 Chr.1: 15481405-15481405 on Build GRCh38
rs373223907 Chr.1: 15481220-15481220 on Build GRCh38
rs576916677 Chr.1: 15481428-15481428 on Build GRCh38
rs764494200 Chr.1: 15481181-15481181 on Build GRCh38
rs150261602 Chr.1: 15481235-15481235 on Build GRCh38
rs950938434 Chr.1: 15481354-15481354 on Build GRCh38
rs757720815 Chr.1: 15481086-15481086 on Build GRCh38

View Full Product Details