Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 26772727-26773309 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GABRB3
Entrez Gene ID
2562
Gene Name
gamma-aminobutyric acid type A receptor beta3 subunit
Gene Aliases
ECA5
UniGene
Hs.302352
Gene Chromosome Location
Chr.15: 26543546-26773788 on Build GRCh38
Transcripts
NM_000814.5 NM_001278631.1 NM_021912.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5054220 c.65T>C p.V22A Chr.15: 26772898-26772898 on Build GRCh38
5971353 c.260C>T p.P87L Chr.15: 26772761-26772761 on Build GRCh38
459069 c.120G>A p.T40T Chr.15: 26772733-26772733 on Build GRCh38
5971351 c.92C>T p.P31L Chr.15: 26772761-26772761 on Build GRCh38
1207778 c.14C>T p.A5V Chr.15: 26772949-26772949 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759931649 Chr.15: 26772944-26772944 on Build GRCh38
rs113775796 Chr.15: 26773279-26773279 on Build GRCh38
rs777263662 Chr.15: 26772943-26772943 on Build GRCh38
rs751139880 Chr.15: 26772800-26772800 on Build GRCh38
rs868185545 Chr.15: 26772947-26772947 on Build GRCh38
rs867963618 Chr.15: 26773145-26773145 on Build GRCh38
rs868043781 Chr.15: 26773240-26773240 on Build GRCh38
rs368259836 Chr.15: 26772872-26772872 on Build GRCh38
rs775128696 Chr.15: 26772862-26772862 on Build GRCh38
rs576071277 Chr.15: 26773017-26773017 on Build GRCh38
rs1022889477 Chr.15: 26773226-26773226 on Build GRCh38
rs765680963 Chr.15: 26772946-26772946 on Build GRCh38
rs1049486764 Chr.15: 26772927-26772927 on Build GRCh38
rs1038242938 Chr.15: 26773185-26773185 on Build GRCh38
rs750860186 Chr.15: 26772798-26772799 on Build GRCh38
rs897943494 Chr.15: 26773219-26773219 on Build GRCh38
rs941187309 Chr.15: 26772988-26772988 on Build GRCh38
rs553697674 Chr.15: 26772832-26772832 on Build GRCh38
rs941054282 Chr.15: 26773177-26773177 on Build GRCh38
rs113689945 Chr.15: 26772996-26772996 on Build GRCh38
rs889516195 Chr.15: 26772985-26772985 on Build GRCh38
rs770927664 Chr.15: 26772838-26772838 on Build GRCh38
rs548775630 Chr.15: 26773041-26773043 on Build GRCh38
rs989453794 Chr.15: 26773126-26773126 on Build GRCh38
rs905992691 Chr.15: 26773230-26773230 on Build GRCh38
rs749065228 Chr.15: 26772870-26772870 on Build GRCh38
rs994490099 Chr.15: 26773069-26773069 on Build GRCh38
rs933662819 Chr.15: 26773139-26773139 on Build GRCh38
rs988966222 Chr.15: 26773251-26773251 on Build GRCh38
rs534064800 Chr.15: 26772829-26772829 on Build GRCh38
rs775267836 Chr.15: 26772794-26772794 on Build GRCh38
rs962491458 Chr.15: 26773262-26773262 on Build GRCh38
rs765658774 Chr.15: 26772795-26772796 on Build GRCh38
rs758722060 Chr.15: 26772968-26772968 on Build GRCh38
rs956391417 Chr.15: 26773246-26773246 on Build GRCh38
rs541676814 Chr.15: 26773255-26773255 on Build GRCh38
rs910976545 Chr.15: 26773153-26773153 on Build GRCh38
rs757381436 Chr.15: 26772986-26772986 on Build GRCh38
rs895794198 Chr.15: 26773093-26773093 on Build GRCh38
rs916110628 Chr.15: 26772852-26772852 on Build GRCh38
rs896372840 Chr.15: 26773220-26773220 on Build GRCh38
rs766998233 Chr.15: 26772802-26772802 on Build GRCh38
rs896976029 Chr.15: 26773186-26773186 on Build GRCh38
rs1014213875 Chr.15: 26773222-26773222 on Build GRCh38
rs1037772930 Chr.15: 26772845-26772845 on Build GRCh38
rs868500530 Chr.15: 26772922-26772922 on Build GRCh38
rs1023026753 Chr.15: 26773101-26773101 on Build GRCh38
rs755429819 Chr.15: 26772805-26772805 on Build GRCh38
rs761370845 Chr.15: 26772801-26772801 on Build GRCh38
rs926231041 Chr.15: 26772871-26772871 on Build GRCh38
rs1056769866 Chr.15: 26773212-26773212 on Build GRCh38
rs1033590940 Chr.15: 26773118-26773118 on Build GRCh38
rs746996582 Chr.15: 26772775-26772775 on Build GRCh38
rs978900908 Chr.15: 26773111-26773111 on Build GRCh38
rs988532310 Chr.15: 26772875-26772875 on Build GRCh38
rs956348540 Chr.15: 26773124-26773124 on Build GRCh38
rs753024924 Chr.15: 26772955-26772955 on Build GRCh38
rs985096456 Chr.15: 26773152-26773157 on Build GRCh38
rs779143981 Chr.15: 26772813-26772813 on Build GRCh38
rs556238396 Chr.15: 26772938-26772938 on Build GRCh38
rs372462529 Chr.15: 26772915-26772915 on Build GRCh38
rs769695299 Chr.15: 26772858-26772858 on Build GRCh38
rs772263479 Chr.15: 26772766-26772766 on Build GRCh38
rs922315216 Chr.15: 26773133-26773133 on Build GRCh38
rs970079419 Chr.15: 26773108-26773108 on Build GRCh38
rs867679278 Chr.15: 26772887-26772887 on Build GRCh38
rs761422251 Chr.15: 26772924-26772924 on Build GRCh38
rs565238494 Chr.15: 26773252-26773252 on Build GRCh38
rs776511065 Chr.15: 26772779-26772779 on Build GRCh38
rs769413892 Chr.15: 26772788-26772788 on Build GRCh38
rs748306024 Chr.15: 26772754-26772754 on Build GRCh38
rs573761918 Chr.15: 26772851-26772851 on Build GRCh38
rs1014318342 Chr.15: 26773094-26773094 on Build GRCh38
rs542737772 Chr.15: 26772879-26772879 on Build GRCh38
rs201579427 Chr.15: 26772777-26772777 on Build GRCh38
rs929932118 Chr.15: 26773195-26773195 on Build GRCh38
rs763748758 Chr.15: 26772798-26772798 on Build GRCh38
rs865875277 Chr.15: 26772883-26772883 on Build GRCh38
rs751719249 Chr.15: 26772984-26772984 on Build GRCh38
rs961562121 Chr.15: 26772803-26772803 on Build GRCh38
rs71651682 Chr.15: 26772759-26772759 on Build GRCh38
rs758595473 Chr.15: 26772823-26772823 on Build GRCh38
rs777797308 Chr.15: 26772770-26772770 on Build GRCh38
rs889087740 Chr.15: 26772797-26772797 on Build GRCh38
rs985132493 Chr.15: 26773253-26773253 on Build GRCh38
rs753002765 Chr.15: 26772814-26772814 on Build GRCh38
rs759375301 Chr.15: 26772783-26772783 on Build GRCh38
rs762506428 Chr.15: 26772795-26772795 on Build GRCh38
rs781116888 Chr.15: 26772843-26772843 on Build GRCh38
rs974209464 Chr.15: 26773284-26773284 on Build GRCh38
rs25407 Chr.15: 26772785-26772785 on Build GRCh38
rs1038176877 Chr.15: 26773020-26773020 on Build GRCh38
rs545012756 Chr.15: 26773072-26773072 on Build GRCh38
rs897073733 Chr.15: 26773062-26773064 on Build GRCh38

View Full Product Details