Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 78131094-78131641 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CIB2
Entrez Gene ID
10518
Gene Name
calcium and integrin binding family member 2
Gene Aliases
DFNB48, KIP2, USH1J
UniGene
Hs.129867
Gene Chromosome Location
Chr.15: 78104606-78131976 on Build GRCh38
Transcripts
NM_001271888.1 NM_001271889.1 NM_001301224.1 NM_006383.3 NR_125435.1 XM_005254126.3 XM_006720374.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1735398 c.45C>G p.N15K Chr.15: 78131171-78131171 on Build GRCh38
6250718 c.42C>A p.D14E Chr.15: 78131174-78131174 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1013909595 Chr.15: 78131558-78131558 on Build GRCh38
rs761904991 Chr.15: 78131265-78131265 on Build GRCh38
rs760756694 Chr.15: 78131434-78131434 on Build GRCh38
rs916538782 Chr.15: 78131439-78131439 on Build GRCh38
rs978790224 Chr.15: 78131596-78131596 on Build GRCh38
rs1029947051 Chr.15: 78131383-78131383 on Build GRCh38
rs934001232 Chr.15: 78131501-78131501 on Build GRCh38
rs916486259 Chr.15: 78131292-78131292 on Build GRCh38
rs969270460 Chr.15: 78131442-78131442 on Build GRCh38
rs201655250 Chr.15: 78131237-78131237 on Build GRCh38
rs764140956 Chr.15: 78131159-78131159 on Build GRCh38
rs998450090 Chr.15: 78131382-78131383 on Build GRCh38
rs368498640 Chr.15: 78131249-78131249 on Build GRCh38
rs757773491 Chr.15: 78131122-78131122 on Build GRCh38
rs559331816 Chr.15: 78131490-78131490 on Build GRCh38
rs772637977 Chr.15: 78131213-78131213 on Build GRCh38
rs1037534834 Chr.15: 78131507-78131507 on Build GRCh38
rs775414156 Chr.15: 78131246-78131246 on Build GRCh38
rs757748764 Chr.15: 78131168-78131168 on Build GRCh38
rs972843105 Chr.15: 78131201-78131201 on Build GRCh38
rs898995405 Chr.15: 78131508-78131508 on Build GRCh38
rs776613190 Chr.15: 78131139-78131139 on Build GRCh38
rs1043539185 Chr.15: 78131312-78131313 on Build GRCh38
rs373826971 Chr.15: 78131204-78131204 on Build GRCh38
rs891768996 Chr.15: 78131360-78131360 on Build GRCh38
rs1017105941 Chr.15: 78131404-78131404 on Build GRCh38
rs765585209 Chr.15: 78131148-78131148 on Build GRCh38
rs772763900 Chr.15: 78131136-78131136 on Build GRCh38
rs768347318 Chr.15: 78131127-78131127 on Build GRCh38
rs536373665 Chr.15: 78131225-78131225 on Build GRCh38
rs1016715787 Chr.15: 78131166-78131166 on Build GRCh38
rs773945311 Chr.15: 78131128-78131128 on Build GRCh38
rs1034525184 Chr.15: 78131550-78131550 on Build GRCh38
rs925103936 Chr.15: 78131481-78131481 on Build GRCh38
rs766309761 Chr.15: 78131138-78131138 on Build GRCh38
rs771750530 Chr.15: 78131207-78131207 on Build GRCh38
rs958456754 Chr.15: 78131618-78131618 on Build GRCh38
rs962784479 Chr.15: 78131412-78131412 on Build GRCh38
rs529776066 Chr.15: 78131516-78131516 on Build GRCh38
rs947915936 Chr.15: 78131303-78131303 on Build GRCh38
rs925436511 Chr.15: 78131601-78131601 on Build GRCh38
rs752897876 Chr.15: 78131153-78131153 on Build GRCh38
rs564839260 Chr.15: 78131324-78131324 on Build GRCh38
rs1023224907 Chr.15: 78131593-78131593 on Build GRCh38
rs566097906 Chr.15: 78131429-78131429 on Build GRCh38
rs913293249 Chr.15: 78131495-78131495 on Build GRCh38
rs770791635 Chr.15: 78131218-78131218 on Build GRCh38
rs746255131 Chr.15: 78131183-78131183 on Build GRCh38
rs747419043 Chr.15: 78131121-78131121 on Build GRCh38
rs369854897 Chr.15: 78131271-78131271 on Build GRCh38
rs1007101767 Chr.15: 78131392-78131392 on Build GRCh38
rs747689693 Chr.15: 78131131-78131131 on Build GRCh38
rs771423166 Chr.15: 78131132-78131132 on Build GRCh38
rs898635908 Chr.15: 78131535-78131535 on Build GRCh38
rs8040395 Chr.15: 78131137-78131137 on Build GRCh38
rs991768892 Chr.15: 78131436-78131436 on Build GRCh38
rs1025745427 Chr.15: 78131431-78131431 on Build GRCh38
rs11547207 Chr.15: 78131254-78131254 on Build GRCh38
rs1007114540 Chr.15: 78131519-78131519 on Build GRCh38
rs966943103 Chr.15: 78131568-78131568 on Build GRCh38
rs1053530066 Chr.15: 78131358-78131358 on Build GRCh38
rs1019989609 Chr.15: 78131567-78131567 on Build GRCh38
rs1051092623 Chr.15: 78131506-78131506 on Build GRCh38
rs759352267 Chr.15: 78131143-78131143 on Build GRCh38
rs890022826 Chr.15: 78131384-78131384 on Build GRCh38
rs781480464 Chr.15: 78131123-78131123 on Build GRCh38
rs758568331 Chr.15: 78131156-78131156 on Build GRCh38
rs1038962223 Chr.15: 78131530-78131530 on Build GRCh38
rs936099795 Chr.15: 78131487-78131487 on Build GRCh38
rs746501975 Chr.15: 78131215-78131215 on Build GRCh38
rs767156167 Chr.15: 78131236-78131237 on Build GRCh38
rs751658542 Chr.15: 78131164-78131164 on Build GRCh38
rs759407248 Chr.15: 78131235-78131235 on Build GRCh38
rs756431884 Chr.15: 78131196-78131196 on Build GRCh38
rs541211286 Chr.15: 78131453-78131453 on Build GRCh38
rs746200171 Chr.15: 78131124-78131124 on Build GRCh38
rs778544125 Chr.15: 78131199-78131199 on Build GRCh38
rs781709015 Chr.15: 78131179-78131179 on Build GRCh38
rs936067811 Chr.15: 78131354-78131354 on Build GRCh38
rs959670950 Chr.15: 78131555-78131555 on Build GRCh38
rs962488743 Chr.15: 78131192-78131192 on Build GRCh38

View Full Product Details