Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 83004045-83004642 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C15orf40
Entrez Gene ID
123207
Gene Name
chromosome 15 open reading frame 40
Gene Aliases
-
UniGene
Hs.352541
Gene Chromosome Location
Chr.15: 82988297-83012298 on Build GRCh38
Transcripts
NM_001160114.1 NM_001160115.1 NM_001160116.1 NM_144597.2 NR_027650.1 XM_006720385.2 XM_011521212.2 XM_011521213.2

SNP Detail

SNP ID Chromosome Location
rs993336462 Chr.15: 83004587-83004587 on Build GRCh38
rs926130616 Chr.15: 83004383-83004383 on Build GRCh38
rs71412288 Chr.15: 83004157-83004157 on Build GRCh38
rs1038136847 Chr.15: 83004234-83004234 on Build GRCh38
rs1047519049 Chr.15: 83004166-83004166 on Build GRCh38
rs966460542 Chr.15: 83004099-83004102 on Build GRCh38
rs533736081 Chr.15: 83004098-83004098 on Build GRCh38
rs556369584 Chr.15: 83004170-83004170 on Build GRCh38
rs974631369 Chr.15: 83004125-83004125 on Build GRCh38
rs974347318 Chr.15: 83004388-83004388 on Build GRCh38
rs35500715 Chr.15: 83004305-83004306 on Build GRCh38
rs202036704 Chr.15: 83004555-83004558 on Build GRCh38
rs993836416 Chr.15: 83004340-83004340 on Build GRCh38
rs917214478 Chr.15: 83004430-83004430 on Build GRCh38
rs1024782755 Chr.15: 83004082-83004082 on Build GRCh38
rs185634900 Chr.15: 83004357-83004357 on Build GRCh38
rs143864972 Chr.15: 83004105-83004105 on Build GRCh38
rs918942721 Chr.15: 83004130-83004130 on Build GRCh38
rs954381675 Chr.15: 83004347-83004347 on Build GRCh38
rs551791638 Chr.15: 83004525-83004525 on Build GRCh38
rs979519036 Chr.15: 83004104-83004104 on Build GRCh38
rs555053403 Chr.15: 83004098-83004099 on Build GRCh38
rs773782702 Chr.15: 83004598-83004598 on Build GRCh38
rs907687551 Chr.15: 83004408-83004408 on Build GRCh38
rs184373304 Chr.15: 83004178-83004178 on Build GRCh38
rs1025903108 Chr.15: 83004344-83004344 on Build GRCh38
rs373538087 Chr.15: 83004167-83004167 on Build GRCh38
rs776077448 Chr.15: 83004474-83004474 on Build GRCh38
rs905975718 Chr.15: 83004472-83004472 on Build GRCh38
rs934636289 Chr.15: 83004220-83004220 on Build GRCh38
rs905022102 Chr.15: 83004257-83004257 on Build GRCh38
rs899002236 Chr.15: 83004579-83004579 on Build GRCh38
rs866703015 Chr.15: 83004122-83004122 on Build GRCh38
rs1026229070 Chr.15: 83004588-83004588 on Build GRCh38
rs555222337 Chr.15: 83004267-83004267 on Build GRCh38
rs763627395 Chr.15: 83004571-83004571 on Build GRCh38
rs1037511771 Chr.15: 83004529-83004529 on Build GRCh38
rs964341373 Chr.15: 83004371-83004371 on Build GRCh38
rs943591920 Chr.15: 83004162-83004162 on Build GRCh38
rs1057130143 Chr.15: 83004555-83004557 on Build GRCh38
rs71412287 Chr.15: 83004151-83004151 on Build GRCh38
rs866740591 Chr.15: 83004561-83004561 on Build GRCh38
rs540765065 Chr.15: 83004493-83004493 on Build GRCh38
rs10083713 Chr.15: 83004403-83004403 on Build GRCh38
rs8035233 Chr.15: 83004613-83004613 on Build GRCh38
rs926257020 Chr.15: 83004209-83004209 on Build GRCh38
rs770998396 Chr.15: 83004120-83004125 on Build GRCh38
rs895831498 Chr.15: 83004240-83004240 on Build GRCh38
rs963366729 Chr.15: 83004110-83004110 on Build GRCh38
rs376811731 Chr.15: 83004464-83004464 on Build GRCh38
rs987190614 Chr.15: 83004141-83004141 on Build GRCh38
rs1053082971 Chr.15: 83004230-83004230 on Build GRCh38
rs571868245 Chr.15: 83004576-83004576 on Build GRCh38
rs937325960 Chr.15: 83004507-83004507 on Build GRCh38
rs1057398088 Chr.15: 83004261-83004261 on Build GRCh38
rs965231548 Chr.15: 83004372-83004372 on Build GRCh38
rs942098506 Chr.15: 83004191-83004191 on Build GRCh38
rs987180819 Chr.15: 83004354-83004354 on Build GRCh38
rs978973846 Chr.15: 83004373-83004373 on Build GRCh38
rs963303253 Chr.15: 83004306-83004306 on Build GRCh38
rs550207447 Chr.15: 83004246-83004246 on Build GRCh38
rs907970419 Chr.15: 83004177-83004177 on Build GRCh38
rs542847954 Chr.15: 83004402-83004402 on Build GRCh38
rs544988980 Chr.15: 83004241-83004241 on Build GRCh38
rs553594344 Chr.15: 83004099-83004099 on Build GRCh38
rs531762503 Chr.15: 83004497-83004497 on Build GRCh38
rs568221390 Chr.15: 83004552-83004556 on Build GRCh38
rs969216538 Chr.15: 83004085-83004085 on Build GRCh38
rs1014313086 Chr.15: 83004255-83004255 on Build GRCh38
rs1020421764 Chr.15: 83004299-83004299 on Build GRCh38
rs910682669 Chr.15: 83004144-83004144 on Build GRCh38
rs905859782 Chr.15: 83004235-83004235 on Build GRCh38
rs923144353 Chr.15: 83004126-83004129 on Build GRCh38
rs950069622 Chr.15: 83004439-83004439 on Build GRCh38
rs193146032 Chr.15: 83004275-83004275 on Build GRCh38
rs74889982 Chr.15: 83004258-83004258 on Build GRCh38
rs146849516 Chr.15: 83004134-83004134 on Build GRCh38
rs187896426 Chr.15: 83004256-83004256 on Build GRCh38
rs761476898 Chr.15: 83004615-83004615 on Build GRCh38
rs942133714 Chr.15: 83004413-83004413 on Build GRCh38
rs1043695757 Chr.15: 83004187-83004187 on Build GRCh38
rs1044490480 Chr.15: 83004467-83004467 on Build GRCh38
rs923195288 Chr.15: 83004397-83004397 on Build GRCh38
rs1000600520 Chr.15: 83004317-83004317 on Build GRCh38
rs910338645 Chr.15: 83004362-83004362 on Build GRCh38
rs1042300159 Chr.15: 83004616-83004616 on Build GRCh38

View Full Product Details