Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 65201434-65202014 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CILP
Entrez Gene ID
8483
Gene Name
cartilage intermediate layer protein
Gene Aliases
CILP-1, HsT18872
UniGene
Hs.442180
Gene Chromosome Location
Chr.15: 65195999-65211608 on Build GRCh38
Transcripts
NM_003613.3 XM_017022678.1 XM_017022679.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5680515 c.1150G>T p.A384S Chr.15: 65201908-65201908 on Build GRCh38
5528702 c.1092G>A p.R364R Chr.15: 65201966-65201966 on Build GRCh38
964032 c.1160C>A p.S387Y Chr.15: 65201898-65201898 on Build GRCh38
4394153 c.1141G>A p.D381N Chr.15: 65201917-65201917 on Build GRCh38
4056279 c.1142A>C p.D381A Chr.15: 65201916-65201916 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779874552 Chr.15: 65201905-65201905 on Build GRCh38
rs201640053 Chr.15: 65201940-65201940 on Build GRCh38
rs386784824 Chr.15: 65201763-65201772 on Build GRCh38
rs530490042 Chr.15: 65201812-65201812 on Build GRCh38
rs971314078 Chr.15: 65201829-65201829 on Build GRCh38
rs551518793 Chr.15: 65201602-65201602 on Build GRCh38
rs549295927 Chr.15: 65201461-65201461 on Build GRCh38
rs2073708 Chr.15: 65201514-65201514 on Build GRCh38
rs1032383182 Chr.15: 65201657-65201657 on Build GRCh38
rs59737757 Chr.15: 65201749-65201750 on Build GRCh38
rs532792713 Chr.15: 65201726-65201726 on Build GRCh38
rs796192701 Chr.15: 65201760-65201772 on Build GRCh38
rs750383086 Chr.15: 65201887-65201887 on Build GRCh38
rs760973054 Chr.15: 65201984-65201984 on Build GRCh38
rs371812207 Chr.15: 65201765-65201772 on Build GRCh38
rs987994481 Chr.15: 65201467-65201467 on Build GRCh38
rs779740186 Chr.15: 65201947-65201948 on Build GRCh38
rs371735478 Chr.15: 65201922-65201922 on Build GRCh38
rs1040879377 Chr.15: 65201584-65201584 on Build GRCh38
rs973345074 Chr.15: 65201522-65201522 on Build GRCh38
rs746915122 Chr.15: 65201952-65201952 on Build GRCh38
rs1030191251 Chr.15: 65201693-65201693 on Build GRCh38
rs560662089 Chr.15: 65201723-65201723 on Build GRCh38
rs138700406 Chr.15: 65201594-65201594 on Build GRCh38
rs980452632 Chr.15: 65201858-65201858 on Build GRCh38
rs746652286 Chr.15: 65201730-65201731 on Build GRCh38
rs375170690 Chr.15: 65201967-65201967 on Build GRCh38
rs368473423 Chr.15: 65201760-65201760 on Build GRCh38
rs201892140 Chr.15: 65201886-65201886 on Build GRCh38
rs760923066 Chr.15: 65201859-65201859 on Build GRCh38
rs887040884 Chr.15: 65201515-65201515 on Build GRCh38
rs376599011 Chr.15: 65201772-65201773 on Build GRCh38
rs145769526 Chr.15: 65201541-65201541 on Build GRCh38
rs753055415 Chr.15: 65201846-65201846 on Build GRCh38
rs956959963 Chr.15: 65201745-65201745 on Build GRCh38
rs376717139 Chr.15: 65201761-65201769 on Build GRCh38
rs2681037 Chr.15: 65201747-65201747 on Build GRCh38
rs963227746 Chr.15: 65201908-65201908 on Build GRCh38
rs942032433 Chr.15: 65201516-65201516 on Build GRCh38
rs186351044 Chr.15: 65201550-65201550 on Build GRCh38
rs772498958 Chr.15: 65201982-65201982 on Build GRCh38
rs764257410 Chr.15: 65201866-65201866 on Build GRCh38
rs762437638 Chr.15: 65201729-65201731 on Build GRCh38
rs530119520 Chr.15: 65201715-65201715 on Build GRCh38
rs575197397 Chr.15: 65201636-65201636 on Build GRCh38
rs757426664 Chr.15: 65201883-65201883 on Build GRCh38
rs904894154 Chr.15: 65201622-65201622 on Build GRCh38
rs778591557 Chr.15: 65201865-65201866 on Build GRCh38
rs1030554217 Chr.15: 65201481-65201481 on Build GRCh38
rs1057315723 Chr.15: 65201578-65201578 on Build GRCh38
rs771830165 Chr.15: 65201940-65201941 on Build GRCh38
rs769294305 Chr.15: 65201950-65201950 on Build GRCh38
rs537053213 Chr.15: 65201462-65201462 on Build GRCh38
rs985996257 Chr.15: 65201505-65201505 on Build GRCh38
rs536411120 Chr.15: 65201549-65201549 on Build GRCh38
rs372982626 Chr.15: 65201656-65201656 on Build GRCh38
rs759827178 Chr.15: 65201835-65201835 on Build GRCh38
rs552862588 Chr.15: 65201743-65201743 on Build GRCh38
rs780729543 Chr.15: 65201926-65201926 on Build GRCh38
rs953335285 Chr.15: 65201615-65201615 on Build GRCh38
rs369880105 Chr.15: 65201499-65201499 on Build GRCh38
rs929361628 Chr.15: 65201555-65201555 on Build GRCh38
rs773393854 Chr.15: 65201959-65201960 on Build GRCh38
rs754617486 Chr.15: 65201924-65201924 on Build GRCh38
rs758470105 Chr.15: 65201888-65201888 on Build GRCh38
rs1000880630 Chr.15: 65201651-65201651 on Build GRCh38
rs1009702017 Chr.15: 65201677-65201677 on Build GRCh38
rs548210093 Chr.15: 65201673-65201673 on Build GRCh38
rs544287318 Chr.15: 65201654-65201654 on Build GRCh38
rs772772706 Chr.15: 65201977-65201977 on Build GRCh38
rs376283956 Chr.15: 65201747-65201748 on Build GRCh38
rs747595889 Chr.15: 65201939-65201939 on Build GRCh38
rs116251736 Chr.15: 65201790-65201790 on Build GRCh38
rs377627736 Chr.15: 65201716-65201716 on Build GRCh38
rs796662345 Chr.15: 65201741-65201747 on Build GRCh38
rs926259871 Chr.15: 65201494-65201494 on Build GRCh38
rs550320492 Chr.15: 65201840-65201840 on Build GRCh38
rs775887125 Chr.15: 65201983-65201983 on Build GRCh38
rs745399926 Chr.15: 65201610-65201610 on Build GRCh38
rs768762257 Chr.15: 65201954-65201955 on Build GRCh38
rs2073711 Chr.15: 65201874-65201874 on Build GRCh38
rs572729999 Chr.15: 65201592-65201592 on Build GRCh38
rs745516539 Chr.15: 65201908-65201908 on Build GRCh38

View Full Product Details