Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 20858043-20858618 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCUN1D3
Entrez Gene ID
123879
Gene Name
defective in cullin neddylation 1 domain containing 3
Gene Aliases
44M2.4, SCCRO3
UniGene
Hs.101007
Gene Chromosome Location
Chr.16: 20854925-20900358 on Build GRCh38
Transcripts
NM_173475.3

SNP Detail

SNP ID Chromosome Location
rs12919570 Chr.16: 20858523-20858523 on Build GRCh38
rs369767029 Chr.16: 20858407-20858407 on Build GRCh38
rs878884322 Chr.16: 20858405-20858405 on Build GRCh38
rs561552421 Chr.16: 20858304-20858304 on Build GRCh38
rs967540017 Chr.16: 20858081-20858081 on Build GRCh38
rs937914430 Chr.16: 20858342-20858342 on Build GRCh38
rs1008335337 Chr.16: 20858520-20858520 on Build GRCh38
rs558706770 Chr.16: 20858085-20858085 on Build GRCh38
rs71374837 Chr.16: 20858353-20858353 on Build GRCh38
rs28418371 Chr.16: 20858570-20858570 on Build GRCh38
rs557086730 Chr.16: 20858524-20858524 on Build GRCh38
rs574547779 Chr.16: 20858325-20858325 on Build GRCh38
rs905055783 Chr.16: 20858339-20858339 on Build GRCh38
rs534829003 Chr.16: 20858187-20858187 on Build GRCh38
rs773634080 Chr.16: 20858358-20858358 on Build GRCh38
rs541647491 Chr.16: 20858355-20858355 on Build GRCh38
rs531497458 Chr.16: 20858116-20858116 on Build GRCh38
rs553057962 Chr.16: 20858223-20858223 on Build GRCh38
rs548043634 Chr.16: 20858294-20858294 on Build GRCh38
rs1012001374 Chr.16: 20858389-20858389 on Build GRCh38
rs28665909 Chr.16: 20858568-20858568 on Build GRCh38
rs71374836 Chr.16: 20858303-20858303 on Build GRCh38
rs12919569 Chr.16: 20858521-20858521 on Build GRCh38
rs912776452 Chr.16: 20858121-20858121 on Build GRCh38
rs949196251 Chr.16: 20858326-20858326 on Build GRCh38
rs975692749 Chr.16: 20858245-20858245 on Build GRCh38
rs903265917 Chr.16: 20858450-20858450 on Build GRCh38
rs1019781021 Chr.16: 20858522-20858522 on Build GRCh38
rs1020320625 Chr.16: 20858423-20858423 on Build GRCh38
rs908297306 Chr.16: 20858305-20858305 on Build GRCh38
rs537733579 Chr.16: 20858070-20858070 on Build GRCh38
rs919564374 Chr.16: 20858279-20858279 on Build GRCh38
rs12919573 Chr.16: 20858533-20858533 on Build GRCh38
rs1046212563 Chr.16: 20858327-20858327 on Build GRCh38

View Full Product Details