Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 35872226-35872749 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCL5
Entrez Gene ID
6352
Gene Name
C-C motif chemokine ligand 5
Gene Aliases
D17S136E, RANTES, SCYA5, SIS-delta, SISd, TCP228, eoCP
UniGene
Hs.514821
Gene Chromosome Location
Chr.17: 35871491-35880373 on Build GRCh38
Transcripts
NM_001278736.1 NM_002985.2

SNP Detail

SNP ID Chromosome Location
rs749276861 Chr.17: 35872292-35872292 on Build GRCh38
rs985838642 Chr.17: 35872620-35872620 on Build GRCh38
rs376930383 Chr.17: 35872451-35872451 on Build GRCh38
rs769571877 Chr.17: 35872333-35872333 on Build GRCh38
rs774402327 Chr.17: 35872309-35872309 on Build GRCh38
rs75062043 Chr.17: 35872691-35872691 on Build GRCh38
rs757287512 Chr.17: 35872488-35872488 on Build GRCh38
rs757614591 Chr.17: 35872286-35872286 on Build GRCh38
rs909202351 Chr.17: 35872710-35872710 on Build GRCh38
rs982361824 Chr.17: 35872650-35872650 on Build GRCh38
rs376576620 Chr.17: 35872513-35872513 on Build GRCh38
rs766974257 Chr.17: 35872484-35872484 on Build GRCh38
rs768718938 Chr.17: 35872257-35872257 on Build GRCh38
rs1018714251 Chr.17: 35872631-35872631 on Build GRCh38
rs370171166 Chr.17: 35872330-35872330 on Build GRCh38
rs78288036 Chr.17: 35872521-35872521 on Build GRCh38
rs149913745 Chr.17: 35872445-35872445 on Build GRCh38
rs768066628 Chr.17: 35872452-35872452 on Build GRCh38
rs1039556077 Chr.17: 35872570-35872570 on Build GRCh38
rs1055806210 Chr.17: 35872465-35872465 on Build GRCh38
rs185969338 Chr.17: 35872313-35872313 on Build GRCh38
rs749528591 Chr.17: 35872253-35872253 on Build GRCh38
rs373485196 Chr.17: 35872442-35872442 on Build GRCh38
rs752777330 Chr.17: 35872515-35872515 on Build GRCh38
rs368100847 Chr.17: 35872481-35872481 on Build GRCh38
rs746386285 Chr.17: 35872289-35872289 on Build GRCh38
rs1032067574 Chr.17: 35872410-35872410 on Build GRCh38
rs765182976 Chr.17: 35872473-35872475 on Build GRCh38
rs751159728 Chr.17: 35872363-35872363 on Build GRCh38
rs533886706 Chr.17: 35872688-35872688 on Build GRCh38
rs1051990001 Chr.17: 35872578-35872578 on Build GRCh38
rs944525463 Chr.17: 35872369-35872369 on Build GRCh38
rs966329985 Chr.17: 35872404-35872404 on Build GRCh38
rs374198299 Chr.17: 35872450-35872450 on Build GRCh38
rs767873033 Chr.17: 35872372-35872372 on Build GRCh38
rs181824835 Chr.17: 35872301-35872301 on Build GRCh38
rs763632936 Chr.17: 35872282-35872282 on Build GRCh38
rs773079594 Chr.17: 35872259-35872259 on Build GRCh38
rs74589003 Chr.17: 35872285-35872285 on Build GRCh38
rs761483805 Chr.17: 35872454-35872454 on Build GRCh38
rs759002804 Chr.17: 35872568-35872568 on Build GRCh38
rs772833516 Chr.17: 35872714-35872714 on Build GRCh38
rs776506453 Chr.17: 35872322-35872322 on Build GRCh38
rs189189494 Chr.17: 35872273-35872273 on Build GRCh38
rs200852773 Chr.17: 35872453-35872453 on Build GRCh38
rs999169591 Chr.17: 35872595-35872595 on Build GRCh38
rs577904395 Chr.17: 35872632-35872632 on Build GRCh38
rs781159560 Chr.17: 35872438-35872438 on Build GRCh38
rs998131322 Chr.17: 35872577-35872577 on Build GRCh38
rs1031995654 Chr.17: 35872597-35872597 on Build GRCh38
rs372562673 Chr.17: 35872345-35872345 on Build GRCh38
rs751997637 Chr.17: 35872283-35872283 on Build GRCh38
rs761931677 Chr.17: 35872369-35872370 on Build GRCh38
rs1015132261 Chr.17: 35872559-35872559 on Build GRCh38
rs755452101 Chr.17: 35872511-35872511 on Build GRCh38
rs35198662 Chr.17: 35872301-35872302 on Build GRCh38
rs760362934 Chr.17: 35872260-35872260 on Build GRCh38
rs147455197 Chr.17: 35872571-35872571 on Build GRCh38
rs528220869 Chr.17: 35872702-35872702 on Build GRCh38
rs758094681 Chr.17: 35872279-35872279 on Build GRCh38
rs770833181 Chr.17: 35872317-35872317 on Build GRCh38
rs756634409 Chr.17: 35872403-35872403 on Build GRCh38
rs764848801 Chr.17: 35872274-35872274 on Build GRCh38
rs997764126 Chr.17: 35872358-35872358 on Build GRCh38
rs1047733705 Chr.17: 35872486-35872486 on Build GRCh38
rs753989105 Chr.17: 35872405-35872405 on Build GRCh38
rs767106130 Chr.17: 35872464-35872464 on Build GRCh38
rs147829173 Chr.17: 35872389-35872389 on Build GRCh38
rs776702497 Chr.17: 35872268-35872268 on Build GRCh38
rs752111320 Chr.17: 35872278-35872278 on Build GRCh38
rs112293582 Chr.17: 35872676-35872676 on Build GRCh38
rs750833551 Chr.17: 35872467-35872467 on Build GRCh38
rs761396978 Chr.17: 35872368-35872368 on Build GRCh38
rs572481560 Chr.17: 35872510-35872510 on Build GRCh38
rs112389936 Chr.17: 35872499-35872499 on Build GRCh38
rs868439163 Chr.17: 35872266-35872266 on Build GRCh38
rs750745230 Chr.17: 35872396-35872396 on Build GRCh38
rs902782816 Chr.17: 35872592-35872592 on Build GRCh38
rs551348072 Chr.17: 35872707-35872707 on Build GRCh38
rs201244295 Chr.17: 35872437-35872437 on Build GRCh38
rs28914812 Chr.17: 35872634-35872634 on Build GRCh38
rs140271646 Chr.17: 35872444-35872444 on Build GRCh38
rs775116117 Chr.17: 35872339-35872339 on Build GRCh38
rs1031034180 Chr.17: 35872362-35872362 on Build GRCh38
rs909277264 Chr.17: 35872545-35872545 on Build GRCh38
rs369280877 Chr.17: 35872408-35872408 on Build GRCh38
rs754081737 Chr.17: 35872459-35872459 on Build GRCh38
rs762340963 Chr.17: 35872344-35872344 on Build GRCh38
rs779249457 Chr.17: 35872409-35872409 on Build GRCh38
rs766454792 Chr.17: 35872262-35872262 on Build GRCh38
rs957794382 Chr.17: 35872413-35872413 on Build GRCh38
rs756546334 Chr.17: 35872290-35872290 on Build GRCh38
rs3817655 Chr.17: 35872637-35872637 on Build GRCh38
rs780947360 Chr.17: 35872656-35872656 on Build GRCh38
rs970546164 Chr.17: 35872646-35872646 on Build GRCh38
rs748390053 Chr.17: 35872429-35872429 on Build GRCh38
rs536988547 Chr.17: 35872613-35872613 on Build GRCh38
rs750690767 Chr.17: 35872477-35872479 on Build GRCh38
rs779091034 Chr.17: 35872562-35872562 on Build GRCh38

View Full Product Details