Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 85775530-85776096 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COL24A1
Entrez Gene ID
255631
Gene Name
collagen type XXIV alpha 1
Gene Aliases
-
UniGene
Hs.659516
Gene Chromosome Location
Chr.1: 85728507-86156987 on Build GRCh38
Transcripts
NM_152890.5 XM_017000924.1 XM_017000925.1 XM_017000926.1 XM_017000927.1 XM_017000928.1 XM_017000929.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7307598 c.4339-8_4339-7insT p.? Chr.1: 85775716-85775717 on Build GRCh38
2244161 c.4374+4A>C p.? Chr.1: 85775670-85775670 on Build GRCh38
4010311 c.4357G>A p.G1453S Chr.1: 85775691-85775691 on Build GRCh38
7261922 c.4339-8delT p.? Chr.1: 85775717-85775717 on Build GRCh38
1639982 c.4355delA p.K1452fs*40 Chr.1: 85775693-85775693 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1008377507 Chr.1: 85775597-85775597 on Build GRCh38
rs944215179 Chr.1: 85775898-85775898 on Build GRCh38
rs781765676 Chr.1: 85775748-85775748 on Build GRCh38
rs996915895 Chr.1: 85775939-85775939 on Build GRCh38
rs368488549 Chr.1: 85775756-85775756 on Build GRCh38
rs374747073 Chr.1: 85775755-85775755 on Build GRCh38
rs760666123 Chr.1: 85775685-85775685 on Build GRCh38
rs181886478 Chr.1: 85776029-85776029 on Build GRCh38
rs777490193 Chr.1: 85775727-85775727 on Build GRCh38
rs928931816 Chr.1: 85776060-85776060 on Build GRCh38
rs991271522 Chr.1: 85775783-85775783 on Build GRCh38
rs970122245 Chr.1: 85776054-85776054 on Build GRCh38
rs913665365 Chr.1: 85775852-85775852 on Build GRCh38
rs779675159 Chr.1: 85775693-85775694 on Build GRCh38
rs550833348 Chr.1: 85775932-85775932 on Build GRCh38
rs1053139237 Chr.1: 85775883-85775883 on Build GRCh38
rs1036047623 Chr.1: 85776062-85776062 on Build GRCh38
rs1043413536 Chr.1: 85775895-85775895 on Build GRCh38
rs1048498128 Chr.1: 85775998-85775998 on Build GRCh38
rs75364489 Chr.1: 85776030-85776030 on Build GRCh38
rs1015661870 Chr.1: 85775635-85775635 on Build GRCh38
rs1053266378 Chr.1: 85775842-85775842 on Build GRCh38
rs959956591 Chr.1: 85776068-85776068 on Build GRCh38
rs370588081 Chr.1: 85775749-85775749 on Build GRCh38
rs934341371 Chr.1: 85775848-85775848 on Build GRCh38
rs995555422 Chr.1: 85775988-85775988 on Build GRCh38
rs571796429 Chr.1: 85775781-85775781 on Build GRCh38
rs887148869 Chr.1: 85776005-85776005 on Build GRCh38
rs1049691242 Chr.1: 85775966-85775966 on Build GRCh38
rs763183949 Chr.1: 85775649-85775649 on Build GRCh38
rs762275919 Chr.1: 85776056-85776056 on Build GRCh38
rs550729519 Chr.1: 85775717-85775717 on Build GRCh38
rs746503445 Chr.1: 85775733-85775733 on Build GRCh38
rs775603305 Chr.1: 85775641-85775641 on Build GRCh38
rs961414551 Chr.1: 85775657-85775660 on Build GRCh38
rs528451622 Chr.1: 85775716-85775716 on Build GRCh38
rs912799718 Chr.1: 85775897-85775897 on Build GRCh38
rs758039551 Chr.1: 85775723-85775723 on Build GRCh38
rs1008031209 Chr.1: 85776004-85776004 on Build GRCh38
rs756767744 Chr.1: 85775736-85775736 on Build GRCh38
rs141767705 Chr.1: 85775716-85775717 on Build GRCh38
rs567463385 Chr.1: 85775975-85775975 on Build GRCh38
rs34471406 Chr.1: 85775933-85775933 on Build GRCh38
rs573113208 Chr.1: 85775779-85775779 on Build GRCh38
rs745370513 Chr.1: 85775671-85775672 on Build GRCh38
rs980491222 Chr.1: 85776058-85776058 on Build GRCh38
rs114744777 Chr.1: 85775803-85775803 on Build GRCh38
rs961443862 Chr.1: 85776010-85776010 on Build GRCh38
rs995516756 Chr.1: 85776021-85776021 on Build GRCh38
rs924237877 Chr.1: 85775833-85775833 on Build GRCh38
rs766320753 Chr.1: 85775691-85775691 on Build GRCh38
rs752342436 Chr.1: 85775720-85775720 on Build GRCh38
rs775227290 Chr.1: 85775987-85775987 on Build GRCh38
rs915676845 Chr.1: 85775788-85775788 on Build GRCh38
rs539689255 Chr.1: 85775717-85775718 on Build GRCh38
rs1026945596 Chr.1: 85775648-85775648 on Build GRCh38
rs746231365 Chr.1: 85775628-85775628 on Build GRCh38
rs200383865 Chr.1: 85775696-85775696 on Build GRCh38
rs754761164 Chr.1: 85775709-85775709 on Build GRCh38
rs374908126 Chr.1: 85775633-85775633 on Build GRCh38
rs189294036 Chr.1: 85775981-85775981 on Build GRCh38
rs774412838 Chr.1: 85775679-85775679 on Build GRCh38
rs764204835 Chr.1: 85775651-85775651 on Build GRCh38
rs184513433 Chr.1: 85775719-85775719 on Build GRCh38
rs1039838754 Chr.1: 85775605-85775605 on Build GRCh38
rs978400554 Chr.1: 85775813-85775813 on Build GRCh38

View Full Product Details