Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 114584164-114584725 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DENND2C
Entrez Gene ID
163259
Gene Name
DENN domain containing 2C
Gene Aliases
dJ1156J9.1
UniGene
Hs.654928
Gene Chromosome Location
Chr.1: 114584575-114670111 on Build GRCh38
Transcripts
NM_001256404.1 NM_198459.3

SNP Detail

SNP ID Chromosome Location
rs1015324571 Chr.1: 114584479-114584479 on Build GRCh38
rs1056956668 Chr.1: 114584192-114584192 on Build GRCh38
rs932899564 Chr.1: 114584639-114584639 on Build GRCh38
rs532143971 Chr.1: 114584236-114584240 on Build GRCh38
rs1049073119 Chr.1: 114584422-114584422 on Build GRCh38
rs143217253 Chr.1: 114584261-114584261 on Build GRCh38
rs148335629 Chr.1: 114584333-114584333 on Build GRCh38
rs1056987639 Chr.1: 114584352-114584352 on Build GRCh38
rs774274575 Chr.1: 114584358-114584358 on Build GRCh38
rs944011037 Chr.1: 114584383-114584383 on Build GRCh38
rs146372952 Chr.1: 114584651-114584651 on Build GRCh38
rs571126922 Chr.1: 114584604-114584604 on Build GRCh38
rs768480481 Chr.1: 114584495-114584495 on Build GRCh38
rs1017086381 Chr.1: 114584465-114584466 on Build GRCh38
rs774741856 Chr.1: 114584334-114584334 on Build GRCh38
rs1041009456 Chr.1: 114584396-114584396 on Build GRCh38
rs185068374 Chr.1: 114584374-114584374 on Build GRCh38
rs1015272043 Chr.1: 114584288-114584288 on Build GRCh38
rs924631988 Chr.1: 114584375-114584375 on Build GRCh38
rs149563733 Chr.1: 114584204-114584204 on Build GRCh38
rs970358390 Chr.1: 114584292-114584292 on Build GRCh38
rs926981527 Chr.1: 114584340-114584340 on Build GRCh38
rs746384674 Chr.1: 114584357-114584357 on Build GRCh38
rs1042882377 Chr.1: 114584364-114584364 on Build GRCh38
rs966491807 Chr.1: 114584558-114584558 on Build GRCh38
rs1007330120 Chr.1: 114584254-114584256 on Build GRCh38
rs1033350135 Chr.1: 114584507-114584507 on Build GRCh38
rs951656168 Chr.1: 114584452-114584452 on Build GRCh38
rs558515803 Chr.1: 114584302-114584302 on Build GRCh38
rs776435322 Chr.1: 114584526-114584526 on Build GRCh38
rs182457108 Chr.1: 114584231-114584231 on Build GRCh38
rs141467047 Chr.1: 114584630-114584630 on Build GRCh38
rs965346871 Chr.1: 114584261-114584273 on Build GRCh38
rs572069958 Chr.1: 114584660-114584660 on Build GRCh38
rs900106167 Chr.1: 114584397-114584397 on Build GRCh38
rs1048617174 Chr.1: 114584640-114584640 on Build GRCh38
rs1025867364 Chr.1: 114584262-114584262 on Build GRCh38
rs908991084 Chr.1: 114584281-114584282 on Build GRCh38
rs984303758 Chr.1: 114584274-114584278 on Build GRCh38
rs900010522 Chr.1: 114584211-114584211 on Build GRCh38
rs1033626118 Chr.1: 114584332-114584332 on Build GRCh38
rs913178229 Chr.1: 114584555-114584555 on Build GRCh38
rs1027136009 Chr.1: 114584258-114584258 on Build GRCh38
rs940201173 Chr.1: 114584688-114584688 on Build GRCh38
rs989381324 Chr.1: 114584528-114584528 on Build GRCh38
rs1018256931 Chr.1: 114584255-114584261 on Build GRCh38
rs976760274 Chr.1: 114584622-114584622 on Build GRCh38
rs760260007 Chr.1: 114584418-114584418 on Build GRCh38
rs369756608 Chr.1: 114584360-114584360 on Build GRCh38
rs1022164619 Chr.1: 114584232-114584232 on Build GRCh38
rs938584633 Chr.1: 114584190-114584190 on Build GRCh38
rs558844238 Chr.1: 114584641-114584641 on Build GRCh38
rs904627506 Chr.1: 114584373-114584373 on Build GRCh38
rs181698563 Chr.1: 114584459-114584459 on Build GRCh38
rs538924555 Chr.1: 114584266-114584266 on Build GRCh38
rs1022549040 Chr.1: 114584556-114584556 on Build GRCh38
rs977488198 Chr.1: 114584593-114584593 on Build GRCh38
rs222494 Chr.1: 114584438-114584438 on Build GRCh38
rs944107249 Chr.1: 114584614-114584614 on Build GRCh38
rs989761608 Chr.1: 114584343-114584343 on Build GRCh38
rs959587805 Chr.1: 114584522-114584522 on Build GRCh38
rs566426184 Chr.1: 114584327-114584327 on Build GRCh38
rs906270435 Chr.1: 114584485-114584485 on Build GRCh38
rs992444080 Chr.1: 114584539-114584539 on Build GRCh38

View Full Product Details