Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 406595-407142 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXO25
Entrez Gene ID
26260
Gene Name
F-box protein 25
Gene Aliases
FBX25
UniGene
Hs.438454
Gene Chromosome Location
Chr.8: 406808-477967 on Build GRCh38
Transcripts
NM_012173.3 NM_183420.1 NM_183421.1 XM_017013313.1 XM_017013314.1

SNP Detail

SNP ID Chromosome Location
rs893918428 Chr.8: 407119-407119 on Build GRCh38
rs575697669 Chr.8: 406642-406642 on Build GRCh38
rs918750774 Chr.8: 406810-406810 on Build GRCh38
rs544725046 Chr.8: 406673-406673 on Build GRCh38
rs897384981 Chr.8: 406620-406620 on Build GRCh38
rs7011258 Chr.8: 406775-406775 on Build GRCh38
rs368089137 Chr.8: 406745-406745 on Build GRCh38
rs916747192 Chr.8: 407070-407070 on Build GRCh38
rs928194092 Chr.8: 407078-407078 on Build GRCh38
rs943753165 Chr.8: 406803-406803 on Build GRCh38
rs563696962 Chr.8: 406768-406768 on Build GRCh38
rs1049943605 Chr.8: 406704-406704 on Build GRCh38
rs985556169 Chr.8: 406874-406874 on Build GRCh38
rs1037192509 Chr.8: 406711-406711 on Build GRCh38
rs556013908 Chr.8: 406775-406776 on Build GRCh38
rs906549754 Chr.8: 406958-406958 on Build GRCh38
rs561233562 Chr.8: 406631-406631 on Build GRCh38
rs1019916646 Chr.8: 407008-407008 on Build GRCh38
rs1025492027 Chr.8: 406751-406751 on Build GRCh38
rs543603967 Chr.8: 406791-406791 on Build GRCh38
rs1037243119 Chr.8: 406804-406804 on Build GRCh38
rs1046863708 Chr.8: 406943-406943 on Build GRCh38
rs991305564 Chr.8: 406644-406644 on Build GRCh38
rs912331775 Chr.8: 406802-406802 on Build GRCh38
rs566207209 Chr.8: 406798-406798 on Build GRCh38
rs1025427176 Chr.8: 407042-407042 on Build GRCh38
rs972485454 Chr.8: 406883-406883 on Build GRCh38
rs1052375018 Chr.8: 407098-407098 on Build GRCh38
rs532457025 Chr.8: 406854-406854 on Build GRCh38
rs530421565 Chr.8: 406741-406741 on Build GRCh38
rs955873952 Chr.8: 406856-406856 on Build GRCh38
rs536563206 Chr.8: 406853-406853 on Build GRCh38
rs569287206 Chr.8: 406940-406940 on Build GRCh38
rs952420524 Chr.8: 407046-407046 on Build GRCh38
rs555152262 Chr.8: 407022-407022 on Build GRCh38
rs1021442753 Chr.8: 406977-406977 on Build GRCh38
rs567407435 Chr.8: 406850-406850 on Build GRCh38
rs944218359 Chr.8: 406710-406710 on Build GRCh38
rs867885372 Chr.8: 406780-406780 on Build GRCh38
rs1349057 Chr.8: 406616-406616 on Build GRCh38
rs866836167 Chr.8: 406754-406754 on Build GRCh38
rs746580732 Chr.8: 406878-406878 on Build GRCh38
rs981263887 Chr.8: 406772-406773 on Build GRCh38
rs954141387 Chr.8: 406989-406989 on Build GRCh38
rs13279681 Chr.8: 406811-406811 on Build GRCh38
rs948328155 Chr.8: 406939-406939 on Build GRCh38
rs1025025628 Chr.8: 406663-406663 on Build GRCh38
rs542926220 Chr.8: 406764-406765 on Build GRCh38
rs866120267 Chr.8: 407103-407103 on Build GRCh38
rs931488547 Chr.8: 406893-406893 on Build GRCh38
rs552690745 Chr.8: 406796-406796 on Build GRCh38
rs1012210582 Chr.8: 406747-406747 on Build GRCh38
rs1052488896 Chr.8: 406965-406965 on Build GRCh38
rs1011339639 Chr.8: 406861-406861 on Build GRCh38
rs13279121 Chr.8: 406619-406619 on Build GRCh38
rs971654546 Chr.8: 406672-406672 on Build GRCh38
rs970823743 Chr.8: 406767-406768 on Build GRCh38
rs990032372 Chr.8: 406787-406787 on Build GRCh38
rs1349059 Chr.8: 406628-406628 on Build GRCh38
rs3840686 Chr.8: 406754-406755 on Build GRCh38
rs955988300 Chr.8: 406781-406781 on Build GRCh38
rs527492082 Chr.8: 406806-406806 on Build GRCh38
rs989811708 Chr.8: 406695-406695 on Build GRCh38
rs935345159 Chr.8: 407085-407085 on Build GRCh38
rs541216023 Chr.8: 407005-407005 on Build GRCh38
rs895147538 Chr.8: 406740-406740 on Build GRCh38
rs1026969835 Chr.8: 406639-406639 on Build GRCh38
rs931436247 Chr.8: 406819-406819 on Build GRCh38
rs541648375 Chr.8: 406713-406713 on Build GRCh38
rs965158880 Chr.8: 406879-406879 on Build GRCh38
rs544109333 Chr.8: 407038-407038 on Build GRCh38
rs953975882 Chr.8: 406865-406865 on Build GRCh38
rs532543748 Chr.8: 407076-407076 on Build GRCh38
rs924560044 Chr.8: 406680-406680 on Build GRCh38
rs796089268 Chr.8: 406730-406730 on Build GRCh38
rs564666131 Chr.8: 406686-406686 on Build GRCh38
rs572150003 Chr.8: 406975-406975 on Build GRCh38
rs1349058 Chr.8: 406618-406618 on Build GRCh38
rs561417815 Chr.8: 406726-406726 on Build GRCh38
rs7844398 Chr.8: 406703-406703 on Build GRCh38
rs575006429 Chr.8: 407027-407027 on Build GRCh38
rs984734285 Chr.8: 407053-407053 on Build GRCh38
rs550167224 Chr.8: 406764-406764 on Build GRCh38
rs916840977 Chr.8: 406930-406930 on Build GRCh38
rs918301902 Chr.8: 406892-406892 on Build GRCh38
rs890857769 Chr.8: 406707-406707 on Build GRCh38
rs564007075 Chr.8: 407051-407051 on Build GRCh38
rs185171001 Chr.8: 406963-406963 on Build GRCh38
rs1049124489 Chr.8: 406848-406848 on Build GRCh38
rs558219775 Chr.8: 406972-406972 on Build GRCh38
rs57766499 Chr.8: 406779-406780 on Build GRCh38
rs1021975461 Chr.8: 406863-406863 on Build GRCh38
rs948182859 Chr.8: 407073-407073 on Build GRCh38
rs985116835 Chr.8: 406797-406797 on Build GRCh38
rs1006799904 Chr.8: 406991-406991 on Build GRCh38
rs868659151 Chr.8: 406772-406772 on Build GRCh38
rs995756063 Chr.8: 406727-406727 on Build GRCh38
rs914212411 Chr.8: 406696-406696 on Build GRCh38
rs990407288 Chr.8: 406903-406903 on Build GRCh38

View Full Product Details