Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 412932-413529 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXO25
Entrez Gene ID
26260
Gene Name
F-box protein 25
Gene Aliases
FBX25
UniGene
Hs.438454
Gene Chromosome Location
Chr.8: 406808-477967 on Build GRCh38
Transcripts
NM_012173.3 NM_183420.1 NM_183421.1 XM_011534748.2 XM_011534749.1 XM_017013313.1 XM_017013314.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1099403 c.75T>C p.C25C Chr.8: 413154-413154 on Build GRCh38
4823885 c.62G>T p.G21V Chr.8: 413141-413141 on Build GRCh38
5574629 c.106A>G p.N36D Chr.8: 413185-413185 on Build GRCh38
327995 c.121_122delAT p.I41fs*8 Chr.8: 413200-413201 on Build GRCh38
3731751 c.15T>C p.G5G Chr.8: 413094-413094 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs561687119 Chr.8: 413369-413369 on Build GRCh38
rs771042619 Chr.8: 413217-413217 on Build GRCh38
rs767600529 Chr.8: 413057-413057 on Build GRCh38
rs760862328 Chr.8: 413138-413138 on Build GRCh38
rs553257241 Chr.8: 413298-413298 on Build GRCh38
rs1028835805 Chr.8: 413500-413500 on Build GRCh38
rs899360385 Chr.8: 413309-413309 on Build GRCh38
rs764951878 Chr.8: 413170-413170 on Build GRCh38
rs17665340 Chr.8: 413185-413185 on Build GRCh38
rs182232826 Chr.8: 413321-413321 on Build GRCh38
rs989298480 Chr.8: 413376-413376 on Build GRCh38
rs777423245 Chr.8: 413097-413097 on Build GRCh38
rs757921735 Chr.8: 413093-413093 on Build GRCh38
rs376724327 Chr.8: 413111-413111 on Build GRCh38
rs191925864 Chr.8: 412997-412997 on Build GRCh38
rs766037228 Chr.8: 413060-413060 on Build GRCh38
rs894595964 Chr.8: 413452-413452 on Build GRCh38
rs183716836 Chr.8: 413021-413021 on Build GRCh38
rs893216067 Chr.8: 413487-413487 on Build GRCh38
rs566868481 Chr.8: 413081-413081 on Build GRCh38
rs1051428540 Chr.8: 413268-413268 on Build GRCh38
rs377460563 Chr.8: 413181-413181 on Build GRCh38
rs770246772 Chr.8: 413029-413029 on Build GRCh38
rs777402645 Chr.8: 413177-413177 on Build GRCh38
rs535410177 Chr.8: 413102-413102 on Build GRCh38
rs776850865 Chr.8: 413232-413232 on Build GRCh38
rs752611656 Chr.8: 413172-413172 on Build GRCh38
rs773300956 Chr.8: 413134-413134 on Build GRCh38
rs745733996 Chr.8: 413283-413283 on Build GRCh38
rs529557014 Chr.8: 413475-413475 on Build GRCh38
rs143072661 Chr.8: 413032-413032 on Build GRCh38
rs533358488 Chr.8: 413440-413440 on Build GRCh38
rs374817871 Chr.8: 413053-413053 on Build GRCh38
rs575338146 Chr.8: 413403-413403 on Build GRCh38
rs192059386 Chr.8: 413296-413296 on Build GRCh38
rs182679161 Chr.8: 413442-413442 on Build GRCh38
rs1055966001 Chr.8: 413451-413451 on Build GRCh38
rs756859801 Chr.8: 413184-413184 on Build GRCh38
rs761939382 Chr.8: 413056-413056 on Build GRCh38
rs371194282 Chr.8: 413256-413256 on Build GRCh38
rs752591305 Chr.8: 413253-413253 on Build GRCh38
rs71202618 Chr.8: 413002-413003 on Build GRCh38
rs373203480 Chr.8: 413173-413173 on Build GRCh38
rs770759492 Chr.8: 413110-413110 on Build GRCh38
rs529618908 Chr.8: 413404-413404 on Build GRCh38
rs1011193632 Chr.8: 413496-413496 on Build GRCh38
rs561727286 Chr.8: 412999-412999 on Build GRCh38
rs773243949 Chr.8: 413207-413207 on Build GRCh38
rs372681706 Chr.8: 413121-413121 on Build GRCh38
rs533607843 Chr.8: 413058-413058 on Build GRCh38
rs930847729 Chr.8: 413444-413444 on Build GRCh38
rs186942153 Chr.8: 413354-413354 on Build GRCh38
rs10090550 Chr.8: 413192-413192 on Build GRCh38
rs890196692 Chr.8: 413044-413044 on Build GRCh38
rs1046375666 Chr.8: 413456-413456 on Build GRCh38
rs546967901 Chr.8: 413068-413068 on Build GRCh38
rs756734462 Chr.8: 413264-413264 on Build GRCh38
rs762314032 Chr.8: 413130-413130 on Build GRCh38
rs998836005 Chr.8: 413473-413473 on Build GRCh38
rs936573716 Chr.8: 412990-412990 on Build GRCh38
rs751846738 Chr.8: 413324-413324 on Build GRCh38
rs543953997 Chr.8: 413420-413420 on Build GRCh38
rs5000141 Chr.8: 413150-413150 on Build GRCh38
rs767545803 Chr.8: 413133-413133 on Build GRCh38
rs994947900 Chr.8: 413315-413315 on Build GRCh38
rs780710927 Chr.8: 413162-413162 on Build GRCh38
rs868329929 Chr.8: 413342-413342 on Build GRCh38
rs957878973 Chr.8: 413373-413373 on Build GRCh38
rs995065440 Chr.8: 413103-413103 on Build GRCh38
rs772614978 Chr.8: 413206-413206 on Build GRCh38
rs71223333 Chr.8: 412999-413000 on Build GRCh38
rs765038708 Chr.8: 413242-413242 on Build GRCh38
rs745520522 Chr.8: 413191-413191 on Build GRCh38
rs191762550 Chr.8: 413436-413436 on Build GRCh38
rs369217867 Chr.8: 413115-413115 on Build GRCh38
rs950337047 Chr.8: 413454-413454 on Build GRCh38
rs764198839 Chr.8: 413257-413257 on Build GRCh38
rs911576325 Chr.8: 413422-413422 on Build GRCh38
rs763351710 Chr.8: 413034-413034 on Build GRCh38
rs1036043206 Chr.8: 413308-413308 on Build GRCh38
rs764705522 Chr.8: 413037-413037 on Build GRCh38
rs759680142 Chr.8: 413237-413237 on Build GRCh38
rs774864422 Chr.8: 413129-413129 on Build GRCh38
rs879340184 Chr.8: 413407-413407 on Build GRCh38
rs747644040 Chr.8: 413085-413085 on Build GRCh38
rs773441077 Chr.8: 413190-413190 on Build GRCh38
rs749731980 Chr.8: 413120-413120 on Build GRCh38
rs10110960 Chr.8: 413465-413465 on Build GRCh38
rs751105125 Chr.8: 413260-413260 on Build GRCh38
rs369423216 Chr.8: 413165-413165 on Build GRCh38
rs753567178 Chr.8: 413158-413158 on Build GRCh38
rs998952299 Chr.8: 413361-413361 on Build GRCh38
rs558268047 Chr.8: 413266-413266 on Build GRCh38
rs779661777 Chr.8: 413355-413355 on Build GRCh38
rs187570858 Chr.8: 413273-413273 on Build GRCh38
rs753716116 Chr.8: 413067-413067 on Build GRCh38
rs931667589 Chr.8: 413230-413230 on Build GRCh38

View Full Product Details