Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 23258755-23259154 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHMP7
Entrez Gene ID
91782
Gene Name
charged multivesicular body protein 7
Gene Aliases
-
UniGene
Hs.5019
Gene Chromosome Location
Chr.8: 23243637-23262000 on Build GRCh38
Transcripts
NM_001317899.1 NM_152272.4 XM_006716415.2 XM_017013961.1 XM_017013962.1 XM_017013963.1 XM_017013964.1 XM_017013965.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6628493 c.993A>T p.A331A Chr.8: 23258764-23258764 on Build GRCh38
6189345 c.1089G>T p.Q363H Chr.8: 23259095-23259095 on Build GRCh38
7268312 c.1016T>C p.V339A Chr.8: 23258787-23258787 on Build GRCh38
454325 c.1048C>G p.Q350E Chr.8: 23258819-23258819 on Build GRCh38
1098237 c.1074G>A p.Q358Q Chr.8: 23259080-23259080 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777937942 Chr.8: 23258859-23258859 on Build GRCh38
rs963462040 Chr.8: 23258974-23258974 on Build GRCh38
rs200476187 Chr.8: 23258789-23258789 on Build GRCh38
rs386723547 Chr.8: 23258916-23258918 on Build GRCh38
rs766203467 Chr.8: 23258834-23258834 on Build GRCh38
rs533496712 Chr.8: 23259031-23259031 on Build GRCh38
rs761352734 Chr.8: 23258818-23258818 on Build GRCh38
rs765475464 Chr.8: 23258888-23258888 on Build GRCh38
rs751480195 Chr.8: 23259057-23259057 on Build GRCh38
rs571576988 Chr.8: 23258842-23258842 on Build GRCh38
rs544753592 Chr.8: 23258909-23258909 on Build GRCh38
rs1048442384 Chr.8: 23258913-23258913 on Build GRCh38
rs4581063 Chr.8: 23258879-23258879 on Build GRCh38
rs572610523 Chr.8: 23258827-23258827 on Build GRCh38
rs146133000 Chr.8: 23258971-23258971 on Build GRCh38
rs376259485 Chr.8: 23259044-23259044 on Build GRCh38
rs565938159 Chr.8: 23259009-23259009 on Build GRCh38
rs573949335 Chr.8: 23258809-23258809 on Build GRCh38
rs767289175 Chr.8: 23259097-23259097 on Build GRCh38
rs762434567 Chr.8: 23259049-23259049 on Build GRCh38
rs1042579946 Chr.8: 23258902-23258904 on Build GRCh38
rs111259946 Chr.8: 23258914-23258914 on Build GRCh38
rs779460830 Chr.8: 23258882-23258882 on Build GRCh38
rs945491894 Chr.8: 23258885-23258885 on Build GRCh38
rs764686668 Chr.8: 23258824-23258824 on Build GRCh38
rs776169955 Chr.8: 23259034-23259034 on Build GRCh38
rs781321305 Chr.8: 23258847-23258847 on Build GRCh38
rs761415397 Chr.8: 23259040-23259040 on Build GRCh38
rs997026881 Chr.8: 23258983-23258983 on Build GRCh38
rs563511724 Chr.8: 23258910-23258910 on Build GRCh38
rs943430344 Chr.8: 23258841-23258841 on Build GRCh38
rs958193807 Chr.8: 23258996-23258996 on Build GRCh38
rs201788600 Chr.8: 23258918-23258919 on Build GRCh38
rs772461741 Chr.8: 23259033-23259033 on Build GRCh38
rs981754373 Chr.8: 23259090-23259090 on Build GRCh38
rs75354577 Chr.8: 23258975-23258975 on Build GRCh38
rs4478582 Chr.8: 23258916-23258916 on Build GRCh38
rs189131066 Chr.8: 23258972-23258972 on Build GRCh38
rs937932848 Chr.8: 23258863-23258863 on Build GRCh38
rs757471282 Chr.8: 23258862-23258864 on Build GRCh38
rs966369156 Chr.8: 23258821-23258821 on Build GRCh38
rs1011499546 Chr.8: 23259026-23259026 on Build GRCh38
rs752940634 Chr.8: 23258848-23258848 on Build GRCh38
rs930488332 Chr.8: 23258912-23258912 on Build GRCh38
rs184597704 Chr.8: 23258948-23258948 on Build GRCh38
rs772649953 Chr.8: 23259046-23259046 on Build GRCh38
rs557929592 Chr.8: 23258884-23258884 on Build GRCh38
rs750382739 Chr.8: 23258825-23258825 on Build GRCh38
rs1028550822 Chr.8: 23258992-23258992 on Build GRCh38
rs551599688 Chr.8: 23259109-23259109 on Build GRCh38
rs751907901 Chr.8: 23258854-23258855 on Build GRCh38
rs964710441 Chr.8: 23259091-23259091 on Build GRCh38
rs369277278 Chr.8: 23259050-23259050 on Build GRCh38
rs774509663 Chr.8: 23258788-23258788 on Build GRCh38
rs374534208 Chr.8: 23258854-23258854 on Build GRCh38
rs35799523 Chr.8: 23258812-23258812 on Build GRCh38
rs749814745 Chr.8: 23258871-23258871 on Build GRCh38
rs987424531 Chr.8: 23258784-23258784 on Build GRCh38
rs771432263 Chr.8: 23258778-23258778 on Build GRCh38
rs4872098 Chr.8: 23258918-23258918 on Build GRCh38
rs781337050 Chr.8: 23258878-23258879 on Build GRCh38

View Full Product Details