Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 87920201-87920750 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC497256
Entrez Gene ID
497256
Gene Name
uncharacterized LOC497256
Gene Aliases
-
UniGene
Hs.584108
Gene Chromosome Location
Chr.9: 87844517-87939244 on Build GRCh38
Transcripts
NM_001145124.1 XM_011518701.2 XM_011518702.1 XM_011518703.1

Gene Symbol
SPATA31C1
Entrez Gene ID
441452
Gene Name
SPATA31 subfamily C member 1
Gene Aliases
FAM75C1, SPATA31C2
UniGene
Hs.652956
Gene Chromosome Location
Chr.9: 87913936-87923657 on Build GRCh38
Transcripts
NM_001145124.1 XM_011518701.2 XM_011518702.1 XM_011518703.1

SNP Detail

SNP ID Chromosome Location
rs755476896 Chr.9: 87920315-87920315 on Build GRCh38
rs779019999 Chr.9: 87920283-87920283 on Build GRCh38
rs185071524 Chr.9: 87920249-87920249 on Build GRCh38
rs757119727 Chr.9: 87920600-87920600 on Build GRCh38
rs538426341 Chr.9: 87920409-87920409 on Build GRCh38
rs775325068 Chr.9: 87920329-87920329 on Build GRCh38
rs185881132 Chr.9: 87920627-87920627 on Build GRCh38
rs754234070 Chr.9: 87920512-87920512 on Build GRCh38
rs562949328 Chr.9: 87920712-87920712 on Build GRCh38
rs746618972 Chr.9: 87920618-87920618 on Build GRCh38
rs753844256 Chr.9: 87920599-87920599 on Build GRCh38
rs749552878 Chr.9: 87920368-87920368 on Build GRCh38
rs753134578 Chr.9: 87920360-87920360 on Build GRCh38
rs748149136 Chr.9: 87920576-87920576 on Build GRCh38
rs778158521 Chr.9: 87920699-87920699 on Build GRCh38
rs576431058 Chr.9: 87920484-87920484 on Build GRCh38
rs539994715 Chr.9: 87920395-87920395 on Build GRCh38
rs761619753 Chr.9: 87920298-87920298 on Build GRCh38
rs759060618 Chr.9: 87920482-87920482 on Build GRCh38
rs370518734 Chr.9: 87920364-87920364 on Build GRCh38
rs369763880 Chr.9: 87920318-87920318 on Build GRCh38
rs397833502 Chr.9: 87920597-87920597 on Build GRCh38
rs776349774 Chr.9: 87920339-87920339 on Build GRCh38
rs748132752 Chr.9: 87920458-87920458 on Build GRCh38
rs769980775 Chr.9: 87920236-87920236 on Build GRCh38
rs755494100 Chr.9: 87920358-87920358 on Build GRCh38
rs776184190 Chr.9: 87920275-87920275 on Build GRCh38
rs748734176 Chr.9: 87920623-87920623 on Build GRCh38
rs3119583 Chr.9: 87920375-87920375 on Build GRCh38
rs775313165 Chr.9: 87920258-87920258 on Build GRCh38
rs375322331 Chr.9: 87920471-87920471 on Build GRCh38
rs775713417 Chr.9: 87920721-87920721 on Build GRCh38
rs1005962830 Chr.9: 87920352-87920352 on Build GRCh38
rs371519082 Chr.9: 87920657-87920657 on Build GRCh38
rs923114567 Chr.9: 87920280-87920280 on Build GRCh38
rs750692988 Chr.9: 87920523-87920523 on Build GRCh38
rs370980409 Chr.9: 87920690-87920690 on Build GRCh38
rs757490383 Chr.9: 87920438-87920438 on Build GRCh38
rs188709636 Chr.9: 87920340-87920340 on Build GRCh38
rs750033682 Chr.9: 87920308-87920308 on Build GRCh38
rs569414999 Chr.9: 87920408-87920408 on Build GRCh38
rs572265381 Chr.9: 87920590-87920590 on Build GRCh38
rs760621475 Chr.9: 87920267-87920267 on Build GRCh38
rs548526723 Chr.9: 87920415-87920415 on Build GRCh38
rs191720172 Chr.9: 87920726-87920726 on Build GRCh38
rs768636892 Chr.9: 87920228-87920228 on Build GRCh38
rs763554208 Chr.9: 87920271-87920271 on Build GRCh38
rs750388759 Chr.9: 87920725-87920725 on Build GRCh38
rs761140136 Chr.9: 87920427-87920427 on Build GRCh38
rs779772861 Chr.9: 87920371-87920371 on Build GRCh38
rs1031825152 Chr.9: 87920505-87920505 on Build GRCh38
rs527614246 Chr.9: 87920337-87920337 on Build GRCh38
rs775901033 Chr.9: 87920562-87920562 on Build GRCh38
rs768954693 Chr.9: 87920567-87920567 on Build GRCh38
rs751928334 Chr.9: 87920351-87920351 on Build GRCh38
rs397833503 Chr.9: 87920528-87920528 on Build GRCh38
rs781175504 Chr.9: 87920316-87920316 on Build GRCh38
rs751248819 Chr.9: 87920676-87920676 on Build GRCh38
rs747487515 Chr.9: 87920226-87920226 on Build GRCh38
rs772179220 Chr.9: 87920718-87920718 on Build GRCh38
rs933118475 Chr.9: 87920307-87920307 on Build GRCh38
rs765030527 Chr.9: 87920723-87920723 on Build GRCh38
rs756393140 Chr.9: 87920319-87920319 on Build GRCh38
rs749765765 Chr.9: 87920709-87920709 on Build GRCh38
rs763321614 Chr.9: 87920587-87920587 on Build GRCh38
rs776834336 Chr.9: 87920622-87920622 on Build GRCh38
rs778543514 Chr.9: 87920602-87920602 on Build GRCh38
rs773017326 Chr.9: 87920349-87920349 on Build GRCh38
rs753902869 Chr.9: 87920434-87920434 on Build GRCh38
rs778246337 Chr.9: 87920538-87920538 on Build GRCh38
rs771832366 Chr.9: 87920252-87920252 on Build GRCh38
rs755985498 Chr.9: 87920595-87920595 on Build GRCh38
rs781289397 Chr.9: 87920229-87920229 on Build GRCh38
rs777778335 Chr.9: 87920639-87920640 on Build GRCh38
rs771961697 Chr.9: 87920617-87920619 on Build GRCh38
rs769828893 Chr.9: 87920459-87920459 on Build GRCh38
rs751763579 Chr.9: 87920594-87920594 on Build GRCh38
rs765185434 Chr.9: 87920521-87920521 on Build GRCh38
rs759896142 Chr.9: 87920713-87920713 on Build GRCh38
rs770278460 Chr.9: 87920480-87920480 on Build GRCh38
rs772814322 Chr.9: 87920396-87920396 on Build GRCh38
rs577427444 Chr.9: 87920322-87920322 on Build GRCh38
rs756198990 Chr.9: 87920363-87920363 on Build GRCh38
rs560034208 Chr.9: 87920631-87920631 on Build GRCh38
rs758041873 Chr.9: 87920503-87920503 on Build GRCh38
rs763990033 Chr.9: 87920656-87920656 on Build GRCh38
rs764787506 Chr.9: 87920343-87920343 on Build GRCh38
rs947700814 Chr.9: 87920638-87920638 on Build GRCh38
rs777975408 Chr.9: 87920321-87920321 on Build GRCh38
rs764979781 Chr.9: 87920306-87920306 on Build GRCh38
rs142470040 Chr.9: 87920359-87920359 on Build GRCh38
rs779496659 Chr.9: 87920672-87920672 on Build GRCh38
rs971555515 Chr.9: 87920468-87920468 on Build GRCh38
rs774738656 Chr.9: 87920648-87920648 on Build GRCh38
rs749120544 Chr.9: 87920535-87920535 on Build GRCh38
rs397832697 Chr.9: 87920527-87920527 on Build GRCh38
rs199983150 Chr.9: 87920564-87920564 on Build GRCh38
rs181014483 Chr.9: 87920232-87920232 on Build GRCh38
rs990582455 Chr.9: 87920227-87920227 on Build GRCh38
rs181056382 Chr.9: 87920386-87920386 on Build GRCh38
rs771700492 Chr.9: 87920545-87920545 on Build GRCh38
rs568708324 Chr.9: 87920457-87920457 on Build GRCh38
rs745584148 Chr.9: 87920541-87920541 on Build GRCh38
rs540972709 Chr.9: 87920608-87920608 on Build GRCh38
rs754569215 Chr.9: 87920686-87920686 on Build GRCh38
rs746818688 Chr.9: 87920373-87920373 on Build GRCh38
rs1032294668 Chr.9: 87920683-87920683 on Build GRCh38
rs750557255 Chr.9: 87920445-87920445 on Build GRCh38
rs757923408 Chr.9: 87920671-87920671 on Build GRCh38
rs368844699 Chr.9: 87920632-87920632 on Build GRCh38
rs757839453 Chr.9: 87920606-87920606 on Build GRCh38
rs377360948 Chr.9: 87920547-87920547 on Build GRCh38
rs534239537 Chr.9: 87920470-87920470 on Build GRCh38
rs754828021 Chr.9: 87920451-87920451 on Build GRCh38
rs765089151 Chr.9: 87920669-87920669 on Build GRCh38
rs745367524 Chr.9: 87920605-87920605 on Build GRCh38
rs377054909 Chr.9: 87920309-87920309 on Build GRCh38
rs781665194 Chr.9: 87920689-87920689 on Build GRCh38
rs781178940 Chr.9: 87920526-87920526 on Build GRCh38
rs865992927 Chr.9: 87920384-87920384 on Build GRCh38
rs756813882 Chr.9: 87920537-87920537 on Build GRCh38
rs638565 Chr.9: 87920573-87920573 on Build GRCh38
rs564452741 Chr.9: 87920655-87920655 on Build GRCh38
rs764208668 Chr.9: 87920433-87920433 on Build GRCh38
rs761970323 Chr.9: 87920514-87920514 on Build GRCh38
rs769044592 Chr.9: 87920619-87920619 on Build GRCh38
rs933211853 Chr.9: 87920588-87920588 on Build GRCh38
rs762559564 Chr.9: 87920407-87920407 on Build GRCh38
rs757284768 Chr.9: 87920659-87920659 on Build GRCh38
rs761230036 Chr.9: 87920495-87920495 on Build GRCh38
rs764582660 Chr.9: 87920509-87920509 on Build GRCh38
rs762651593 Chr.9: 87920350-87920350 on Build GRCh38
rs202077057 Chr.9: 87920569-87920569 on Build GRCh38
rs781295129 Chr.9: 87920376-87920376 on Build GRCh38
rs759005325 Chr.9: 87920233-87920234 on Build GRCh38
rs774188022 Chr.9: 87920241-87920241 on Build GRCh38
rs771794931 Chr.9: 87920326-87920326 on Build GRCh38
rs779204293 Chr.9: 87920439-87920439 on Build GRCh38
rs943286396 Chr.9: 87920263-87920263 on Build GRCh38
rs546253951 Chr.9: 87920691-87920691 on Build GRCh38

View Full Product Details