Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 87922714-87923256 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC497256
Entrez Gene ID
497256
Gene Name
uncharacterized LOC497256
Gene Aliases
-
UniGene
Hs.584108
Gene Chromosome Location
Chr.9: 87844517-87939244 on Build GRCh38
Transcripts
NM_001145124.1 XM_011518701.2 XM_011518702.1 XM_011518703.1

Gene Symbol
SPATA31C1
Entrez Gene ID
441452
Gene Name
SPATA31 subfamily C member 1
Gene Aliases
FAM75C1, SPATA31C2
UniGene
Hs.652956
Gene Chromosome Location
Chr.9: 87913936-87923657 on Build GRCh38
Transcripts
NM_001145124.1 XM_011518701.2 XM_011518702.1 XM_011518703.1

SNP Detail

SNP ID Chromosome Location
rs759922870 Chr.9: 87923168-87923168 on Build GRCh38
rs544294092 Chr.9: 87923036-87923036 on Build GRCh38
rs535801745 Chr.9: 87922976-87922976 on Build GRCh38
rs775359118 Chr.9: 87923042-87923042 on Build GRCh38
rs374819681 Chr.9: 87922854-87922854 on Build GRCh38
rs560862305 Chr.9: 87923055-87923055 on Build GRCh38
rs760588266 Chr.9: 87923097-87923097 on Build GRCh38
rs761165911 Chr.9: 87923109-87923109 on Build GRCh38
rs760212436 Chr.9: 87923049-87923049 on Build GRCh38
rs764962511 Chr.9: 87923112-87923112 on Build GRCh38
rs746783081 Chr.9: 87922873-87922874 on Build GRCh38
rs774193171 Chr.9: 87923029-87923029 on Build GRCh38
rs770060204 Chr.9: 87923083-87923083 on Build GRCh38
rs778089347 Chr.9: 87923134-87923134 on Build GRCh38
rs748357277 Chr.9: 87923013-87923013 on Build GRCh38
rs753451889 Chr.9: 87922919-87922919 on Build GRCh38
rs596236 Chr.9: 87923113-87923113 on Build GRCh38
rs746425496 Chr.9: 87922963-87922963 on Build GRCh38
rs779815979 Chr.9: 87923141-87923141 on Build GRCh38
rs551044351 Chr.9: 87923220-87923220 on Build GRCh38
rs757572071 Chr.9: 87922845-87922845 on Build GRCh38
rs542892181 Chr.9: 87922890-87922890 on Build GRCh38
rs757469374 Chr.9: 87923218-87923218 on Build GRCh38
rs529885550 Chr.9: 87923057-87923057 on Build GRCh38
rs751975988 Chr.9: 87923163-87923163 on Build GRCh38
rs775449181 Chr.9: 87923095-87923095 on Build GRCh38
rs759189099 Chr.9: 87923030-87923030 on Build GRCh38
rs755536585 Chr.9: 87923124-87923124 on Build GRCh38
rs752835468 Chr.9: 87923132-87923132 on Build GRCh38
rs747019285 Chr.9: 87923142-87923142 on Build GRCh38
rs776119670 Chr.9: 87923106-87923106 on Build GRCh38
rs756924363 Chr.9: 87923056-87923056 on Build GRCh38
rs777655689 Chr.9: 87922765-87922765 on Build GRCh38
rs771953119 Chr.9: 87923140-87923140 on Build GRCh38
rs764060754 Chr.9: 87923196-87923196 on Build GRCh38
rs780734906 Chr.9: 87923059-87923059 on Build GRCh38
rs546507988 Chr.9: 87923058-87923058 on Build GRCh38
rs777034084 Chr.9: 87922983-87922983 on Build GRCh38
rs780729903 Chr.9: 87922783-87922783 on Build GRCh38
rs776940440 Chr.9: 87922884-87922884 on Build GRCh38
rs769152285 Chr.9: 87923152-87923152 on Build GRCh38
rs9410980 Chr.9: 87922824-87922824 on Build GRCh38
rs879050319 Chr.9: 87922874-87922874 on Build GRCh38
rs367759932 Chr.9: 87922869-87922869 on Build GRCh38
rs902646792 Chr.9: 87922987-87922987 on Build GRCh38
rs918203451 Chr.9: 87922971-87922971 on Build GRCh38
rs530578796 Chr.9: 87922981-87922981 on Build GRCh38
rs763414556 Chr.9: 87923114-87923114 on Build GRCh38
rs512437 Chr.9: 87922941-87922941 on Build GRCh38
rs372224653 Chr.9: 87922888-87922888 on Build GRCh38
rs755564536 Chr.9: 87922905-87922905 on Build GRCh38
rs771727632 Chr.9: 87923094-87923094 on Build GRCh38
rs1006833178 Chr.9: 87922835-87922835 on Build GRCh38
rs756318440 Chr.9: 87923031-87923032 on Build GRCh38
rs551322010 Chr.9: 87923153-87923153 on Build GRCh38
rs748638313 Chr.9: 87923156-87923156 on Build GRCh38
rs374279707 Chr.9: 87923222-87923222 on Build GRCh38
rs752080118 Chr.9: 87923066-87923066 on Build GRCh38
rs749573500 Chr.9: 87923057-87923058 on Build GRCh38
rs752352376 Chr.9: 87922790-87922790 on Build GRCh38
rs769009439 Chr.9: 87922879-87922879 on Build GRCh38
rs202204642 Chr.9: 87923091-87923091 on Build GRCh38
rs766916650 Chr.9: 87923157-87923157 on Build GRCh38
rs770688994 Chr.9: 87923020-87923020 on Build GRCh38
rs774477597 Chr.9: 87923097-87923098 on Build GRCh38
rs781407505 Chr.9: 87923068-87923068 on Build GRCh38
rs768590970 Chr.9: 87923143-87923143 on Build GRCh38
rs571313061 Chr.9: 87923155-87923155 on Build GRCh38
rs557751528 Chr.9: 87922746-87922746 on Build GRCh38
rs762498265 Chr.9: 87923156-87923156 on Build GRCh38
rs756261094 Chr.9: 87923188-87923188 on Build GRCh38
rs512327 Chr.9: 87922977-87922977 on Build GRCh38
rs751473988 Chr.9: 87922759-87922759 on Build GRCh38
rs369542775 Chr.9: 87923105-87923105 on Build GRCh38
rs595689 Chr.9: 87923234-87923234 on Build GRCh38
rs768687079 Chr.9: 87923096-87923097 on Build GRCh38
rs571374231 Chr.9: 87922813-87922813 on Build GRCh38
rs747016576 Chr.9: 87922871-87922871 on Build GRCh38
rs754485312 Chr.9: 87922769-87922769 on Build GRCh38
rs866918527 Chr.9: 87922779-87922779 on Build GRCh38
rs745698976 Chr.9: 87923092-87923092 on Build GRCh38
rs763568771 Chr.9: 87923050-87923050 on Build GRCh38
rs752208303 Chr.9: 87922904-87922904 on Build GRCh38
rs1053847797 Chr.9: 87923021-87923021 on Build GRCh38
rs553613518 Chr.9: 87922891-87922891 on Build GRCh38
rs557226812 Chr.9: 87922858-87922858 on Build GRCh38
rs767003629 Chr.9: 87922895-87922895 on Build GRCh38
rs766826579 Chr.9: 87923117-87923117 on Build GRCh38
rs766249433 Chr.9: 87922752-87922752 on Build GRCh38
rs749443208 Chr.9: 87923138-87923138 on Build GRCh38
rs532290260 Chr.9: 87923133-87923133 on Build GRCh38
rs776498663 Chr.9: 87923149-87923149 on Build GRCh38
rs763762479 Chr.9: 87922909-87922909 on Build GRCh38
rs755761027 Chr.9: 87922794-87922794 on Build GRCh38
rs762774562 Chr.9: 87922747-87922747 on Build GRCh38
rs757661603 Chr.9: 87922932-87922932 on Build GRCh38
rs754151745 Chr.9: 87923215-87923215 on Build GRCh38
rs752170026 Chr.9: 87923118-87923118 on Build GRCh38
rs772095627 Chr.9: 87922865-87922865 on Build GRCh38
rs781737459 Chr.9: 87923125-87923125 on Build GRCh38
rs780160336 Chr.9: 87923039-87923039 on Build GRCh38
rs768163628 Chr.9: 87923173-87923173 on Build GRCh38
rs537097011 Chr.9: 87923178-87923178 on Build GRCh38
rs376807700 Chr.9: 87923071-87923071 on Build GRCh38

View Full Product Details