Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 34723764-34724314 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM205A
Entrez Gene ID
259308
Gene Name
family with sequence similarity 205 member A
Gene Aliases
C9orf144B
UniGene
Hs.522093
Gene Chromosome Location
Chr.9: 34723053-34729538 on Build GRCh38
Transcripts
NM_001141917.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2773508 c.3277C>A p.P1093T Chr.9: 34723963-34723963 on Build GRCh38
2773504 c.3345T>A p.H1115Q Chr.9: 34723895-34723895 on Build GRCh38
7244421 c.3100A>C p.K1034Q Chr.9: 34724140-34724140 on Build GRCh38
4929444 c.3021G>C p.M1007I Chr.9: 34724219-34724219 on Build GRCh38
7230551 c.3109G>C p.D1037H Chr.9: 34724131-34724131 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs12338985 Chr.9: 34723991-34723991 on Build GRCh38
rs548462411 Chr.9: 34724093-34724093 on Build GRCh38
rs764937565 Chr.9: 34724037-34724037 on Build GRCh38
rs562820384 Chr.9: 34724034-34724034 on Build GRCh38
rs769135100 Chr.9: 34724176-34724176 on Build GRCh38
rs930143968 Chr.9: 34723984-34723984 on Build GRCh38
rs757994563 Chr.9: 34723860-34723860 on Build GRCh38
rs372434535 Chr.9: 34724249-34724249 on Build GRCh38
rs768876289 Chr.9: 34723953-34723953 on Build GRCh38
rs777436926 Chr.9: 34724097-34724097 on Build GRCh38
rs988932449 Chr.9: 34723878-34723878 on Build GRCh38
rs762765364 Chr.9: 34724283-34724283 on Build GRCh38
rs922841412 Chr.9: 34724218-34724218 on Build GRCh38
rs201435661 Chr.9: 34724183-34724183 on Build GRCh38
rs472644 Chr.9: 34724062-34724062 on Build GRCh38
rs927004294 Chr.9: 34724253-34724253 on Build GRCh38
rs1035534039 Chr.9: 34723850-34723850 on Build GRCh38
rs750409219 Chr.9: 34723895-34723895 on Build GRCh38
rs199566731 Chr.9: 34724186-34724186 on Build GRCh38
rs867666621 Chr.9: 34724103-34724103 on Build GRCh38
rs951581790 Chr.9: 34724226-34724226 on Build GRCh38
rs772267726 Chr.9: 34723963-34723963 on Build GRCh38
rs1001836326 Chr.9: 34723802-34723802 on Build GRCh38
rs973325537 Chr.9: 34723875-34723889 on Build GRCh38
rs3739881 Chr.9: 34724244-34724244 on Build GRCh38
rs765163019 Chr.9: 34724275-34724275 on Build GRCh38
rs1018288366 Chr.9: 34724184-34724184 on Build GRCh38
rs760787461 Chr.9: 34724010-34724010 on Build GRCh38
rs895693499 Chr.9: 34724106-34724106 on Build GRCh38
rs746467654 Chr.9: 34723880-34723880 on Build GRCh38
rs978769829 Chr.9: 34723916-34723916 on Build GRCh38
rs759462630 Chr.9: 34724036-34724036 on Build GRCh38
rs1037391919 Chr.9: 34724045-34724045 on Build GRCh38
rs568651249 Chr.9: 34724116-34724116 on Build GRCh38
rs777657887 Chr.9: 34724119-34724119 on Build GRCh38
rs539416645 Chr.9: 34724196-34724196 on Build GRCh38
rs2297134 Chr.9: 34723938-34723938 on Build GRCh38
rs775167632 Chr.9: 34724281-34724281 on Build GRCh38
rs753804355 Chr.9: 34724032-34724032 on Build GRCh38
rs556144530 Chr.9: 34724233-34724233 on Build GRCh38
rs372939967 Chr.9: 34724267-34724267 on Build GRCh38
rs761934176 Chr.9: 34723962-34723962 on Build GRCh38
rs982919129 Chr.9: 34724228-34724228 on Build GRCh38
rs540930000 Chr.9: 34723920-34723920 on Build GRCh38
rs922829664 Chr.9: 34723983-34723983 on Build GRCh38
rs879025235 Chr.9: 34723982-34723982 on Build GRCh38
rs759394332 Chr.9: 34724268-34724268 on Build GRCh38
rs62622788 Chr.9: 34724023-34724023 on Build GRCh38
rs199932531 Chr.9: 34724140-34724140 on Build GRCh38
rs749785835 Chr.9: 34724170-34724170 on Build GRCh38
rs532363576 Chr.9: 34723959-34723959 on Build GRCh38
rs759170233 Chr.9: 34724022-34724022 on Build GRCh38
rs555223535 Chr.9: 34724291-34724291 on Build GRCh38
rs534449693 Chr.9: 34724120-34724120 on Build GRCh38
rs977374433 Chr.9: 34723960-34723960 on Build GRCh38
rs1001490272 Chr.9: 34724071-34724071 on Build GRCh38
rs763690164 Chr.9: 34724286-34724286 on Build GRCh38
rs397834353 Chr.9: 34723784-34723784 on Build GRCh38
rs474408 Chr.9: 34723885-34723885 on Build GRCh38
rs777154514 Chr.9: 34723861-34723861 on Build GRCh38
rs766450365 Chr.9: 34724011-34724011 on Build GRCh38

View Full Product Details