Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 35818282-35818759 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM221B
Entrez Gene ID
392307
Gene Name
family with sequence similarity 221 member B
Gene Aliases
C9orf128
UniGene
Hs.128075
Gene Chromosome Location
Chr.9: 35817017-35829148 on Build GRCh38
Transcripts
NM_001012446.3 NR_052026.1 XM_006716768.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4460854 c.1181C>G p.T394S Chr.9: 35818497-35818497 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs779444502 Chr.9: 35818536-35818536 on Build GRCh38
rs904102835 Chr.9: 35818540-35818540 on Build GRCh38
rs767000046 Chr.9: 35818471-35818471 on Build GRCh38
rs1043897543 Chr.9: 35818480-35818480 on Build GRCh38
rs1005115854 Chr.9: 35818679-35818679 on Build GRCh38
rs1047404707 Chr.9: 35818565-35818565 on Build GRCh38
rs531866398 Chr.9: 35818486-35818486 on Build GRCh38
rs997557460 Chr.9: 35818549-35818549 on Build GRCh38
rs60095255 Chr.9: 35818582-35818582 on Build GRCh38
rs1013358154 Chr.9: 35818621-35818621 on Build GRCh38
rs752924318 Chr.9: 35818439-35818439 on Build GRCh38
rs974603661 Chr.9: 35818720-35818720 on Build GRCh38
rs778073111 Chr.9: 35818673-35818673 on Build GRCh38
rs1000561440 Chr.9: 35818575-35818575 on Build GRCh38
rs912733032 Chr.9: 35818491-35818491 on Build GRCh38
rs1024825335 Chr.9: 35818356-35818356 on Build GRCh38
rs369645477 Chr.9: 35818664-35818664 on Build GRCh38
rs748532323 Chr.9: 35818593-35818593 on Build GRCh38
rs886170203 Chr.9: 35818574-35818574 on Build GRCh38
rs79293885 Chr.9: 35818336-35818336 on Build GRCh38
rs574160598 Chr.9: 35818708-35818708 on Build GRCh38
rs1013452555 Chr.9: 35818319-35818319 on Build GRCh38
rs992182181 Chr.9: 35818432-35818432 on Build GRCh38
rs925469578 Chr.9: 35818485-35818485 on Build GRCh38
rs538536469 Chr.9: 35818594-35818594 on Build GRCh38
rs2281646 Chr.9: 35818650-35818650 on Build GRCh38
rs370063671 Chr.9: 35818545-35818545 on Build GRCh38
rs558625509 Chr.9: 35818613-35818613 on Build GRCh38
rs983389244 Chr.9: 35818379-35818379 on Build GRCh38
rs1016074809 Chr.9: 35818303-35818303 on Build GRCh38
rs1016220572 Chr.9: 35818680-35818680 on Build GRCh38
rs190986475 Chr.9: 35818657-35818657 on Build GRCh38
rs150993528 Chr.9: 35818397-35818397 on Build GRCh38
rs747187292 Chr.9: 35818732-35818732 on Build GRCh38
rs945502997 Chr.9: 35818506-35818506 on Build GRCh38
rs909089051 Chr.9: 35818384-35818384 on Build GRCh38
rs950579602 Chr.9: 35818378-35818378 on Build GRCh38
rs969905504 Chr.9: 35818626-35818626 on Build GRCh38
rs557399419 Chr.9: 35818660-35818660 on Build GRCh38
rs375633119 Chr.9: 35818712-35818712 on Build GRCh38
rs372571681 Chr.9: 35818576-35818576 on Build GRCh38
rs372748150 Chr.9: 35818435-35818435 on Build GRCh38
rs959094473 Chr.9: 35818309-35818309 on Build GRCh38
rs375980989 Chr.9: 35818511-35818511 on Build GRCh38
rs1039341796 Chr.9: 35818513-35818513 on Build GRCh38
rs921371816 Chr.9: 35818461-35818461 on Build GRCh38

View Full Product Details