Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 65284626-65285171 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FOXD4L5
Entrez Gene ID
653427
Gene Name
forkhead box D4-like 5
Gene Aliases
bA15J10.2
UniGene
Hs.714683
Gene Chromosome Location
Chr.9: 65282101-65285209 on Build GRCh38
Transcripts
NM_001126334.1

SNP Detail

SNP ID Chromosome Location
rs867870755 Chr.9: 65284823-65284823 on Build GRCh38
rs1007930470 Chr.9: 65284803-65284803 on Build GRCh38
rs1001041499 Chr.9: 65285106-65285106 on Build GRCh38
rs963852212 Chr.9: 65285097-65285097 on Build GRCh38
rs868781435 Chr.9: 65284871-65284871 on Build GRCh38
rs1018344940 Chr.9: 65284854-65284854 on Build GRCh38
rs891381017 Chr.9: 65284693-65284693 on Build GRCh38
rs3000496 Chr.9: 65284644-65284644 on Build GRCh38
rs111389044 Chr.9: 65284887-65284887 on Build GRCh38

View Full Product Details