Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 78246660-78247059 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CEP78
Entrez Gene ID
84131
Gene Name
centrosomal protein 78
Gene Aliases
C9orf81, IP63
UniGene
Hs.374421
Gene Chromosome Location
Chr.9: 78236062-78278431 on Build GRCh38
Transcripts
NM_001098802.1 NM_032171.1 XM_005252263.4 XM_005252265.2 XM_005252266.4 XM_017015192.1 XM_017015193.1 XM_017015194.1 XM_017015195.1 XM_017015196.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5876433 c.824C>T p.A275V Chr.9: 78246714-78246714 on Build GRCh38
3664428 c.835G>A p.A279T Chr.9: 78246725-78246725 on Build GRCh38
4881566 c.811G>T p.E271* Chr.9: 78246701-78246701 on Build GRCh38
3664427 c.835G>A p.A279T Chr.9: 78246725-78246725 on Build GRCh38
4765318 c.876_877insA p.N294fs*5 Chr.9: 78246766-78246767 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs572291046 Chr.9: 78246867-78246868 on Build GRCh38
rs369562785 Chr.9: 78247023-78247023 on Build GRCh38
rs772888220 Chr.9: 78246889-78246889 on Build GRCh38
rs910320278 Chr.9: 78246932-78246932 on Build GRCh38
rs764200008 Chr.9: 78246694-78246694 on Build GRCh38
rs778593372 Chr.9: 78246826-78246826 on Build GRCh38
rs527535007 Chr.9: 78246749-78246749 on Build GRCh38
rs776868804 Chr.9: 78246786-78246786 on Build GRCh38
rs756899539 Chr.9: 78246822-78246822 on Build GRCh38
rs531627492 Chr.9: 78246849-78246849 on Build GRCh38
rs568432558 Chr.9: 78246865-78246865 on Build GRCh38
rs1050438867 Chr.9: 78246928-78246928 on Build GRCh38
rs943196154 Chr.9: 78246950-78246950 on Build GRCh38
rs541626020 Chr.9: 78246724-78246724 on Build GRCh38
rs777280029 Chr.9: 78246818-78246818 on Build GRCh38
rs971672954 Chr.9: 78246841-78246841 on Build GRCh38
rs373047264 Chr.9: 78247031-78247031 on Build GRCh38
rs746297934 Chr.9: 78246760-78246760 on Build GRCh38
rs779953987 Chr.9: 78246752-78246752 on Build GRCh38
rs916983397 Chr.9: 78246696-78246696 on Build GRCh38
rs754854216 Chr.9: 78246796-78246796 on Build GRCh38
rs760639106 Chr.9: 78246773-78246773 on Build GRCh38
rs535963173 Chr.9: 78246890-78246890 on Build GRCh38
rs552100057 Chr.9: 78246799-78246799 on Build GRCh38
rs201356997 Chr.9: 78246688-78246688 on Build GRCh38
rs80012402 Chr.9: 78246863-78246863 on Build GRCh38
rs772442602 Chr.9: 78246765-78246765 on Build GRCh38
rs372819004 Chr.9: 78246790-78246790 on Build GRCh38
rs746208048 Chr.9: 78246829-78246829 on Build GRCh38
rs756028255 Chr.9: 78246800-78246800 on Build GRCh38
rs958872466 Chr.9: 78246783-78246783 on Build GRCh38
rs944741561 Chr.9: 78246961-78246961 on Build GRCh38
rs377012729 Chr.9: 78246741-78246741 on Build GRCh38
rs901789347 Chr.9: 78246685-78246685 on Build GRCh38
rs375279599 Chr.9: 78247016-78247016 on Build GRCh38
rs766225621 Chr.9: 78246703-78246703 on Build GRCh38
rs777180782 Chr.9: 78246747-78246747 on Build GRCh38
rs372759241 Chr.9: 78246699-78246699 on Build GRCh38
rs764191140 Chr.9: 78246778-78246778 on Build GRCh38
rs564127305 Chr.9: 78246832-78246832 on Build GRCh38
rs547637403 Chr.9: 78246891-78246891 on Build GRCh38
rs770586355 Chr.9: 78246755-78246755 on Build GRCh38
rs751389334 Chr.9: 78246794-78246794 on Build GRCh38
rs767748856 Chr.9: 78246765-78246766 on Build GRCh38
rs762067410 Chr.9: 78246789-78246789 on Build GRCh38
rs979341905 Chr.9: 78246726-78246726 on Build GRCh38
rs752714111 Chr.9: 78246689-78246689 on Build GRCh38
rs750698150 Chr.9: 78246832-78246832 on Build GRCh38
rs369627645 Chr.9: 78246768-78246768 on Build GRCh38
rs200279297 Chr.9: 78246756-78246756 on Build GRCh38
rs566283017 Chr.9: 78246934-78246934 on Build GRCh38
rs748803864 Chr.9: 78246819-78246819 on Build GRCh38
rs887116759 Chr.9: 78246738-78246738 on Build GRCh38
rs767330368 Chr.9: 78246798-78246798 on Build GRCh38
rs1040652066 Chr.9: 78246953-78246953 on Build GRCh38
rs751411534 Chr.9: 78246720-78246720 on Build GRCh38
rs781251521 Chr.9: 78246730-78246730 on Build GRCh38
rs989884873 Chr.9: 78246895-78246895 on Build GRCh38
rs369779785 Chr.9: 78246692-78246692 on Build GRCh38

View Full Product Details