Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 34942770-34943304 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM47B
Entrez Gene ID
170062
Gene Name
family with sequence similarity 47 member B
Gene Aliases
-
UniGene
Hs.197801
Gene Chromosome Location
Chr.X: 34942796-34944917 on Build GRCh38
Transcripts
NM_152631.2

SNP Detail

SNP ID Chromosome Location
rs758648172 Chr.X: 34942853-34942853 on Build GRCh38
rs773446343 Chr.X: 34943046-34943046 on Build GRCh38
rs748064935 Chr.X: 34943229-34943229 on Build GRCh38
rs371399704 Chr.X: 34942824-34942824 on Build GRCh38
rs981149569 Chr.X: 34942792-34942792 on Build GRCh38
rs189661576 Chr.X: 34943084-34943084 on Build GRCh38
rs776598580 Chr.X: 34943224-34943224 on Build GRCh38
rs753382563 Chr.X: 34942917-34942917 on Build GRCh38
rs887918142 Chr.X: 34943246-34943246 on Build GRCh38
rs368029767 Chr.X: 34943094-34943094 on Build GRCh38
rs761083228 Chr.X: 34943257-34943257 on Build GRCh38
rs752740595 Chr.X: 34942894-34942894 on Build GRCh38
rs753064894 Chr.X: 34943155-34943155 on Build GRCh38
rs1040414763 Chr.X: 34942860-34942860 on Build GRCh38
rs148435716 Chr.X: 34942842-34942842 on Build GRCh38
rs780488641 Chr.X: 34942854-34942854 on Build GRCh38
rs754354675 Chr.X: 34943162-34943162 on Build GRCh38
rs764410326 Chr.X: 34943279-34943279 on Build GRCh38
rs769556600 Chr.X: 34943013-34943013 on Build GRCh38
rs755391165 Chr.X: 34942930-34942930 on Build GRCh38
rs1006435222 Chr.X: 34942932-34942932 on Build GRCh38
rs752260164 Chr.X: 34942841-34942841 on Build GRCh38
rs146758510 Chr.X: 34943172-34943172 on Build GRCh38
rs769963911 Chr.X: 34942870-34942870 on Build GRCh38
rs757331630 Chr.X: 34943169-34943169 on Build GRCh38
rs776489198 Chr.X: 34943118-34943118 on Build GRCh38
rs112122426 Chr.X: 34943247-34943247 on Build GRCh38
rs781710998 Chr.X: 34942867-34942867 on Build GRCh38
rs746806586 Chr.X: 34943215-34943215 on Build GRCh38
rs764653185 Chr.X: 34943161-34943161 on Build GRCh38
rs763270543 Chr.X: 34942880-34942880 on Build GRCh38
rs1050943953 Chr.X: 34943245-34943245 on Build GRCh38
rs41305183 Chr.X: 34943189-34943189 on Build GRCh38
rs991326082 Chr.X: 34942816-34942816 on Build GRCh38
rs773538087 Chr.X: 34942822-34942822 on Build GRCh38
rs756546854 Chr.X: 34942882-34942882 on Build GRCh38
rs367704127 Chr.X: 34942815-34942815 on Build GRCh38
rs769247178 Chr.X: 34943240-34943240 on Build GRCh38
rs143433018 Chr.X: 34943134-34943134 on Build GRCh38
rs868846253 Chr.X: 34943036-34943036 on Build GRCh38
rs558565488 Chr.X: 34943061-34943061 on Build GRCh38
rs756658360 Chr.X: 34943068-34943068 on Build GRCh38
rs759572902 Chr.X: 34942832-34942832 on Build GRCh38
rs758694214 Chr.X: 34943188-34943188 on Build GRCh38
rs185208351 Chr.X: 34943049-34943049 on Build GRCh38
rs914415272 Chr.X: 34942821-34942821 on Build GRCh38
rs745488925 Chr.X: 34942945-34942945 on Build GRCh38
rs199666967 Chr.X: 34943146-34943146 on Build GRCh38
rs753566019 Chr.X: 34942845-34942845 on Build GRCh38
rs760241906 Chr.X: 34942898-34942898 on Build GRCh38
rs140472896 Chr.X: 34943179-34943179 on Build GRCh38
rs781427149 Chr.X: 34942808-34942808 on Build GRCh38
rs764311139 Chr.X: 34942838-34942838 on Build GRCh38
rs751415500 Chr.X: 34942827-34942827 on Build GRCh38
rs775436289 Chr.X: 34943099-34943099 on Build GRCh38
rs746135217 Chr.X: 34943092-34943092 on Build GRCh38
rs755303384 Chr.X: 34943030-34943030 on Build GRCh38
rs189542319 Chr.X: 34942888-34942888 on Build GRCh38
rs756506886 Chr.X: 34943160-34943160 on Build GRCh38
rs766240290 Chr.X: 34942825-34942825 on Build GRCh38
rs750129618 Chr.X: 34943231-34943232 on Build GRCh38
rs773277904 Chr.X: 34942871-34942871 on Build GRCh38
rs754333558 Chr.X: 34943281-34943281 on Build GRCh38
rs371703573 Chr.X: 34943120-34943120 on Build GRCh38
rs770566122 Chr.X: 34943249-34943249 on Build GRCh38
rs139593378 Chr.X: 34943029-34943029 on Build GRCh38
rs759524668 Chr.X: 34943155-34943163 on Build GRCh38
rs372462849 Chr.X: 34942933-34942933 on Build GRCh38
rs775526309 Chr.X: 34942997-34942997 on Build GRCh38
rs763688738 Chr.X: 34942906-34942906 on Build GRCh38
rs746924051 Chr.X: 34942956-34942956 on Build GRCh38
rs991679831 Chr.X: 34943065-34943065 on Build GRCh38
rs772809450 Chr.X: 34943133-34943133 on Build GRCh38
rs759395119 Chr.X: 34942983-34942983 on Build GRCh38
rs757343285 Chr.X: 34943082-34943082 on Build GRCh38
rs777279225 Chr.X: 34942843-34942843 on Build GRCh38
rs776418751 Chr.X: 34942999-34942999 on Build GRCh38
rs779573594 Chr.X: 34942947-34942947 on Build GRCh38
rs4999833 Chr.X: 34943147-34943147 on Build GRCh38
rs932082927 Chr.X: 34943206-34943206 on Build GRCh38
rs756946994 Chr.X: 34942922-34942922 on Build GRCh38
rs746850714 Chr.X: 34943100-34943100 on Build GRCh38
rs376051927 Chr.X: 34942884-34942884 on Build GRCh38
rs761951872 Chr.X: 34942790-34942790 on Build GRCh38
rs374173252 Chr.X: 34943012-34943012 on Build GRCh38
rs142579345 Chr.X: 34942885-34942885 on Build GRCh38
rs749385786 Chr.X: 34943080-34943080 on Build GRCh38
rs748209524 Chr.X: 34942869-34942869 on Build GRCh38
rs771188684 Chr.X: 34942975-34942975 on Build GRCh38
rs762526686 Chr.X: 34943248-34943248 on Build GRCh38
rs773937908 Chr.X: 34943251-34943251 on Build GRCh38
rs761296509 Chr.X: 34943016-34943016 on Build GRCh38
rs200255995 Chr.X: 34943222-34943222 on Build GRCh38
rs751913424 Chr.X: 34942929-34942929 on Build GRCh38
rs769151246 Chr.X: 34942864-34942864 on Build GRCh38
rs777648365 Chr.X: 34943078-34943078 on Build GRCh38
rs764929549 Chr.X: 34943019-34943019 on Build GRCh38
rs1002557863 Chr.X: 34942990-34942990 on Build GRCh38
rs747456969 Chr.X: 34942861-34942861 on Build GRCh38
rs764766129 Chr.X: 34943123-34943123 on Build GRCh38
rs867590745 Chr.X: 34943087-34943087 on Build GRCh38
rs774781860 Chr.X: 34942976-34942976 on Build GRCh38
rs368344355 Chr.X: 34942928-34942928 on Build GRCh38
rs79825675 Chr.X: 34942987-34942987 on Build GRCh38

View Full Product Details