Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 72160974-72161485 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FLJ44635
Entrez Gene ID
392490
Gene Name
TPT1-like protein
Gene Aliases
-
UniGene
Hs.660859
Gene Chromosome Location
Chr.X: 72144184-72161750 on Build GRCh38
Transcripts
NM_207422.2

SNP Detail

SNP ID Chromosome Location
rs894023299 Chr.X: 72161143-72161143 on Build GRCh38
rs909757823 Chr.X: 72161003-72161003 on Build GRCh38
rs918528674 Chr.X: 72161458-72161458 on Build GRCh38
rs922059658 Chr.X: 72161034-72161034 on Build GRCh38
rs141641908 Chr.X: 72161016-72161016 on Build GRCh38
rs138666060 Chr.X: 72161266-72161266 on Build GRCh38
rs761954553 Chr.X: 72161122-72161122 on Build GRCh38
rs768109938 Chr.X: 72161067-72161067 on Build GRCh38
rs929339410 Chr.X: 72161051-72161051 on Build GRCh38
rs962660996 Chr.X: 72161006-72161006 on Build GRCh38
rs750810597 Chr.X: 72161452-72161452 on Build GRCh38
rs150158509 Chr.X: 72161130-72161130 on Build GRCh38
rs762540338 Chr.X: 72161033-72161033 on Build GRCh38
rs987601981 Chr.X: 72161072-72161072 on Build GRCh38
rs912021042 Chr.X: 72161080-72161080 on Build GRCh38
rs1016786108 Chr.X: 72161376-72161376 on Build GRCh38
rs6624604 Chr.X: 72161399-72161399 on Build GRCh38
rs1039203300 Chr.X: 72161133-72161133 on Build GRCh38
rs779296620 Chr.X: 72161436-72161436 on Build GRCh38
rs6624602 Chr.X: 72161108-72161108 on Build GRCh38
rs191918317 Chr.X: 72161304-72161304 on Build GRCh38
rs1011176006 Chr.X: 72161152-72161152 on Build GRCh38
rs6624603 Chr.X: 72161180-72161180 on Build GRCh38
rs959161743 Chr.X: 72161305-72161305 on Build GRCh38
rs752719823 Chr.X: 72161224-72161224 on Build GRCh38
rs919275048 Chr.X: 72161028-72161028 on Build GRCh38
rs184007945 Chr.X: 72161389-72161389 on Build GRCh38
rs182965916 Chr.X: 72161179-72161179 on Build GRCh38
rs1012455268 Chr.X: 72161364-72161364 on Build GRCh38
rs767537688 Chr.X: 72161212-72161212 on Build GRCh38

View Full Product Details