Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 32166458-32166962 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EGFL8
Entrez Gene ID
80864
Gene Name
EGF like domain multiple 8
Gene Aliases
C6orf8, NG3
UniGene
Hs.731888
Gene Chromosome Location
Chr.6: 32164605-32168285 on Build GRCh38
Transcripts
NM_030652.3 NR_037860.1 NR_037861.1

Gene Symbol
PPT2-EGFL8
Entrez Gene ID
100532746
Gene Name
PPT2-EGFL8 readthrough (NMD candidate)
Gene Aliases
-
UniGene
Hs.731888
Gene Chromosome Location
Chr.6: 32153999-32168285 on Build GRCh38
Transcripts
NM_030652.3 NR_037860.1 NR_037861.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6652185 c.170T>C p.V57A Chr.6: 32166566-32166566 on Build GRCh38
6204595 c.252G>A p.E84E Chr.6: 32166728-32166728 on Build GRCh38
6193247 c.117C>T p.S39S Chr.6: 32166513-32166513 on Build GRCh38
3872933 c.260G>A p.R87Q Chr.6: 32166736-32166736 on Build GRCh38
6754695 c.102-4G>A p.? Chr.6: 32166494-32166494 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780581280 Chr.6: 32166911-32166911 on Build GRCh38
rs768104469 Chr.6: 32166517-32166517 on Build GRCh38
rs756107785 Chr.6: 32166853-32166853 on Build GRCh38
rs748400386 Chr.6: 32166706-32166710 on Build GRCh38
rs575900965 Chr.6: 32166534-32166534 on Build GRCh38
rs771318855 Chr.6: 32166688-32166688 on Build GRCh38
rs867897965 Chr.6: 32166780-32166780 on Build GRCh38
rs567711648 Chr.6: 32166494-32166494 on Build GRCh38
rs368674055 Chr.6: 32166845-32166845 on Build GRCh38
rs900817931 Chr.6: 32166915-32166915 on Build GRCh38
rs769920311 Chr.6: 32166493-32166493 on Build GRCh38
rs377454015 Chr.6: 32166547-32166547 on Build GRCh38
rs762314805 Chr.6: 32166504-32166504 on Build GRCh38
rs148110134 Chr.6: 32166518-32166518 on Build GRCh38
rs3096697 Chr.6: 32166733-32166733 on Build GRCh38
rs147273977 Chr.6: 32166875-32166875 on Build GRCh38
rs781644515 Chr.6: 32166886-32166886 on Build GRCh38
rs758132074 Chr.6: 32166619-32166619 on Build GRCh38
rs371155632 Chr.6: 32166665-32166665 on Build GRCh38
rs746963804 Chr.6: 32166564-32166564 on Build GRCh38
rs768210351 Chr.6: 32166920-32166920 on Build GRCh38
rs954256205 Chr.6: 32166742-32166742 on Build GRCh38
rs772497702 Chr.6: 32166678-32166678 on Build GRCh38
rs34898211 Chr.6: 32166696-32166697 on Build GRCh38
rs146391240 Chr.6: 32166735-32166735 on Build GRCh38
rs748340981 Chr.6: 32166744-32166744 on Build GRCh38
rs371176366 Chr.6: 32166796-32166796 on Build GRCh38
rs762259856 Chr.6: 32166594-32166594 on Build GRCh38
rs145530750 Chr.6: 32166803-32166803 on Build GRCh38
rs763832118 Chr.6: 32166724-32166724 on Build GRCh38
rs376635812 Chr.6: 32166723-32166723 on Build GRCh38
rs948574175 Chr.6: 32166593-32166593 on Build GRCh38
rs773311019 Chr.6: 32166721-32166732 on Build GRCh38
rs368801471 Chr.6: 32166856-32166856 on Build GRCh38
rs777015456 Chr.6: 32166827-32166835 on Build GRCh38
rs753309190 Chr.6: 32166602-32166602 on Build GRCh38
rs533046653 Chr.6: 32166790-32166790 on Build GRCh38
rs764817340 Chr.6: 32166717-32166717 on Build GRCh38
rs981686624 Chr.6: 32166913-32166913 on Build GRCh38
rs35319377 Chr.6: 32166500-32166501 on Build GRCh38
rs761496982 Chr.6: 32166843-32166843 on Build GRCh38
rs777263645 Chr.6: 32166701-32166701 on Build GRCh38
rs1028040431 Chr.6: 32166714-32166714 on Build GRCh38
rs749293324 Chr.6: 32166654-32166654 on Build GRCh38
rs376321383 Chr.6: 32166629-32166629 on Build GRCh38
rs759555973 Chr.6: 32166827-32166827 on Build GRCh38
rs779259395 Chr.6: 32166664-32166664 on Build GRCh38
rs778719150 Chr.6: 32166543-32166543 on Build GRCh38
rs374168906 Chr.6: 32166685-32166685 on Build GRCh38
rs556255985 Chr.6: 32166936-32166936 on Build GRCh38
rs1010773218 Chr.6: 32166758-32166758 on Build GRCh38
rs769257440 Chr.6: 32166812-32166812 on Build GRCh38
rs777552173 Chr.6: 32166627-32166627 on Build GRCh38
rs3130347 Chr.6: 32166879-32166879 on Build GRCh38
rs545453429 Chr.6: 32166542-32166542 on Build GRCh38
rs775058202 Chr.6: 32166718-32166718 on Build GRCh38
rs566703545 Chr.6: 32166837-32166837 on Build GRCh38
rs747201782 Chr.6: 32166902-32166902 on Build GRCh38
rs754773356 Chr.6: 32166720-32166721 on Build GRCh38
rs367939373 Chr.6: 32166844-32166844 on Build GRCh38
rs551640096 Chr.6: 32166836-32166836 on Build GRCh38
rs747332700 Chr.6: 32166784-32166784 on Build GRCh38
rs765820216 Chr.6: 32166521-32166521 on Build GRCh38
rs577709421 Chr.6: 32166734-32166734 on Build GRCh38
rs768844994 Chr.6: 32166663-32166663 on Build GRCh38
rs748795876 Chr.6: 32166918-32166918 on Build GRCh38
rs749479796 Chr.6: 32166586-32166586 on Build GRCh38
rs765199076 Chr.6: 32166772-32166772 on Build GRCh38
rs372390354 Chr.6: 32166871-32166871 on Build GRCh38
rs1052984738 Chr.6: 32166892-32166892 on Build GRCh38
rs778188254 Chr.6: 32166783-32166783 on Build GRCh38
rs774672313 Chr.6: 32166592-32166592 on Build GRCh38
rs752459687 Chr.6: 32166546-32166546 on Build GRCh38
rs150029816 Chr.6: 32166797-32166797 on Build GRCh38
rs745434046 Chr.6: 32166503-32166503 on Build GRCh38
rs762501828 Chr.6: 32166834-32166834 on Build GRCh38
rs772467659 Chr.6: 32166711-32166712 on Build GRCh38
rs746315178 Chr.6: 32166898-32166898 on Build GRCh38
rs143726348 Chr.6: 32166497-32166497 on Build GRCh38
rs769849513 Chr.6: 32166566-32166566 on Build GRCh38
rs141849753 Chr.6: 32166601-32166601 on Build GRCh38
rs752801440 Chr.6: 32166872-32166872 on Build GRCh38
rs960048319 Chr.6: 32166775-32166775 on Build GRCh38
rs527481138 Chr.6: 32166852-32166852 on Build GRCh38
rs759856358 Chr.6: 32166853-32166860 on Build GRCh38
rs141826798 Chr.6: 32166618-32166618 on Build GRCh38
rs766894626 Chr.6: 32166787-32166788 on Build GRCh38
rs549541405 Chr.6: 32166874-32166874 on Build GRCh38
rs756383733 Chr.6: 32166935-32166935 on Build GRCh38
rs754952287 Chr.6: 32166736-32166736 on Build GRCh38
rs992884896 Chr.6: 32166794-32166794 on Build GRCh38
rs764794536 Chr.6: 32166615-32166615 on Build GRCh38
rs774671618 Chr.6: 32166542-32166544 on Build GRCh38
rs140183306 Chr.6: 32166712-32166712 on Build GRCh38
rs758714469 Chr.6: 32166752-32166752 on Build GRCh38
rs371197629 Chr.6: 32166591-32166591 on Build GRCh38
rs201259021 Chr.6: 32166585-32166585 on Build GRCh38
rs781243251 Chr.6: 32166642-32166642 on Build GRCh38
rs757166744 Chr.6: 32166630-32166630 on Build GRCh38
rs1018861803 Chr.6: 32166848-32166848 on Build GRCh38
rs753463061 Chr.6: 32166525-32166525 on Build GRCh38
rs768293732 Chr.6: 32166684-32166684 on Build GRCh38
rs776945797 Chr.6: 32166795-32166795 on Build GRCh38
rs919294715 Chr.6: 32166510-32166510 on Build GRCh38
rs370779103 Chr.6: 32166600-32166600 on Build GRCh38
rs375116850 Chr.6: 32166523-32166523 on Build GRCh38
rs772620132 Chr.6: 32166596-32166596 on Build GRCh38
rs113910833 Chr.6: 32166808-32166808 on Build GRCh38
rs779034202 Chr.6: 32166666-32166666 on Build GRCh38
rs114706893 Chr.6: 32166743-32166743 on Build GRCh38
rs1027595328 Chr.6: 32166739-32166739 on Build GRCh38
rs757496939 Chr.6: 32166882-32166882 on Build GRCh38

View Full Product Details