Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 32168615-32169125 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AGPAT1
Entrez Gene ID
10554
Gene Name
1-acylglycerol-3-phosphate O-acyltransferase 1
Gene Aliases
1-AGPAT1, G15, LPAAT-alpha, LPAATA
UniGene
Hs.409230
Gene Chromosome Location
Chr.6: 32168206-32178111 on Build GRCh38
Transcripts
NM_006411.3 NM_032741.4 XM_005248805.3 XM_005248806.2 XM_011514234.1

SNP Detail

SNP ID Chromosome Location
rs1061807 Chr.6: 32169061-32169061 on Build GRCh38
rs11553430 Chr.6: 32168994-32168994 on Build GRCh38
rs1040202996 Chr.6: 32168913-32168913 on Build GRCh38
rs765871801 Chr.6: 32168913-32168914 on Build GRCh38
rs572055172 Chr.6: 32168833-32168833 on Build GRCh38
rs1061808 Chr.6: 32168770-32168770 on Build GRCh38
rs960240546 Chr.6: 32168711-32168711 on Build GRCh38
rs934140946 Chr.6: 32168914-32168914 on Build GRCh38
rs764571328 Chr.6: 32168672-32168672 on Build GRCh38
rs754581642 Chr.6: 32168641-32168641 on Build GRCh38
rs184093218 Chr.6: 32169069-32169069 on Build GRCh38
rs1017007755 Chr.6: 32168650-32168650 on Build GRCh38
rs139427026 Chr.6: 32168816-32168816 on Build GRCh38
rs562654671 Chr.6: 32168943-32168943 on Build GRCh38
rs925702509 Chr.6: 32168774-32168774 on Build GRCh38
rs992786438 Chr.6: 32168722-32168722 on Build GRCh38
rs936180944 Chr.6: 32168691-32168691 on Build GRCh38
rs995320112 Chr.6: 32168990-32168990 on Build GRCh38
rs1034162586 Chr.6: 32168690-32168691 on Build GRCh38
rs543561330 Chr.6: 32169053-32169053 on Build GRCh38
rs887355030 Chr.6: 32168957-32168957 on Build GRCh38
rs990309564 Chr.6: 32168754-32168754 on Build GRCh38
rs777568527 Chr.6: 32168928-32168928 on Build GRCh38
rs762200991 Chr.6: 32168911-32168911 on Build GRCh38
rs555996405 Chr.6: 32168687-32168687 on Build GRCh38
rs1014499814 Chr.6: 32169016-32169016 on Build GRCh38
rs538661229 Chr.6: 32168781-32168781 on Build GRCh38
rs959921767 Chr.6: 32169000-32169000 on Build GRCh38
rs978429344 Chr.6: 32169084-32169084 on Build GRCh38
rs369014153 Chr.6: 32168998-32168998 on Build GRCh38
rs1005517045 Chr.6: 32168958-32168958 on Build GRCh38
rs578022721 Chr.6: 32168962-32168962 on Build GRCh38
rs769539488 Chr.6: 32168912-32168912 on Build GRCh38
rs1001325703 Chr.6: 32168680-32168680 on Build GRCh38
rs576195877 Chr.6: 32168999-32168999 on Build GRCh38
rs1038293701 Chr.6: 32168983-32168983 on Build GRCh38
rs913120063 Chr.6: 32168752-32168752 on Build GRCh38
rs969410887 Chr.6: 32168662-32168662 on Build GRCh38
rs541114057 Chr.6: 32169099-32169099 on Build GRCh38
rs1024605509 Chr.6: 32169004-32169004 on Build GRCh38
rs1001931923 Chr.6: 32169035-32169035 on Build GRCh38
rs969479479 Chr.6: 32169032-32169032 on Build GRCh38
rs967072098 Chr.6: 32168762-32168762 on Build GRCh38
rs891121229 Chr.6: 32168891-32168891 on Build GRCh38

View Full Product Details