Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 32175572-32176066 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AGPAT1
Entrez Gene ID
10554
Gene Name
1-acylglycerol-3-phosphate O-acyltransferase 1
Gene Aliases
1-AGPAT1, G15, LPAAT-alpha, LPAATA
UniGene
Hs.409230
Gene Chromosome Location
Chr.6: 32168206-32178111 on Build GRCh38
Transcripts
NM_006411.3 NM_032741.4 XM_005248805.3 XM_005248806.2 XM_011514234.1

SNP Detail

SNP ID Chromosome Location
rs533166631 Chr.6: 32175828-32175828 on Build GRCh38
rs943503035 Chr.6: 32175903-32175903 on Build GRCh38
rs969848229 Chr.6: 32175717-32175717 on Build GRCh38
rs3134948 Chr.6: 32175646-32175646 on Build GRCh38
rs887502296 Chr.6: 32175969-32175969 on Build GRCh38
rs1003155253 Chr.6: 32176022-32176022 on Build GRCh38
rs895825422 Chr.6: 32175989-32175989 on Build GRCh38
rs1037906536 Chr.6: 32175913-32175913 on Build GRCh38
rs776057141 Chr.6: 32175656-32175656 on Build GRCh38
rs557433073 Chr.6: 32175631-32175631 on Build GRCh38
rs562763358 Chr.6: 32175765-32175765 on Build GRCh38
rs188890271 Chr.6: 32175695-32175695 on Build GRCh38
rs1006805155 Chr.6: 32175982-32175982 on Build GRCh38
rs899411379 Chr.6: 32175920-32175920 on Build GRCh38
rs549390649 Chr.6: 32175600-32175600 on Build GRCh38
rs929547981 Chr.6: 32175936-32175936 on Build GRCh38
rs985225061 Chr.6: 32175823-32175823 on Build GRCh38
rs567521653 Chr.6: 32175647-32175647 on Build GRCh38
rs1035925903 Chr.6: 32176024-32176024 on Build GRCh38
rs13205654 Chr.6: 32175988-32175988 on Build GRCh38
rs933726348 Chr.6: 32175778-32175778 on Build GRCh38
rs1047686857 Chr.6: 32175987-32175987 on Build GRCh38
rs988456570 Chr.6: 32175763-32175763 on Build GRCh38
rs139168537 Chr.6: 32175732-32175732 on Build GRCh38
rs1014283103 Chr.6: 32176001-32176001 on Build GRCh38
rs899442710 Chr.6: 32175594-32175594 on Build GRCh38
rs112120659 Chr.6: 32175806-32175806 on Build GRCh38
rs573557569 Chr.6: 32175729-32175729 on Build GRCh38
rs115463529 Chr.6: 32175701-32175701 on Build GRCh38
rs768728263 Chr.6: 32175820-32175820 on Build GRCh38
rs1032884799 Chr.6: 32175734-32175734 on Build GRCh38
rs183995385 Chr.6: 32175677-32175677 on Build GRCh38
rs1022994190 Chr.6: 32176020-32176020 on Build GRCh38
rs1047501837 Chr.6: 32175835-32175835 on Build GRCh38
rs956191306 Chr.6: 32175744-32175744 on Build GRCh38
rs1040255355 Chr.6: 32175797-32175797 on Build GRCh38
rs34586370 Chr.6: 32176018-32176019 on Build GRCh38

View Full Product Details