Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 32175882-32176389 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AGPAT1
Entrez Gene ID
10554
Gene Name
1-acylglycerol-3-phosphate O-acyltransferase 1
Gene Aliases
1-AGPAT1, G15, LPAAT-alpha, LPAATA
UniGene
Hs.409230
Gene Chromosome Location
Chr.6: 32168206-32178111 on Build GRCh38
Transcripts
NM_006411.3 NM_032741.4 XM_005248805.3 XM_005248806.2 XM_011514234.1

SNP Detail

SNP ID Chromosome Location
rs896100246 Chr.6: 32176249-32176249 on Build GRCh38
rs887502296 Chr.6: 32175969-32175969 on Build GRCh38
rs895825422 Chr.6: 32175989-32175989 on Build GRCh38
rs1003155253 Chr.6: 32176022-32176022 on Build GRCh38
rs989148136 Chr.6: 32176110-32176110 on Build GRCh38
rs550214821 Chr.6: 32176273-32176273 on Build GRCh38
rs1047348012 Chr.6: 32176193-32176193 on Build GRCh38
rs955949279 Chr.6: 32176075-32176075 on Build GRCh38
rs538868383 Chr.6: 32176351-32176351 on Build GRCh38
rs527821586 Chr.6: 32176202-32176202 on Build GRCh38
rs964880937 Chr.6: 32176088-32176088 on Build GRCh38
rs560586629 Chr.6: 32176167-32176167 on Build GRCh38
rs551576878 Chr.6: 32176068-32176068 on Build GRCh38
rs910965054 Chr.6: 32176182-32176182 on Build GRCh38
rs1029307087 Chr.6: 32176116-32176116 on Build GRCh38
rs1054189070 Chr.6: 32176322-32176322 on Build GRCh38
rs905992565 Chr.6: 32176065-32176065 on Build GRCh38
rs1014283103 Chr.6: 32176001-32176001 on Build GRCh38
rs567600571 Chr.6: 32176258-32176258 on Build GRCh38
rs1006996099 Chr.6: 32176328-32176328 on Build GRCh38
rs571479116 Chr.6: 32176215-32176215 on Build GRCh38
rs955195137 Chr.6: 32176146-32176146 on Build GRCh38
rs1048430254 Chr.6: 32176248-32176248 on Build GRCh38
rs1022994190 Chr.6: 32176020-32176020 on Build GRCh38
rs770328373 Chr.6: 32176099-32176099 on Build GRCh38
rs571560310 Chr.6: 32176292-32176292 on Build GRCh38
rs985151956 Chr.6: 32176180-32176180 on Build GRCh38
rs910414547 Chr.6: 32176083-32176083 on Build GRCh38
rs1037906536 Chr.6: 32175913-32175913 on Build GRCh38
rs777877740 Chr.6: 32176276-32176276 on Build GRCh38
rs1010258029 Chr.6: 32176262-32176262 on Build GRCh38
rs766282633 Chr.6: 32176060-32176060 on Build GRCh38
rs1006805155 Chr.6: 32175982-32175982 on Build GRCh38
rs899411379 Chr.6: 32175920-32175920 on Build GRCh38
rs204997 Chr.6: 32176234-32176234 on Build GRCh38
rs905918906 Chr.6: 32176283-32176283 on Build GRCh38
rs753930537 Chr.6: 32176296-32176296 on Build GRCh38
rs929547981 Chr.6: 32175936-32175936 on Build GRCh38
rs373229117 Chr.6: 32176066-32176066 on Build GRCh38
rs1035925903 Chr.6: 32176024-32176024 on Build GRCh38
rs13205654 Chr.6: 32175988-32175988 on Build GRCh38
rs923182046 Chr.6: 32176125-32176125 on Build GRCh38
rs1047686857 Chr.6: 32175987-32175987 on Build GRCh38
rs538230942 Chr.6: 32176260-32176260 on Build GRCh38
rs1057416470 Chr.6: 32176259-32176259 on Build GRCh38
rs891641017 Chr.6: 32176346-32176346 on Build GRCh38
rs929553912 Chr.6: 32176225-32176225 on Build GRCh38
rs16869385 Chr.6: 32176214-32176214 on Build GRCh38
rs920765933 Chr.6: 32176218-32176218 on Build GRCh38
rs868709824 Chr.6: 32176109-32176109 on Build GRCh38
rs34586370 Chr.6: 32176018-32176019 on Build GRCh38

View Full Product Details