Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 32368649-32369162 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C6orf10
Entrez Gene ID
10665
Gene Name
chromosome 6 open reading frame 10
Gene Aliases
TSBP
UniGene
Hs.567414
Gene Chromosome Location
Chr.6: 32290523-32371912 on Build GRCh38
Transcripts
NM_001286474.1 NM_001286475.1 NM_006781.4 XM_017010185.1 NR_136244.1 NR_136245.1

Gene Symbol
LOC101929163
Entrez Gene ID
101929163
Gene Name
uncharacterized LOC101929163
Gene Aliases
-
UniGene
Hs.57692
Gene Chromosome Location
Chr.6: 32255173-32407730 on Build GRCh38
Transcripts
NM_001286474.1 NM_001286475.1 NM_006781.4 XM_017010185.1 NR_136244.1 NR_136245.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5959837 c.106T>C p.Y36H Chr.6: 32368809-32368809 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs563122445 Chr.6: 32368939-32368939 on Build GRCh38
rs940655477 Chr.6: 32368862-32368862 on Build GRCh38
rs967422434 Chr.6: 32368710-32368710 on Build GRCh38
rs747720315 Chr.6: 32368764-32368769 on Build GRCh38
rs907332191 Chr.6: 32368902-32368902 on Build GRCh38
rs9268384 Chr.6: 32368809-32368809 on Build GRCh38
rs113857285 Chr.6: 32368768-32368768 on Build GRCh38
rs1041076128 Chr.6: 32368676-32368676 on Build GRCh38
rs564997537 Chr.6: 32368686-32368686 on Build GRCh38
rs769874016 Chr.6: 32368770-32368770 on Build GRCh38
rs771978326 Chr.6: 32368761-32368761 on Build GRCh38
rs6903816 Chr.6: 32368740-32368740 on Build GRCh38
rs768195327 Chr.6: 32368735-32368735 on Build GRCh38
rs576198629 Chr.6: 32368780-32368780 on Build GRCh38
rs758340094 Chr.6: 32368784-32368788 on Build GRCh38
rs986556869 Chr.6: 32368711-32368711 on Build GRCh38
rs763169843 Chr.6: 32368778-32368778 on Build GRCh38
rs3129938 Chr.6: 32368718-32368718 on Build GRCh38
rs1020589857 Chr.6: 32368960-32368960 on Build GRCh38
rs773176464 Chr.6: 32368764-32368764 on Build GRCh38
rs748909691 Chr.6: 32368731-32368731 on Build GRCh38
rs569865178 Chr.6: 32369084-32369084 on Build GRCh38
rs140156094 Chr.6: 32368793-32368793 on Build GRCh38
rs762306607 Chr.6: 32368799-32368799 on Build GRCh38
rs200083918 Chr.6: 32368744-32368744 on Build GRCh38
rs3129939 Chr.6: 32368989-32368989 on Build GRCh38
rs183137162 Chr.6: 32368916-32368916 on Build GRCh38
rs974935036 Chr.6: 32368737-32368737 on Build GRCh38
rs1049748587 Chr.6: 32368777-32368777 on Build GRCh38
rs1055577032 Chr.6: 32368918-32368918 on Build GRCh38
rs542793423 Chr.6: 32368775-32368775 on Build GRCh38
rs745374453 Chr.6: 32369123-32369123 on Build GRCh38
rs576631957 Chr.6: 32368804-32368804 on Build GRCh38
rs901745041 Chr.6: 32368677-32368677 on Build GRCh38
rs1035388815 Chr.6: 32368874-32368874 on Build GRCh38
rs963779584 Chr.6: 32368726-32368726 on Build GRCh38
rs570867502 Chr.6: 32368715-32368715 on Build GRCh38
rs866186638 Chr.6: 32369063-32369063 on Build GRCh38
rs764291835 Chr.6: 32368789-32368789 on Build GRCh38
rs759374091 Chr.6: 32368808-32368808 on Build GRCh38
rs753354315 Chr.6: 32368860-32368860 on Build GRCh38
rs947531820 Chr.6: 32368988-32368989 on Build GRCh38
rs1011507122 Chr.6: 32368954-32368954 on Build GRCh38
rs967074690 Chr.6: 32368974-32368974 on Build GRCh38
rs756588512 Chr.6: 32368841-32368841 on Build GRCh38
rs778360295 Chr.6: 32368750-32368753 on Build GRCh38
rs551936546 Chr.6: 32369028-32369028 on Build GRCh38
rs779549018 Chr.6: 32368849-32368849 on Build GRCh38
rs565289863 Chr.6: 32368866-32368866 on Build GRCh38
rs559979875 Chr.6: 32368951-32368951 on Build GRCh38
rs978157168 Chr.6: 32369032-32369048 on Build GRCh38
rs778671262 Chr.6: 32368738-32368738 on Build GRCh38
rs35272353 Chr.6: 32368744-32368744 on Build GRCh38

View Full Product Details