Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 32968486-32968973 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BRD2
Entrez Gene ID
6046
Gene Name
bromodomain containing 2
Gene Aliases
D6S113E, FSH, FSRG1, NAT, O27.1.1, RING3, RNF3
UniGene
Hs.75243
Gene Chromosome Location
Chr.6: 32968660-32981505 on Build GRCh38
Transcripts
NM_005104.3

SNP Detail

SNP ID Chromosome Location
rs1040297619 Chr.6: 32968617-32968617 on Build GRCh38
rs1020733212 Chr.6: 32968933-32968933 on Build GRCh38
rs1020209560 Chr.6: 32968710-32968710 on Build GRCh38
rs1002539686 Chr.6: 32968891-32968891 on Build GRCh38
rs981042552 Chr.6: 32968555-32968555 on Build GRCh38
rs949431256 Chr.6: 32968851-32968851 on Build GRCh38
rs1025687526 Chr.6: 32968516-32968516 on Build GRCh38
rs557083524 Chr.6: 32968804-32968804 on Build GRCh38
rs3918149 Chr.6: 32968596-32968596 on Build GRCh38
rs942950383 Chr.6: 32968605-32968605 on Build GRCh38
rs1033540034 Chr.6: 32968747-32968754 on Build GRCh38
rs992759355 Chr.6: 32968591-32968591 on Build GRCh38
rs893650644 Chr.6: 32968905-32968905 on Build GRCh38
rs552557898 Chr.6: 32968782-32968782 on Build GRCh38
rs1000575569 Chr.6: 32968900-32968900 on Build GRCh38
rs528716521 Chr.6: 32968548-32968548 on Build GRCh38
rs547068767 Chr.6: 32968562-32968562 on Build GRCh38
rs941109630 Chr.6: 32968652-32968652 on Build GRCh38
rs774599277 Chr.6: 32968716-32968716 on Build GRCh38
rs1013568706 Chr.6: 32968650-32968650 on Build GRCh38
rs967821451 Chr.6: 32968715-32968715 on Build GRCh38
rs570285722 Chr.6: 32968681-32968681 on Build GRCh38
rs34854167 Chr.6: 32968875-32968877 on Build GRCh38
rs111227999 Chr.6: 32968771-32968771 on Build GRCh38
rs569183584 Chr.6: 32968651-32968651 on Build GRCh38
rs1000266150 Chr.6: 32968746-32968747 on Build GRCh38
rs868610272 Chr.6: 32968829-32968829 on Build GRCh38
rs1047143320 Chr.6: 32968643-32968643 on Build GRCh38
rs3918150 Chr.6: 32968658-32968658 on Build GRCh38
rs1033031951 Chr.6: 32968908-32968908 on Build GRCh38
rs975677676 Chr.6: 32968641-32968642 on Build GRCh38
rs1002126341 Chr.6: 32968667-32968667 on Build GRCh38
rs528656351 Chr.6: 32968844-32968844 on Build GRCh38
rs1009329683 Chr.6: 32968911-32968911 on Build GRCh38
rs987854127 Chr.6: 32968707-32968707 on Build GRCh38
rs562313132 Chr.6: 32968860-32968860 on Build GRCh38
rs1046473377 Chr.6: 32968876-32968876 on Build GRCh38
rs987123785 Chr.6: 32968769-32968769 on Build GRCh38
rs987901170 Chr.6: 32968594-32968594 on Build GRCh38
rs1047197747 Chr.6: 32968841-32968841 on Build GRCh38
rs529383010 Chr.6: 32968882-32968882 on Build GRCh38
rs960019714 Chr.6: 32968583-32968583 on Build GRCh38
rs552624755 Chr.6: 32968522-32968522 on Build GRCh38
rs534513793 Chr.6: 32968686-32968686 on Build GRCh38
rs3135331 Chr.6: 32968947-32968947 on Build GRCh38
rs951351744 Chr.6: 32968528-32968528 on Build GRCh38
rs114383918 Chr.6: 32968830-32968830 on Build GRCh38
rs540818158 Chr.6: 32968846-32968846 on Build GRCh38
rs964174584 Chr.6: 32968757-32968757 on Build GRCh38
rs564667023 Chr.6: 32968531-32968531 on Build GRCh38
rs958067144 Chr.6: 32968576-32968576 on Build GRCh38
rs1051307705 Chr.6: 32968600-32968600 on Build GRCh38
rs144708781 Chr.6: 32968761-32968761 on Build GRCh38
rs931422190 Chr.6: 32968624-32968624 on Build GRCh38
rs1020639995 Chr.6: 32968755-32968755 on Build GRCh38
rs879532932 Chr.6: 32968901-32968902 on Build GRCh38
rs577212235 Chr.6: 32968821-32968821 on Build GRCh38
rs905009485 Chr.6: 32968659-32968659 on Build GRCh38
rs898659082 Chr.6: 32968620-32968620 on Build GRCh38
rs546447196 Chr.6: 32968821-32968821 on Build GRCh38
rs1042307625 Chr.6: 32968888-32968888 on Build GRCh38
rs527532696 Chr.6: 32968819-32968819 on Build GRCh38
rs1036168253 Chr.6: 32968948-32968948 on Build GRCh38
rs1009442813 Chr.6: 32968705-32968705 on Build GRCh38
rs954392635 Chr.6: 32968756-32968756 on Build GRCh38
rs552612384 Chr.6: 32968714-32968714 on Build GRCh38
rs370700605 Chr.6: 32968688-32968688 on Build GRCh38
rs535991051 Chr.6: 32968598-32968598 on Build GRCh38
rs768201728 Chr.6: 32968747-32968747 on Build GRCh38
rs113472313 Chr.6: 32968799-32968799 on Build GRCh38
rs1024858095 Chr.6: 32968534-32968534 on Build GRCh38
rs1034829715 Chr.6: 32968673-32968673 on Build GRCh38
rs954483636 Chr.6: 32968941-32968941 on Build GRCh38
rs964433676 Chr.6: 32968922-32968922 on Build GRCh38
rs922820561 Chr.6: 32968643-32968643 on Build GRCh38
rs1032625535 Chr.6: 32968682-32968682 on Build GRCh38
rs912787493 Chr.6: 32968604-32968604 on Build GRCh38
rs746348154 Chr.6: 32968561-32968561 on Build GRCh38
rs920095491 Chr.6: 32968833-32968833 on Build GRCh38
rs192593423 Chr.6: 32968778-32968778 on Build GRCh38
rs370095948 Chr.6: 32968902-32968902 on Build GRCh38
rs987176121 Chr.6: 32968949-32968949 on Build GRCh38
rs887109146 Chr.6: 32968645-32968645 on Build GRCh38
rs550946560 Chr.6: 32968642-32968642 on Build GRCh38
rs9276935 Chr.6: 32968664-32968664 on Build GRCh38
rs931457933 Chr.6: 32968835-32968835 on Build GRCh38

View Full Product Details