Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 184791723-184792322 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM129A
Entrez Gene ID
116496
Gene Name
family with sequence similarity 129 member A
Gene Aliases
C1orf24, GIG39, NIBAN
UniGene
Hs.518662
Gene Chromosome Location
Chr.1: 184791025-184975132 on Build GRCh38
Transcripts
NM_052966.3 XM_011509140.2 XM_011509141.2

SNP Detail

SNP ID Chromosome Location
rs562056428 Chr.1: 184792237-184792237 on Build GRCh38
rs1041438519 Chr.1: 184791959-184791963 on Build GRCh38
rs947595629 Chr.1: 184792263-184792263 on Build GRCh38
rs764397058 Chr.1: 184791756-184791756 on Build GRCh38
rs945723040 Chr.1: 184791927-184791927 on Build GRCh38
rs112320565 Chr.1: 184792095-184792095 on Build GRCh38
rs772041682 Chr.1: 184791962-184791962 on Build GRCh38
rs682331 Chr.1: 184792231-184792231 on Build GRCh38
rs981529269 Chr.1: 184792176-184792176 on Build GRCh38
rs897059517 Chr.1: 184792204-184792204 on Build GRCh38
rs869026227 Chr.1: 184791974-184791974 on Build GRCh38
rs376989781 Chr.1: 184791983-184791983 on Build GRCh38
rs1139074 Chr.1: 184791991-184791991 on Build GRCh38
rs896175087 Chr.1: 184792105-184792105 on Build GRCh38
rs1053402600 Chr.1: 184791959-184791959 on Build GRCh38
rs1019122052 Chr.1: 184792068-184792068 on Build GRCh38
rs978936860 Chr.1: 184792266-184792266 on Build GRCh38
rs985533776 Chr.1: 184792225-184792225 on Build GRCh38
rs944018795 Chr.1: 184791956-184791956 on Build GRCh38
rs869287222 Chr.1: 184791960-184791961 on Build GRCh38
rs983868514 Chr.1: 184792154-184792154 on Build GRCh38
rs1139068 Chr.1: 184792186-184792186 on Build GRCh38
rs995302637 Chr.1: 184792206-184792206 on Build GRCh38
rs974304416 Chr.1: 184791879-184791879 on Build GRCh38
rs492126 Chr.1: 184792052-184792052 on Build GRCh38
rs776829698 Chr.1: 184791960-184791962 on Build GRCh38
rs1043209500 Chr.1: 184792018-184792018 on Build GRCh38
rs1015544446 Chr.1: 184792147-184792147 on Build GRCh38
rs756043735 Chr.1: 184792081-184792081 on Build GRCh38
rs530963382 Chr.1: 184792224-184792224 on Build GRCh38
rs181090379 Chr.1: 184791828-184791828 on Build GRCh38
rs1054812193 Chr.1: 184792066-184792066 on Build GRCh38
rs577041862 Chr.1: 184791803-184791803 on Build GRCh38
rs531537611 Chr.1: 184792230-184792230 on Build GRCh38
rs74132038 Chr.1: 184791875-184791875 on Build GRCh38
rs376890841 Chr.1: 184792000-184792000 on Build GRCh38
rs1139072 Chr.1: 184792045-184792045 on Build GRCh38
rs1015907235 Chr.1: 184791867-184791867 on Build GRCh38
rs913468355 Chr.1: 184791973-184791973 on Build GRCh38
rs3187911 Chr.1: 184792028-184792028 on Build GRCh38
rs555687511 Chr.1: 184791961-184791961 on Build GRCh38
rs954089387 Chr.1: 184791902-184791902 on Build GRCh38
rs1139070 Chr.1: 184792140-184792140 on Build GRCh38
rs1139069 Chr.1: 184792178-184792178 on Build GRCh38
rs184889610 Chr.1: 184792034-184792034 on Build GRCh38
rs1026740677 Chr.1: 184792211-184792211 on Build GRCh38
rs545929681 Chr.1: 184792118-184792118 on Build GRCh38
rs149007931 Chr.1: 184792108-184792108 on Build GRCh38
rs912539991 Chr.1: 184791945-184791945 on Build GRCh38
rs1014973626 Chr.1: 184792160-184792160 on Build GRCh38
rs145686869 Chr.1: 184791895-184791895 on Build GRCh38
rs145066089 Chr.1: 184791973-184791974 on Build GRCh38
rs981247413 Chr.1: 184791773-184791773 on Build GRCh38
rs55815111 Chr.1: 184791779-184791779 on Build GRCh38
rs370954516 Chr.1: 184792169-184792169 on Build GRCh38
rs763434573 Chr.1: 184791969-184791979 on Build GRCh38
rs3180402 Chr.1: 184792054-184792054 on Build GRCh38
rs564140105 Chr.1: 184792202-184792202 on Build GRCh38
rs996416227 Chr.1: 184792094-184792094 on Build GRCh38
rs985439111 Chr.1: 184791934-184791934 on Build GRCh38
rs529170823 Chr.1: 184792243-184792243 on Build GRCh38
rs892787745 Chr.1: 184791913-184791913 on Build GRCh38
rs952054890 Chr.1: 184792139-184792139 on Build GRCh38
rs778508375 Chr.1: 184791961-184791962 on Build GRCh38
rs572204639 Chr.1: 184792103-184792103 on Build GRCh38
rs990061552 Chr.1: 184792268-184792268 on Build GRCh38
rs913490001 Chr.1: 184792257-184792257 on Build GRCh38
rs954146849 Chr.1: 184792223-184792223 on Build GRCh38
rs11338849 Chr.1: 184791960-184791960 on Build GRCh38
rs779458288 Chr.1: 184791781-184791781 on Build GRCh38
rs149769722 Chr.1: 184791759-184791759 on Build GRCh38
rs558762772 Chr.1: 184791780-184791780 on Build GRCh38

View Full Product Details