Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 26496068-26496609 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APBB1IP
Entrez Gene ID
54518
Gene Name
amyloid beta precursor protein binding family B member 1 interacting protein
Gene Aliases
INAG1, PREL1, RARP1, RIAM
UniGene
Hs.310421
Gene Chromosome Location
Chr.10: 26438324-26567879 on Build GRCh38
Transcripts
NM_019043.3 XM_006717451.2 XM_011519514.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5130164 c.100C>G p.P34A Chr.10: 26496331-26496331 on Build GRCh38
4984512 c.116C>A p.P39H Chr.10: 26496347-26496347 on Build GRCh38
4984511 c.116C>A p.P39H Chr.10: 26496347-26496347 on Build GRCh38
1603355 c.122C>T p.A41V Chr.10: 26496353-26496353 on Build GRCh38
3436872 c.73-1G>A p.? Chr.10: 26496303-26496303 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs560488377 Chr.10: 26496189-26496189 on Build GRCh38
rs933310739 Chr.10: 26496383-26496383 on Build GRCh38
rs980296190 Chr.10: 26496504-26496504 on Build GRCh38
rs767353585 Chr.10: 26496270-26496270 on Build GRCh38
rs546300624 Chr.10: 26496245-26496245 on Build GRCh38
rs369446926 Chr.10: 26496283-26496283 on Build GRCh38
rs763263345 Chr.10: 26496111-26496111 on Build GRCh38
rs575031195 Chr.10: 26496152-26496152 on Build GRCh38
rs745309388 Chr.10: 26496292-26496293 on Build GRCh38
rs757132629 Chr.10: 26496269-26496272 on Build GRCh38
rs369222330 Chr.10: 26496214-26496214 on Build GRCh38
rs757420534 Chr.10: 26496298-26496298 on Build GRCh38
rs781120621 Chr.10: 26496283-26496284 on Build GRCh38
rs996731621 Chr.10: 26496345-26496345 on Build GRCh38
rs899321335 Chr.10: 26496555-26496555 on Build GRCh38
rs948215476 Chr.10: 26496569-26496569 on Build GRCh38
rs750734525 Chr.10: 26496319-26496319 on Build GRCh38
rs942070081 Chr.10: 26496567-26496567 on Build GRCh38
rs374864811 Chr.10: 26496321-26496321 on Build GRCh38
rs570838395 Chr.10: 26496208-26496208 on Build GRCh38
rs1048307911 Chr.10: 26496557-26496557 on Build GRCh38
rs921855367 Chr.10: 26496354-26496354 on Build GRCh38
rs955930488 Chr.10: 26496463-26496465 on Build GRCh38
rs7900091 Chr.10: 26496260-26496260 on Build GRCh38
rs777890848 Chr.10: 26496266-26496266 on Build GRCh38
rs1012488751 Chr.10: 26496098-26496098 on Build GRCh38
rs928828644 Chr.10: 26496550-26496550 on Build GRCh38
rs753824149 Chr.10: 26496292-26496292 on Build GRCh38
rs369272082 Chr.10: 26496333-26496333 on Build GRCh38
rs202129957 Chr.10: 26496288-26496288 on Build GRCh38
rs547028219 Chr.10: 26496568-26496568 on Build GRCh38
rs200343167 Chr.10: 26496312-26496312 on Build GRCh38
rs987877849 Chr.10: 26496452-26496452 on Build GRCh38
rs372848780 Chr.10: 26496285-26496285 on Build GRCh38
rs556843650 Chr.10: 26496336-26496336 on Build GRCh38
rs532689072 Chr.10: 26496216-26496216 on Build GRCh38
rs914186079 Chr.10: 26496489-26496489 on Build GRCh38
rs905498775 Chr.10: 26496200-26496200 on Build GRCh38
rs557557089 Chr.10: 26496135-26496135 on Build GRCh38
rs990834599 Chr.10: 26496170-26496170 on Build GRCh38
rs907911480 Chr.10: 26496465-26496465 on Build GRCh38
rs1028172656 Chr.10: 26496353-26496353 on Build GRCh38
rs368860973 Chr.10: 26496505-26496505 on Build GRCh38
rs940755062 Chr.10: 26496487-26496487 on Build GRCh38
rs540678068 Chr.10: 26496171-26496171 on Build GRCh38
rs781737065 Chr.10: 26496351-26496351 on Build GRCh38
rs970293578 Chr.10: 26496496-26496496 on Build GRCh38
rs779220445 Chr.10: 26496307-26496307 on Build GRCh38
rs201471966 Chr.10: 26496329-26496329 on Build GRCh38
rs568265142 Chr.10: 26496338-26496338 on Build GRCh38
rs1037816254 Chr.10: 26496552-26496552 on Build GRCh38
rs898416599 Chr.10: 26496322-26496322 on Build GRCh38
rs770437464 Chr.10: 26496375-26496375 on Build GRCh38
rs771819264 Chr.10: 26496441-26496441 on Build GRCh38
rs907946029 Chr.10: 26496559-26496559 on Build GRCh38
rs745583284 Chr.10: 26496394-26496394 on Build GRCh38
rs748677908 Chr.10: 26496358-26496358 on Build GRCh38
rs773977870 Chr.10: 26496380-26496380 on Build GRCh38
rs373211324 Chr.10: 26496382-26496382 on Build GRCh38

View Full Product Details