Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 110119080-110119619 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADD3
Entrez Gene ID
120
Gene Name
adducin 3
Gene Aliases
ADDL, CPSQ3
UniGene
Hs.501012
Gene Chromosome Location
Chr.10: 109996350-110135565 on Build GRCh38
Transcripts
NM_001121.3 NM_001320591.1 NM_001320592.1 NM_001320593.1 NM_001320594.1 NM_016824.4 NM_019903.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4506683 c.726C>T p.S242S Chr.10: 110119219-110119219 on Build GRCh38
69562 c.782C>T p.A261V Chr.10: 110119275-110119275 on Build GRCh38
4560560 c.848G>A p.G283E Chr.10: 110119341-110119341 on Build GRCh38
7354565 c.899G>T p.G300V Chr.10: 110119503-110119503 on Build GRCh38
3434083 c.792C>T p.D264D Chr.10: 110119285-110119285 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760330800 Chr.10: 110119308-110119310 on Build GRCh38
rs750379380 Chr.10: 110119493-110119493 on Build GRCh38
rs771306473 Chr.10: 110119582-110119582 on Build GRCh38
rs556055990 Chr.10: 110119191-110119191 on Build GRCh38
rs375290612 Chr.10: 110119218-110119218 on Build GRCh38
rs971844490 Chr.10: 110119407-110119407 on Build GRCh38
rs755933623 Chr.10: 110119311-110119311 on Build GRCh38
rs775766673 Chr.10: 110119220-110119220 on Build GRCh38
rs764861697 Chr.10: 110119274-110119274 on Build GRCh38
rs754628673 Chr.10: 110119186-110119186 on Build GRCh38
rs747577326 Chr.10: 110119581-110119581 on Build GRCh38
rs766784177 Chr.10: 110119406-110119406 on Build GRCh38
rs370227323 Chr.10: 110119240-110119240 on Build GRCh38
rs751121786 Chr.10: 110119459-110119459 on Build GRCh38
rs770803143 Chr.10: 110119204-110119204 on Build GRCh38
rs779929567 Chr.10: 110119503-110119503 on Build GRCh38
rs879512943 Chr.10: 110119320-110119320 on Build GRCh38
rs548954210 Chr.10: 110119357-110119357 on Build GRCh38
rs369831974 Chr.10: 110119416-110119416 on Build GRCh38
rs543910414 Chr.10: 110119413-110119413 on Build GRCh38
rs761016545 Chr.10: 110119397-110119397 on Build GRCh38
rs566980206 Chr.10: 110119144-110119144 on Build GRCh38
rs772278639 Chr.10: 110119563-110119563 on Build GRCh38
rs749003106 Chr.10: 110119520-110119520 on Build GRCh38
rs199951875 Chr.10: 110119208-110119208 on Build GRCh38
rs981470000 Chr.10: 110119573-110119573 on Build GRCh38
rs576580333 Chr.10: 110119371-110119371 on Build GRCh38
rs920218365 Chr.10: 110119147-110119147 on Build GRCh38
rs763619672 Chr.10: 110119282-110119282 on Build GRCh38
rs543005015 Chr.10: 110119281-110119281 on Build GRCh38
rs1045849873 Chr.10: 110119221-110119221 on Build GRCh38
rs768795762 Chr.10: 110119226-110119226 on Build GRCh38
rs752394118 Chr.10: 110119278-110119278 on Build GRCh38
rs745630145 Chr.10: 110119212-110119212 on Build GRCh38
rs773847777 Chr.10: 110119381-110119381 on Build GRCh38
rs898391855 Chr.10: 110119469-110119469 on Build GRCh38
rs751032166 Chr.10: 110119284-110119284 on Build GRCh38
rs987302841 Chr.10: 110119107-110119107 on Build GRCh38
rs772761212 Chr.10: 110119375-110119375 on Build GRCh38
rs778608761 Chr.10: 110119188-110119188 on Build GRCh38
rs55763936 Chr.10: 110119269-110119269 on Build GRCh38
rs141823944 Chr.10: 110119499-110119499 on Build GRCh38
rs773363394 Chr.10: 110119579-110119579 on Build GRCh38
rs184766988 Chr.10: 110119441-110119441 on Build GRCh38
rs73352915 Chr.10: 110119122-110119122 on Build GRCh38
rs777096336 Chr.10: 110119584-110119584 on Build GRCh38
rs559948935 Chr.10: 110119538-110119538 on Build GRCh38
rs768718996 Chr.10: 110119358-110119358 on Build GRCh38
rs781485508 Chr.10: 110119277-110119277 on Build GRCh38
rs533538256 Chr.10: 110119106-110119106 on Build GRCh38
rs746958016 Chr.10: 110119200-110119200 on Build GRCh38
rs762701623 Chr.10: 110119436-110119436 on Build GRCh38
rs745708662 Chr.10: 110119308-110119308 on Build GRCh38
rs755298352 Chr.10: 110119527-110119527 on Build GRCh38
rs779656216 Chr.10: 110119323-110119323 on Build GRCh38
rs567558965 Chr.10: 110119574-110119574 on Build GRCh38
rs767510759 Chr.10: 110119486-110119486 on Build GRCh38
rs538498655 Chr.10: 110119255-110119255 on Build GRCh38
rs748302229 Chr.10: 110119540-110119540 on Build GRCh38
rs765964612 Chr.10: 110119391-110119393 on Build GRCh38
rs201055519 Chr.10: 110119271-110119271 on Build GRCh38
rs930321630 Chr.10: 110119154-110119154 on Build GRCh38
rs757228397 Chr.10: 110119293-110119293 on Build GRCh38
rs376190196 Chr.10: 110119480-110119480 on Build GRCh38
rs538893180 Chr.10: 110119108-110119108 on Build GRCh38
rs1033055536 Chr.10: 110119262-110119262 on Build GRCh38
rs781086274 Chr.10: 110119299-110119299 on Build GRCh38
rs537564625 Chr.10: 110119189-110119189 on Build GRCh38
rs774612012 Chr.10: 110119360-110119360 on Build GRCh38
rs933703374 Chr.10: 110119401-110119401 on Build GRCh38
rs763009836 Chr.10: 110119225-110119225 on Build GRCh38

View Full Product Details