Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 119034872-119035411 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EIF3A
Entrez Gene ID
8661
Gene Name
eukaryotic translation initiation factor 3 subunit A
Gene Aliases
EIF3, EIF3S10, P167, TIF32, eIF3-p170, eIF3-theta, p180, p185
UniGene
Hs.523299
Gene Chromosome Location
Chr.10: 119035029-119080822 on Build GRCh38
Transcripts
NM_003750.2

SNP Detail

SNP ID Chromosome Location
rs1020516605 Chr.10: 119034912-119034912 on Build GRCh38
rs1040024828 Chr.10: 119035120-119035120 on Build GRCh38
rs528379109 Chr.10: 119035252-119035252 on Build GRCh38
rs922682946 Chr.10: 119035229-119035235 on Build GRCh38
rs920035077 Chr.10: 119035009-119035009 on Build GRCh38
rs930477203 Chr.10: 119035060-119035060 on Build GRCh38
rs71480777 Chr.10: 119034936-119034937 on Build GRCh38
rs113627946 Chr.10: 119035375-119035375 on Build GRCh38
rs989666344 Chr.10: 119035188-119035188 on Build GRCh38
rs567907143 Chr.10: 119035307-119035307 on Build GRCh38
rs958742073 Chr.10: 119034936-119034936 on Build GRCh38
rs1014335007 Chr.10: 119035333-119035333 on Build GRCh38
rs569392718 Chr.10: 119035353-119035353 on Build GRCh38
rs940597475 Chr.10: 119035113-119035113 on Build GRCh38
rs1031146036 Chr.10: 119035388-119035388 on Build GRCh38
rs967642423 Chr.10: 119034916-119034916 on Build GRCh38
rs547940813 Chr.10: 119035257-119035257 on Build GRCh38
rs998531020 Chr.10: 119035380-119035380 on Build GRCh38
rs546695622 Chr.10: 119035221-119035221 on Build GRCh38
rs867823299 Chr.10: 119035087-119035087 on Build GRCh38
rs1014034199 Chr.10: 119034950-119034950 on Build GRCh38
rs999650720 Chr.10: 119035119-119035119 on Build GRCh38
rs1031094016 Chr.10: 119035144-119035144 on Build GRCh38
rs530666834 Chr.10: 119035352-119035352 on Build GRCh38
rs760463488 Chr.10: 119034929-119034929 on Build GRCh38
rs911243672 Chr.10: 119035208-119035211 on Build GRCh38
rs991604555 Chr.10: 119034976-119034976 on Build GRCh38
rs894176629 Chr.10: 119035330-119035330 on Build GRCh38
rs559570125 Chr.10: 119035225-119035225 on Build GRCh38
rs371698144 Chr.10: 119035091-119035091 on Build GRCh38
rs780483346 Chr.10: 119035042-119035042 on Build GRCh38
rs201811447 Chr.10: 119034968-119034969 on Build GRCh38
rs1055706872 Chr.10: 119035309-119035309 on Build GRCh38
rs1049408080 Chr.10: 119035062-119035062 on Build GRCh38
rs976904619 Chr.10: 119035227-119035227 on Build GRCh38
rs1045363580 Chr.10: 119035351-119035351 on Build GRCh38
rs547918213 Chr.10: 119034933-119034934 on Build GRCh38
rs970959124 Chr.10: 119035052-119035052 on Build GRCh38
rs578853 Chr.10: 119035161-119035161 on Build GRCh38
rs747247803 Chr.10: 119035269-119035269 on Build GRCh38
rs981398522 Chr.10: 119034923-119034923 on Build GRCh38
rs533336401 Chr.10: 119034925-119034925 on Build GRCh38
rs930043729 Chr.10: 119035245-119035245 on Build GRCh38

View Full Product Details