Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 113755190-113755747 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PLEKHS1
Entrez Gene ID
79949
Gene Name
pleckstrin homology domain containing S1
Gene Aliases
C10orf81, HEL185, bA211N11.2
UniGene
Hs.228320
Gene Chromosome Location
Chr.10: 113751262-113782433 on Build GRCh38
Transcripts
NM_001193434.1 NM_001193435.1 NM_024889.4 XM_005270162.2 XM_005270163.2 XM_011540171.1 XM_011540172.1 XM_011540173.1 XM_011540174.2 XM_011540175.1 XM_011540176.1 XM_017016666.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7149958 c.28+7G>T p.? Chr.10: 113755312-113755312 on Build GRCh38
3434233 c.25C>T p.P9S Chr.10: 113755302-113755302 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs567338252 Chr.10: 113755325-113755325 on Build GRCh38
rs149550272 Chr.10: 113755577-113755577 on Build GRCh38
rs898426405 Chr.10: 113755285-113755285 on Build GRCh38
rs768810216 Chr.10: 113755439-113755439 on Build GRCh38
rs112939714 Chr.10: 113755299-113755299 on Build GRCh38
rs933732667 Chr.10: 113755410-113755411 on Build GRCh38
rs112105976 Chr.10: 113755699-113755699 on Build GRCh38
rs932459703 Chr.10: 113755222-113755222 on Build GRCh38
rs573206858 Chr.10: 113755449-113755449 on Build GRCh38
rs7920463 Chr.10: 113755453-113755453 on Build GRCh38
rs553601270 Chr.10: 113755409-113755409 on Build GRCh38
rs745689815 Chr.10: 113755324-113755324 on Build GRCh38
rs778992517 Chr.10: 113755296-113755296 on Build GRCh38
rs895027498 Chr.10: 113755593-113755593 on Build GRCh38
rs753904465 Chr.10: 113755271-113755271 on Build GRCh38
rs3981351 Chr.10: 113755361-113755361 on Build GRCh38
rs113287812 Chr.10: 113755474-113755474 on Build GRCh38
rs779928788 Chr.10: 113755244-113755247 on Build GRCh38
rs773673727 Chr.10: 113755357-113755357 on Build GRCh38
rs977573381 Chr.10: 113755372-113755372 on Build GRCh38
rs773842189 Chr.10: 113755353-113755353 on Build GRCh38
rs767964255 Chr.10: 113755261-113755261 on Build GRCh38
rs751282020 Chr.10: 113755229-113755229 on Build GRCh38
rs957349010 Chr.10: 113755360-113755360 on Build GRCh38
rs186726207 Chr.10: 113755632-113755632 on Build GRCh38
rs780737262 Chr.10: 113755270-113755270 on Build GRCh38
rs4918866 Chr.10: 113755647-113755647 on Build GRCh38
rs907327270 Chr.10: 113755637-113755637 on Build GRCh38
rs548845116 Chr.10: 113755226-113755226 on Build GRCh38
rs761575986 Chr.10: 113755354-113755354 on Build GRCh38
rs971432539 Chr.10: 113755368-113755368 on Build GRCh38
rs548940755 Chr.10: 113755706-113755706 on Build GRCh38
rs756640873 Chr.10: 113755269-113755269 on Build GRCh38
rs757947913 Chr.10: 113755701-113755701 on Build GRCh38
rs990106079 Chr.10: 113755603-113755603 on Build GRCh38
rs946257391 Chr.10: 113755382-113755382 on Build GRCh38
rs748472338 Chr.10: 113755300-113755300 on Build GRCh38
rs1036893399 Chr.10: 113755563-113755563 on Build GRCh38
rs954298934 Chr.10: 113755712-113755712 on Build GRCh38
rs781093639 Chr.10: 113755312-113755312 on Build GRCh38
rs1034832409 Chr.10: 113755564-113755564 on Build GRCh38
rs537676649 Chr.10: 113755346-113755346 on Build GRCh38
rs762077450 Chr.10: 113755238-113755238 on Build GRCh38
rs890847946 Chr.10: 113755254-113755254 on Build GRCh38
rs1001648813 Chr.10: 113755530-113755530 on Build GRCh38
rs778554710 Chr.10: 113755466-113755466 on Build GRCh38
rs894463579 Chr.10: 113755339-113755339 on Build GRCh38
rs563092617 Chr.10: 113755576-113755576 on Build GRCh38
rs767097546 Chr.10: 113755355-113755355 on Build GRCh38
rs374937238 Chr.10: 113755334-113755334 on Build GRCh38
rs370175748 Chr.10: 113755311-113755311 on Build GRCh38
rs7920441 Chr.10: 113755404-113755404 on Build GRCh38
rs939721381 Chr.10: 113755527-113755527 on Build GRCh38
rs138237417 Chr.10: 113755475-113755475 on Build GRCh38
rs990806839 Chr.10: 113755411-113755411 on Build GRCh38
rs1036840929 Chr.10: 113755279-113755279 on Build GRCh38
rs898311649 Chr.10: 113755570-113755570 on Build GRCh38
rs750942741 Chr.10: 113755268-113755268 on Build GRCh38
rs748779808 Chr.10: 113755343-113755343 on Build GRCh38
rs994417926 Chr.10: 113755477-113755477 on Build GRCh38
rs1045704440 Chr.10: 113755612-113755612 on Build GRCh38
rs971469277 Chr.10: 113755274-113755274 on Build GRCh38
rs1031840392 Chr.10: 113755649-113755649 on Build GRCh38
rs1013334894 Chr.10: 113755594-113755594 on Build GRCh38
rs980520842 Chr.10: 113755284-113755284 on Build GRCh38
rs755124446 Chr.10: 113755290-113755290 on Build GRCh38
rs920910637 Chr.10: 113755215-113755215 on Build GRCh38
rs571072329 Chr.10: 113755385-113755385 on Build GRCh38
rs377595573 Chr.10: 113755253-113755253 on Build GRCh38
rs1024111353 Chr.10: 113755483-113755483 on Build GRCh38

View Full Product Details