Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 33091996-33092592 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CSTF3
Entrez Gene ID
1479
Gene Name
cleavage stimulation factor subunit 3
Gene Aliases
CSTF-77
UniGene
Hs.44402
Gene Chromosome Location
Chr.11: 33084584-33161491 on Build GRCh38
Transcripts
NM_001326.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4835976 c.1376-1G>A p.? Chr.11: 33092341-33092341 on Build GRCh38
5640149 c.1435G>A p.E479K Chr.11: 33092281-33092281 on Build GRCh38
3809108 c.1414A>C p.T472P Chr.11: 33092302-33092302 on Build GRCh38
4836262 c.1402G>C p.E468Q Chr.11: 33092314-33092314 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs922419315 Chr.11: 33092456-33092456 on Build GRCh38
rs1010554351 Chr.11: 33092128-33092128 on Build GRCh38
rs764501503 Chr.11: 33092247-33092247 on Build GRCh38
rs12276773 Chr.11: 33092156-33092156 on Build GRCh38
rs761564137 Chr.11: 33092385-33092385 on Build GRCh38
rs78493253 Chr.11: 33092066-33092066 on Build GRCh38
rs1010905404 Chr.11: 33092148-33092148 on Build GRCh38
rs765871237 Chr.11: 33092103-33092108 on Build GRCh38
rs771602234 Chr.11: 33092376-33092376 on Build GRCh38
rs749680289 Chr.11: 33092352-33092352 on Build GRCh38
rs893466028 Chr.11: 33092119-33092119 on Build GRCh38
rs374946238 Chr.11: 33092205-33092205 on Build GRCh38
rs1040842928 Chr.11: 33092027-33092027 on Build GRCh38
rs781528275 Chr.11: 33092273-33092273 on Build GRCh38
rs978922389 Chr.11: 33092184-33092184 on Build GRCh38
rs373923109 Chr.11: 33092541-33092541 on Build GRCh38
rs571388005 Chr.11: 33092311-33092311 on Build GRCh38
rs1042173508 Chr.11: 33092133-33092133 on Build GRCh38
rs1057176317 Chr.11: 33092564-33092564 on Build GRCh38
rs922568581 Chr.11: 33092044-33092044 on Build GRCh38
rs113144162 Chr.11: 33092364-33092365 on Build GRCh38
rs775074543 Chr.11: 33092377-33092377 on Build GRCh38
rs755149209 Chr.11: 33092265-33092265 on Build GRCh38
rs760332671 Chr.11: 33092378-33092378 on Build GRCh38
rs763971493 Chr.11: 33092231-33092231 on Build GRCh38
rs747063822 Chr.11: 33092262-33092262 on Build GRCh38
rs987209279 Chr.11: 33092334-33092334 on Build GRCh38
rs566888133 Chr.11: 33092024-33092024 on Build GRCh38
rs987523702 Chr.11: 33092026-33092026 on Build GRCh38
rs1036199327 Chr.11: 33092042-33092044 on Build GRCh38
rs908607188 Chr.11: 33092022-33092022 on Build GRCh38
rs188385129 Chr.11: 33092211-33092211 on Build GRCh38
rs999050451 Chr.11: 33092201-33092201 on Build GRCh38
rs769999645 Chr.11: 33092326-33092326 on Build GRCh38
rs757217023 Chr.11: 33092249-33092249 on Build GRCh38
rs767694246 Chr.11: 33092554-33092554 on Build GRCh38
rs551421480 Chr.11: 33092484-33092484 on Build GRCh38
rs764375258 Chr.11: 33092363-33092364 on Build GRCh38
rs771452356 Chr.11: 33092357-33092357 on Build GRCh38
rs940622509 Chr.11: 33092034-33092034 on Build GRCh38
rs946394287 Chr.11: 33092533-33092533 on Build GRCh38
rs34354154 Chr.11: 33092183-33092184 on Build GRCh38
rs895791724 Chr.11: 33092120-33092120 on Build GRCh38
rs773633893 Chr.11: 33092339-33092339 on Build GRCh38
rs1044866481 Chr.11: 33092447-33092447 on Build GRCh38
rs73482844 Chr.11: 33092403-33092403 on Build GRCh38
rs757429737 Chr.11: 33092485-33092485 on Build GRCh38
rs976593657 Chr.11: 33092438-33092438 on Build GRCh38
rs371063770 Chr.11: 33092388-33092388 on Build GRCh38
rs376585351 Chr.11: 33092384-33092384 on Build GRCh38
rs571373722 Chr.11: 33092527-33092527 on Build GRCh38
rs542255754 Chr.11: 33092250-33092250 on Build GRCh38
rs374330250 Chr.11: 33092394-33092394 on Build GRCh38
rs1042059616 Chr.11: 33092537-33092537 on Build GRCh38
rs952839299 Chr.11: 33092360-33092360 on Build GRCh38
rs915134143 Chr.11: 33092507-33092507 on Build GRCh38
rs748406781 Chr.11: 33092316-33092316 on Build GRCh38
rs756984161 Chr.11: 33092067-33092072 on Build GRCh38
rs1048502957 Chr.11: 33092086-33092086 on Build GRCh38
rs985800153 Chr.11: 33092501-33092501 on Build GRCh38
rs7950763 Chr.11: 33092107-33092107 on Build GRCh38
rs968907122 Chr.11: 33092170-33092170 on Build GRCh38
rs896105222 Chr.11: 33092212-33092212 on Build GRCh38
rs763726992 Chr.11: 33092380-33092380 on Build GRCh38
rs143361944 Chr.11: 33092085-33092086 on Build GRCh38
rs1023175477 Chr.11: 33092359-33092359 on Build GRCh38
rs1013595978 Chr.11: 33092300-33092300 on Build GRCh38
rs932467609 Chr.11: 33092467-33092467 on Build GRCh38
rs71034650 Chr.11: 33092371-33092372 on Build GRCh38
rs562458789 Chr.11: 33092371-33092371 on Build GRCh38
rs386373520 Chr.11: 33092365-33092366 on Build GRCh38
rs774884565 Chr.11: 33092364-33092364 on Build GRCh38
rs961422610 Chr.11: 33092410-33092410 on Build GRCh38

View Full Product Details