Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 33025291-33025876 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DEPDC7
Entrez Gene ID
91614
Gene Name
DEP domain containing 7
Gene Aliases
TR2, dJ85M6.4
UniGene
Hs.280990
Gene Chromosome Location
Chr.11: 33015864-33033582 on Build GRCh38
Transcripts
NM_001077242.1 NM_139160.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5030509 c.277G>A p.A93T Chr.11: 33025862-33025862 on Build GRCh38
4483941 c.247C>T p.R83* Chr.11: 33025859-33025859 on Build GRCh38
1475379 c.163C>T p.R55* Chr.11: 33025748-33025748 on Build GRCh38
5838432 c.94C>T p.P32S Chr.11: 33025679-33025679 on Build GRCh38
6642723 c.80C>T p.T27M Chr.11: 33025692-33025692 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs867836227 Chr.11: 33025424-33025424 on Build GRCh38
rs1012222916 Chr.11: 33025512-33025512 on Build GRCh38
rs1021337485 Chr.11: 33025653-33025653 on Build GRCh38
rs746938818 Chr.11: 33025749-33025749 on Build GRCh38
rs887776444 Chr.11: 33025815-33025815 on Build GRCh38
rs552446140 Chr.11: 33025758-33025758 on Build GRCh38
rs369107128 Chr.11: 33025325-33025325 on Build GRCh38
rs780581236 Chr.11: 33025671-33025671 on Build GRCh38
rs779134492 Chr.11: 33025843-33025843 on Build GRCh38
rs760564144 Chr.11: 33025693-33025694 on Build GRCh38
rs369053359 Chr.11: 33025763-33025763 on Build GRCh38
rs561420236 Chr.11: 33025619-33025619 on Build GRCh38
rs375593314 Chr.11: 33025666-33025666 on Build GRCh38
rs749968641 Chr.11: 33025640-33025640 on Build GRCh38
rs760595441 Chr.11: 33025618-33025618 on Build GRCh38
rs749424506 Chr.11: 33025738-33025738 on Build GRCh38
rs752126886 Chr.11: 33025778-33025778 on Build GRCh38
rs745587922 Chr.11: 33025665-33025665 on Build GRCh38
rs746084543 Chr.11: 33025464-33025464 on Build GRCh38
rs910402509 Chr.11: 33025832-33025832 on Build GRCh38
rs138325318 Chr.11: 33025782-33025782 on Build GRCh38
rs958821425 Chr.11: 33025516-33025516 on Build GRCh38
rs767591203 Chr.11: 33025652-33025654 on Build GRCh38
rs753468814 Chr.11: 33025836-33025836 on Build GRCh38
rs775534730 Chr.11: 33025610-33025610 on Build GRCh38
rs914221046 Chr.11: 33025339-33025339 on Build GRCh38
rs748018531 Chr.11: 33025848-33025848 on Build GRCh38
rs753497220 Chr.11: 33025621-33025621 on Build GRCh38
rs901679227 Chr.11: 33025581-33025581 on Build GRCh38
rs755481363 Chr.11: 33025797-33025797 on Build GRCh38
rs868433858 Chr.11: 33025788-33025788 on Build GRCh38
rs547941463 Chr.11: 33025574-33025574 on Build GRCh38
rs527684582 Chr.11: 33025376-33025376 on Build GRCh38
rs768733647 Chr.11: 33025847-33025847 on Build GRCh38
rs778104021 Chr.11: 33025830-33025830 on Build GRCh38
rs74559182 Chr.11: 33025822-33025822 on Build GRCh38
rs997320165 Chr.11: 33025588-33025588 on Build GRCh38
rs745881333 Chr.11: 33025846-33025846 on Build GRCh38
rs368794067 Chr.11: 33025691-33025691 on Build GRCh38
rs749457582 Chr.11: 33025833-33025833 on Build GRCh38
rs939496250 Chr.11: 33025447-33025447 on Build GRCh38
rs888734692 Chr.11: 33025443-33025443 on Build GRCh38
rs887776562 Chr.11: 33025544-33025544 on Build GRCh38
rs1041461749 Chr.11: 33025421-33025421 on Build GRCh38
rs765864249 Chr.11: 33025772-33025772 on Build GRCh38
rs777811808 Chr.11: 33025730-33025730 on Build GRCh38
rs1056650836 Chr.11: 33025452-33025452 on Build GRCh38
rs754602099 Chr.11: 33025854-33025855 on Build GRCh38
rs200073795 Chr.11: 33025724-33025724 on Build GRCh38
rs924146176 Chr.11: 33025442-33025442 on Build GRCh38
rs757478085 Chr.11: 33025842-33025842 on Build GRCh38
rs756375437 Chr.11: 33025728-33025728 on Build GRCh38
rs977580251 Chr.11: 33025692-33025692 on Build GRCh38
rs945817357 Chr.11: 33025360-33025360 on Build GRCh38
rs780288746 Chr.11: 33025515-33025515 on Build GRCh38
rs939528353 Chr.11: 33025709-33025709 on Build GRCh38
rs957553535 Chr.11: 33025819-33025819 on Build GRCh38
rs749314158 Chr.11: 33025693-33025693 on Build GRCh38
rs776495819 Chr.11: 33025754-33025754 on Build GRCh38
rs79199473 Chr.11: 33025697-33025697 on Build GRCh38
rs769954673 Chr.11: 33025467-33025467 on Build GRCh38
rs761527124 Chr.11: 33025625-33025625 on Build GRCh38
rs775626512 Chr.11: 33025748-33025748 on Build GRCh38
rs374733737 Chr.11: 33025757-33025757 on Build GRCh38
rs764736913 Chr.11: 33025628-33025628 on Build GRCh38
rs768643664 Chr.11: 33025753-33025753 on Build GRCh38
rs771111220 Chr.11: 33025745-33025745 on Build GRCh38
rs757636821 Chr.11: 33025324-33025324 on Build GRCh38
rs765946746 Chr.11: 33025808-33025809 on Build GRCh38
rs201178241 Chr.11: 33025561-33025561 on Build GRCh38
rs767956674 Chr.11: 33025803-33025803 on Build GRCh38
rs1047038350 Chr.11: 33025433-33025433 on Build GRCh38
rs781533300 Chr.11: 33025699-33025699 on Build GRCh38
rs773107100 Chr.11: 33025676-33025676 on Build GRCh38
rs1014970362 Chr.11: 33025559-33025559 on Build GRCh38
rs1005416744 Chr.11: 33025550-33025550 on Build GRCh38
rs1049298013 Chr.11: 33025759-33025759 on Build GRCh38
rs570369904 Chr.11: 33025555-33025555 on Build GRCh38
rs772165913 Chr.11: 33025609-33025609 on Build GRCh38
rs533528680 Chr.11: 33025823-33025823 on Build GRCh38
rs200057806 Chr.11: 33025767-33025767 on Build GRCh38

View Full Product Details