Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 22622703-22623251 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FANCF
Entrez Gene ID
2188
Gene Name
Fanconi anemia complementation group F
Gene Aliases
FAF
UniGene
Hs.632151
Gene Chromosome Location
Chr.11: 22622533-22625841 on Build GRCh38
Transcripts
NM_022725.3

SNP Detail

SNP ID Chromosome Location
rs10500938 Chr.11: 22623057-22623057 on Build GRCh38
rs555654133 Chr.11: 22622772-22622772 on Build GRCh38
rs1025850797 Chr.11: 22622910-22622910 on Build GRCh38
rs34198575 Chr.11: 22623154-22623155 on Build GRCh38
rs923194357 Chr.11: 22622736-22622736 on Build GRCh38
rs886048153 Chr.11: 22623177-22623177 on Build GRCh38
rs572694104 Chr.11: 22622777-22622777 on Build GRCh38
rs898565751 Chr.11: 22623218-22623218 on Build GRCh38
rs186333671 Chr.11: 22623226-22623226 on Build GRCh38
rs868634019 Chr.11: 22622763-22622763 on Build GRCh38
rs942675219 Chr.11: 22623176-22623176 on Build GRCh38
rs933206380 Chr.11: 22623170-22623170 on Build GRCh38
rs961929762 Chr.11: 22622816-22622816 on Build GRCh38
rs542719930 Chr.11: 22623032-22623032 on Build GRCh38
rs886048152 Chr.11: 22623161-22623161 on Build GRCh38
rs965847461 Chr.11: 22623222-22623222 on Build GRCh38
rs947080653 Chr.11: 22623049-22623049 on Build GRCh38
rs965462899 Chr.11: 22622980-22622980 on Build GRCh38
rs1040193693 Chr.11: 22622728-22622728 on Build GRCh38
rs570056034 Chr.11: 22623207-22623207 on Build GRCh38
rs913742659 Chr.11: 22623152-22623152 on Build GRCh38
rs976338984 Chr.11: 22623007-22623007 on Build GRCh38
rs45553340 Chr.11: 22622807-22622807 on Build GRCh38
rs528951933 Chr.11: 22623018-22623018 on Build GRCh38
rs45624941 Chr.11: 22623037-22623040 on Build GRCh38
rs994385621 Chr.11: 22622847-22622847 on Build GRCh38
rs1006908460 Chr.11: 22623210-22623210 on Build GRCh38
rs578259902 Chr.11: 22622907-22622907 on Build GRCh38
rs1051008203 Chr.11: 22622773-22622773 on Build GRCh38
rs1057485119 Chr.11: 22623096-22623096 on Build GRCh38
rs980487442 Chr.11: 22623054-22623054 on Build GRCh38
rs571200561 Chr.11: 22623174-22623174 on Build GRCh38
rs1018802413 Chr.11: 22622975-22622976 on Build GRCh38
rs1052722344 Chr.11: 22622753-22622753 on Build GRCh38
rs563296397 Chr.11: 22623017-22623017 on Build GRCh38
rs774789696 Chr.11: 22622765-22622765 on Build GRCh38
rs1044165541 Chr.11: 22622769-22622769 on Build GRCh38
rs923090835 Chr.11: 22623030-22623030 on Build GRCh38
rs891105129 Chr.11: 22623175-22623175 on Build GRCh38
rs557247779 Chr.11: 22622956-22622956 on Build GRCh38
rs1024611706 Chr.11: 22622990-22622990 on Build GRCh38
rs866690784 Chr.11: 22622737-22622737 on Build GRCh38
rs551211564 Chr.11: 22623125-22623125 on Build GRCh38
rs181285048 Chr.11: 22623202-22623202 on Build GRCh38
rs190919478 Chr.11: 22622845-22622845 on Build GRCh38
rs892376002 Chr.11: 22622764-22622764 on Build GRCh38
rs938424178 Chr.11: 22623070-22623070 on Build GRCh38
rs914487796 Chr.11: 22623044-22623044 on Build GRCh38
rs536079455 Chr.11: 22623203-22623203 on Build GRCh38
rs886048151 Chr.11: 22623151-22623151 on Build GRCh38
rs898601881 Chr.11: 22622905-22622905 on Build GRCh38
rs559054290 Chr.11: 22623056-22623056 on Build GRCh38
rs771298710 Chr.11: 22622723-22622723 on Build GRCh38
rs1052304887 Chr.11: 22622759-22622759 on Build GRCh38
rs925110798 Chr.11: 22622734-22622734 on Build GRCh38
rs951605084 Chr.11: 22622942-22622942 on Build GRCh38
rs934580777 Chr.11: 22623043-22623044 on Build GRCh38
rs886048149 Chr.11: 22622990-22622990 on Build GRCh38
rs763604119 Chr.11: 22623155-22623155 on Build GRCh38
rs1027230581 Chr.11: 22622848-22622848 on Build GRCh38
rs886048150 Chr.11: 22623054-22623054 on Build GRCh38
rs987932925 Chr.11: 22623088-22623088 on Build GRCh38
rs1018795675 Chr.11: 22622887-22622887 on Build GRCh38
rs760103091 Chr.11: 22622802-22622802 on Build GRCh38

View Full Product Details