Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 47245144-47245741 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACP2
Entrez Gene ID
53
Gene Name
acid phosphatase 2, lysosomal
Gene Aliases
LAP
UniGene
Hs.532492
Gene Chromosome Location
Chr.11: 47239302-47248847 on Build GRCh38
Transcripts
NM_001302489.1 NM_001302490.1 NM_001302491.1 NM_001302492.1 NM_001610.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1354040 c.581G>T p.G194V Chr.11: 47245363-47245363 on Build GRCh38
5999804 c.537T>C p.S179S Chr.11: 47245486-47245486 on Build GRCh38
4658929 c.417T>C p.I139I Chr.11: 47245715-47245715 on Build GRCh38
1354041 c.542G>A p.R181Q Chr.11: 47245481-47245481 on Build GRCh38
6023299 c.616G>C p.V206L Chr.11: 47245328-47245328 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773070237 Chr.11: 47245429-47245429 on Build GRCh38
rs778713650 Chr.11: 47245288-47245288 on Build GRCh38
rs751568789 Chr.11: 47245231-47245231 on Build GRCh38
rs770219586 Chr.11: 47245171-47245171 on Build GRCh38
rs751211991 Chr.11: 47245683-47245683 on Build GRCh38
rs558648785 Chr.11: 47245216-47245216 on Build GRCh38
rs748939874 Chr.11: 47245440-47245440 on Build GRCh38
rs369792561 Chr.11: 47245510-47245510 on Build GRCh38
rs2242261 Chr.11: 47245257-47245257 on Build GRCh38
rs185966000 Chr.11: 47245344-47245344 on Build GRCh38
rs748424544 Chr.11: 47245170-47245170 on Build GRCh38
rs764658808 Chr.11: 47245469-47245469 on Build GRCh38
rs776348174 Chr.11: 47245337-47245337 on Build GRCh38
rs745572250 Chr.11: 47245403-47245403 on Build GRCh38
rs779668251 Chr.11: 47245422-47245422 on Build GRCh38
rs201635592 Chr.11: 47245313-47245313 on Build GRCh38
rs373531382 Chr.11: 47245445-47245445 on Build GRCh38
rs760364954 Chr.11: 47245210-47245210 on Build GRCh38
rs761406762 Chr.11: 47245188-47245188 on Build GRCh38
rs139616510 Chr.11: 47245204-47245204 on Build GRCh38
rs998259629 Chr.11: 47245285-47245285 on Build GRCh38
rs768511268 Chr.11: 47245439-47245439 on Build GRCh38
rs373260498 Chr.11: 47245647-47245647 on Build GRCh38
rs751805071 Chr.11: 47245368-47245368 on Build GRCh38
rs562621346 Chr.11: 47245515-47245515 on Build GRCh38
rs757842561 Chr.11: 47245409-47245409 on Build GRCh38
rs200104071 Chr.11: 47245458-47245458 on Build GRCh38
rs770561560 Chr.11: 47245182-47245182 on Build GRCh38
rs771660391 Chr.11: 47245292-47245292 on Build GRCh38
rs1013575677 Chr.11: 47245250-47245250 on Build GRCh38
rs199680796 Chr.11: 47245268-47245268 on Build GRCh38
rs760789795 Chr.11: 47245537-47245537 on Build GRCh38
rs181634705 Chr.11: 47245281-47245281 on Build GRCh38
rs758006727 Chr.11: 47245503-47245503 on Build GRCh38
rs767481773 Chr.11: 47245463-47245463 on Build GRCh38
rs10838677 Chr.11: 47245528-47245528 on Build GRCh38
rs777395443 Chr.11: 47245400-47245400 on Build GRCh38
rs759247125 Chr.11: 47245705-47245705 on Build GRCh38
rs755119226 Chr.11: 47245237-47245237 on Build GRCh38
rs754443245 Chr.11: 47245479-47245479 on Build GRCh38
rs942193347 Chr.11: 47245505-47245505 on Build GRCh38
rs766697551 Chr.11: 47245668-47245668 on Build GRCh38
rs765762417 Chr.11: 47245482-47245482 on Build GRCh38
rs757729647 Chr.11: 47245380-47245380 on Build GRCh38
rs763351467 Chr.11: 47245666-47245666 on Build GRCh38
rs751221357 Chr.11: 47245567-47245567 on Build GRCh38
rs1034114790 Chr.11: 47245283-47245283 on Build GRCh38
rs550354036 Chr.11: 47245559-47245559 on Build GRCh38
rs754382383 Chr.11: 47245609-47245609 on Build GRCh38
rs768422993 Chr.11: 47245322-47245322 on Build GRCh38
rs773120760 Chr.11: 47245521-47245521 on Build GRCh38
rs912963658 Chr.11: 47245421-47245421 on Build GRCh38
rs749050118 Chr.11: 47245525-47245525 on Build GRCh38
rs375804831 Chr.11: 47245601-47245601 on Build GRCh38
rs773464046 Chr.11: 47245221-47245222 on Build GRCh38
rs780290242 Chr.11: 47245602-47245602 on Build GRCh38
rs775519096 Chr.11: 47245201-47245201 on Build GRCh38
rs982261479 Chr.11: 47245198-47245198 on Build GRCh38
rs775259733 Chr.11: 47245293-47245293 on Build GRCh38
rs754502962 Chr.11: 47245514-47245514 on Build GRCh38
rs768842094 Chr.11: 47245632-47245632 on Build GRCh38
rs780493231 Chr.11: 47245613-47245613 on Build GRCh38
rs560625251 Chr.11: 47245371-47245371 on Build GRCh38
rs143294431 Chr.11: 47245519-47245519 on Build GRCh38
rs754455970 Chr.11: 47245581-47245581 on Build GRCh38
rs781505049 Chr.11: 47245238-47245238 on Build GRCh38
rs1041750676 Chr.11: 47245618-47245618 on Build GRCh38
rs755988176 Chr.11: 47245587-47245587 on Build GRCh38
rs760842915 Chr.11: 47245467-47245467 on Build GRCh38
rs750030860 Chr.11: 47245487-47245487 on Build GRCh38
rs753301207 Chr.11: 47245256-47245256 on Build GRCh38
rs766545927 Chr.11: 47245362-47245362 on Build GRCh38
rs375915924 Chr.11: 47245529-47245529 on Build GRCh38
rs368941003 Chr.11: 47245620-47245620 on Build GRCh38
rs770648129 Chr.11: 47245465-47245465 on Build GRCh38
rs190450604 Chr.11: 47245183-47245183 on Build GRCh38
rs777271588 Chr.11: 47245542-47245542 on Build GRCh38
rs540880065 Chr.11: 47245370-47245370 on Build GRCh38
rs767175413 Chr.11: 47245722-47245722 on Build GRCh38
rs761985649 Chr.11: 47245657-47245657 on Build GRCh38
rs376709487 Chr.11: 47245663-47245663 on Build GRCh38
rs930207388 Chr.11: 47245208-47245208 on Build GRCh38
rs942975772 Chr.11: 47245548-47245548 on Build GRCh38
rs745313795 Chr.11: 47245290-47245290 on Build GRCh38
rs971336695 Chr.11: 47245258-47245258 on Build GRCh38
rs748992995 Chr.11: 47245179-47245179 on Build GRCh38
rs757664236 Chr.11: 47245481-47245481 on Build GRCh38
rs201770743 Chr.11: 47245444-47245444 on Build GRCh38
rs202079999 Chr.11: 47245541-47245541 on Build GRCh38
rs774410105 Chr.11: 47245454-47245454 on Build GRCh38
rs778684657 Chr.11: 47245384-47245384 on Build GRCh38
rs751006145 Chr.11: 47245430-47245430 on Build GRCh38
rs780750863 Chr.11: 47245582-47245582 on Build GRCh38
rs552764293 Chr.11: 47245424-47245424 on Build GRCh38
rs372851690 Chr.11: 47245633-47245633 on Build GRCh38
rs750729492 Chr.11: 47245556-47245556 on Build GRCh38
rs11039146 Chr.11: 47245211-47245211 on Build GRCh38
rs761597969 Chr.11: 47245338-47245338 on Build GRCh38
rs749071542 Chr.11: 47245600-47245600 on Build GRCh38
rs756678544 Chr.11: 47245539-47245539 on Build GRCh38
rs577134798 Chr.11: 47245375-47245375 on Build GRCh38
rs981895039 Chr.11: 47245491-47245491 on Build GRCh38
rs757969651 Chr.11: 47245205-47245205 on Build GRCh38
rs746485467 Chr.11: 47245513-47245513 on Build GRCh38
rs762468028 Chr.11: 47245549-47245549 on Build GRCh38
rs763254822 Chr.11: 47245558-47245558 on Build GRCh38
rs375076117 Chr.11: 47245658-47245658 on Build GRCh38
rs747709245 Chr.11: 47245218-47245218 on Build GRCh38
rs765671353 Chr.11: 47245551-47245551 on Build GRCh38
rs766599070 Chr.11: 47245225-47245225 on Build GRCh38
rs138260098 Chr.11: 47245566-47245566 on Build GRCh38
rs759101018 Chr.11: 47245193-47245193 on Build GRCh38
rs376413659 Chr.11: 47245178-47245178 on Build GRCh38
rs373389955 Chr.11: 47245595-47245595 on Build GRCh38
rs752621427 Chr.11: 47245586-47245586 on Build GRCh38
rs770651512 Chr.11: 47245287-47245287 on Build GRCh38

View Full Product Details