Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 68096856-68097190 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHKA
Entrez Gene ID
1119
Gene Name
choline kinase alpha
Gene Aliases
CHK, CK, CKI, EK
UniGene
Hs.77221
Gene Chromosome Location
Chr.11: 68052859-68121391 on Build GRCh38
Transcripts
NM_001277.2 NM_212469.1 XM_017017147.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4672165 c.436C>T p.R146W Chr.11: 68097045-68097045 on Build GRCh38
5619603 c.380G>A p.C127Y Chr.11: 68097101-68097101 on Build GRCh38
6070239 c.430C>T p.L144F Chr.11: 68097051-68097051 on Build GRCh38
5659059 c.425A>T p.K142I Chr.11: 68097056-68097056 on Build GRCh38
5505321 c.351A>G p.R117R Chr.11: 68097130-68097130 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749211062 Chr.11: 68097126-68097126 on Build GRCh38
rs892900675 Chr.11: 68096976-68096976 on Build GRCh38
rs375551488 Chr.11: 68097040-68097040 on Build GRCh38
rs779736042 Chr.11: 68097113-68097113 on Build GRCh38
rs201644597 Chr.11: 68097055-68097055 on Build GRCh38
rs767244257 Chr.11: 68096983-68096983 on Build GRCh38
rs747156380 Chr.11: 68097159-68097159 on Build GRCh38
rs751213726 Chr.11: 68096992-68096992 on Build GRCh38
rs17857113 Chr.11: 68097021-68097021 on Build GRCh38
rs1019950605 Chr.11: 68097004-68097004 on Build GRCh38
rs886222654 Chr.11: 68096946-68096946 on Build GRCh38
rs754949909 Chr.11: 68097127-68097127 on Build GRCh38
rs765529707 Chr.11: 68097060-68097060 on Build GRCh38
rs751166615 Chr.11: 68097102-68097102 on Build GRCh38
rs745737649 Chr.11: 68097007-68097007 on Build GRCh38
rs759877440 Chr.11: 68097059-68097059 on Build GRCh38
rs749196970 Chr.11: 68097020-68097020 on Build GRCh38
rs994955467 Chr.11: 68096927-68096927 on Build GRCh38
rs773313547 Chr.11: 68097031-68097031 on Build GRCh38
rs546088815 Chr.11: 68097164-68097164 on Build GRCh38
rs572084295 Chr.11: 68097076-68097076 on Build GRCh38
rs763398771 Chr.11: 68097081-68097081 on Build GRCh38
rs187957797 Chr.11: 68097032-68097032 on Build GRCh38
rs777815732 Chr.11: 68097136-68097136 on Build GRCh38
rs779976460 Chr.11: 68097016-68097016 on Build GRCh38
rs1019148520 Chr.11: 68096994-68096994 on Build GRCh38
rs537284288 Chr.11: 68096929-68096929 on Build GRCh38
rs777443234 Chr.11: 68096988-68096988 on Build GRCh38
rs769618494 Chr.11: 68097009-68097009 on Build GRCh38
rs748308087 Chr.11: 68096987-68096988 on Build GRCh38
rs777724194 Chr.11: 68097135-68097148 on Build GRCh38
rs747032997 Chr.11: 68097141-68097142 on Build GRCh38
rs769981779 Chr.11: 68096945-68096945 on Build GRCh38
rs370056742 Chr.11: 68096985-68096985 on Build GRCh38
rs184506502 Chr.11: 68096993-68096993 on Build GRCh38
rs776783159 Chr.11: 68096934-68096935 on Build GRCh38
rs763663084 Chr.11: 68097087-68097087 on Build GRCh38
rs375011666 Chr.11: 68097106-68097106 on Build GRCh38
rs767349832 Chr.11: 68097105-68097105 on Build GRCh38
rs199679026 Chr.11: 68097025-68097025 on Build GRCh38
rs139911822 Chr.11: 68097104-68097104 on Build GRCh38
rs781115042 Chr.11: 68097003-68097003 on Build GRCh38

View Full Product Details