Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 66345451-66346029 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
B4GAT1
Entrez Gene ID
11041
Gene Name
beta-1,4-glucuronyltransferase 1
Gene Aliases
B3GN-T1, B3GNT1, B3GNT6, BETA3GNTI, MDDGA13, iGAT, iGNT
UniGene
Hs.8526
Gene Chromosome Location
Chr.11: 66345372-66347690 on Build GRCh38
Transcripts
NM_006876.2

SNP Detail

SNP ID Chromosome Location
rs944499374 Chr.11: 66345614-66345614 on Build GRCh38
rs950537498 Chr.11: 66345812-66345812 on Build GRCh38
rs778363284 Chr.11: 66345806-66345806 on Build GRCh38
rs555584686 Chr.11: 66345593-66345593 on Build GRCh38
rs758646031 Chr.11: 66345738-66345738 on Build GRCh38
rs757331942 Chr.11: 66345967-66345967 on Build GRCh38
rs557755436 Chr.11: 66345793-66345793 on Build GRCh38
rs764013182 Chr.11: 66346000-66346000 on Build GRCh38
rs573756401 Chr.11: 66346002-66346002 on Build GRCh38
rs145270731 Chr.11: 66345682-66345684 on Build GRCh38
rs1038796940 Chr.11: 66345709-66345709 on Build GRCh38
rs974179465 Chr.11: 66345977-66345977 on Build GRCh38
rs1024714350 Chr.11: 66345633-66345633 on Build GRCh38
rs182104341 Chr.11: 66345727-66345727 on Build GRCh38
rs540174173 Chr.11: 66345866-66345866 on Build GRCh38
rs765758195 Chr.11: 66345720-66345720 on Build GRCh38
rs987294289 Chr.11: 66345819-66345819 on Build GRCh38
rs963160349 Chr.11: 66345959-66345959 on Build GRCh38
rs929244986 Chr.11: 66345584-66345584 on Build GRCh38
rs892748915 Chr.11: 66345921-66345921 on Build GRCh38
rs912639728 Chr.11: 66345823-66345830 on Build GRCh38
rs974505663 Chr.11: 66345500-66345500 on Build GRCh38
rs936902616 Chr.11: 66345508-66345508 on Build GRCh38
rs761563670 Chr.11: 66345914-66345914 on Build GRCh38
rs962752710 Chr.11: 66345497-66345497 on Build GRCh38
rs1039741581 Chr.11: 66345490-66345490 on Build GRCh38
rs944166352 Chr.11: 66345838-66345838 on Build GRCh38
rs767678516 Chr.11: 66345685-66345687 on Build GRCh38
rs141108052 Chr.11: 66345856-66345856 on Build GRCh38
rs1026669958 Chr.11: 66345766-66345766 on Build GRCh38
rs186852695 Chr.11: 66345555-66345555 on Build GRCh38
rs906312674 Chr.11: 66345669-66345669 on Build GRCh38
rs902178934 Chr.11: 66345788-66345788 on Build GRCh38
rs117338501 Chr.11: 66345986-66345986 on Build GRCh38
rs537869583 Chr.11: 66345760-66345760 on Build GRCh38
rs960876205 Chr.11: 66345801-66345801 on Build GRCh38
rs756119324 Chr.11: 66345694-66345696 on Build GRCh38
rs936685583 Chr.11: 66345907-66345907 on Build GRCh38
rs975907394 Chr.11: 66345855-66345855 on Build GRCh38
rs766917755 Chr.11: 66345937-66345937 on Build GRCh38
rs1043821704 Chr.11: 66345763-66345763 on Build GRCh38
rs1035193672 Chr.11: 66345794-66345794 on Build GRCh38
rs567111399 Chr.11: 66345575-66345575 on Build GRCh38
rs191735108 Chr.11: 66345586-66345586 on Build GRCh38

View Full Product Details