Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 78106657-78107048 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALG8
Entrez Gene ID
79053
Gene Name
ALG8, alpha-1,3-glucosyltransferase
Gene Aliases
CDG1H
UniGene
Hs.503368
Gene Chromosome Location
Chr.11: 78100942-78139660 on Build GRCh38
Transcripts
NM_001007027.2 NM_024079.4 XM_005274247.3 XM_011545252.2 XM_017018274.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5999846 c.1126T>C p.F376L Chr.11: 78106859-78106859 on Build GRCh38
1357151 c.1054C>A p.L352I Chr.11: 78106931-78106931 on Build GRCh38
7291715 c.1178+3A>G p.? Chr.11: 78106804-78106804 on Build GRCh38
5827054 c.1054C>A p.L352I Chr.11: 78106931-78106931 on Build GRCh38
309048 c.1091G>A p.R364Q Chr.11: 78106894-78106894 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751021064 Chr.11: 78106968-78106968 on Build GRCh38
rs943776989 Chr.11: 78106806-78106806 on Build GRCh38
rs367549562 Chr.11: 78106924-78106924 on Build GRCh38
rs368431889 Chr.11: 78106788-78106788 on Build GRCh38
rs878929156 Chr.11: 78106879-78106879 on Build GRCh38
rs777335080 Chr.11: 78106823-78106823 on Build GRCh38
rs375315639 Chr.11: 78106780-78106780 on Build GRCh38
rs1034748777 Chr.11: 78106725-78106725 on Build GRCh38
rs758100604 Chr.11: 78106759-78106759 on Build GRCh38
rs765044195 Chr.11: 78106811-78106811 on Build GRCh38
rs398124390 Chr.11: 78106807-78106807 on Build GRCh38
rs1005132694 Chr.11: 78106900-78106900 on Build GRCh38
rs759289772 Chr.11: 78106920-78106920 on Build GRCh38
rs776598783 Chr.11: 78106909-78106909 on Build GRCh38
rs772107842 Chr.11: 78106892-78106892 on Build GRCh38
rs535366536 Chr.11: 78106820-78106820 on Build GRCh38
rs771225674 Chr.11: 78106908-78106908 on Build GRCh38
rs763527492 Chr.11: 78106965-78106965 on Build GRCh38
rs397961874 Chr.11: 78106685-78106686 on Build GRCh38
rs977617976 Chr.11: 78106799-78106799 on Build GRCh38
rs756846211 Chr.11: 78106827-78106827 on Build GRCh38
rs907399574 Chr.11: 78106926-78106926 on Build GRCh38
rs958716703 Chr.11: 78106736-78106736 on Build GRCh38
rs867450807 Chr.11: 78106851-78106851 on Build GRCh38
rs3831385 Chr.11: 78106681-78106682 on Build GRCh38
rs780331751 Chr.11: 78106773-78106773 on Build GRCh38
rs777330036 Chr.11: 78106760-78106760 on Build GRCh38
rs780826167 Chr.11: 78106854-78106854 on Build GRCh38
rs372626745 Chr.11: 78106817-78106817 on Build GRCh38
rs193037393 Chr.11: 78106983-78106983 on Build GRCh38
rs12284153 Chr.11: 78106762-78106762 on Build GRCh38
rs779340536 Chr.11: 78106869-78106869 on Build GRCh38
rs558379335 Chr.11: 78106868-78106868 on Build GRCh38
rs773367764 Chr.11: 78106786-78106786 on Build GRCh38
rs766238263 Chr.11: 78106894-78106894 on Build GRCh38
rs749810326 Chr.11: 78106774-78106774 on Build GRCh38
rs749616227 Chr.11: 78106854-78106854 on Build GRCh38
rs748678669 Chr.11: 78106890-78106890 on Build GRCh38
rs751218300 Chr.11: 78106824-78106824 on Build GRCh38
rs528175233 Chr.11: 78107024-78107024 on Build GRCh38
rs760971527 Chr.11: 78106789-78106789 on Build GRCh38
rs67803998 Chr.11: 78106682-78106683 on Build GRCh38
rs761842418 Chr.11: 78106781-78106781 on Build GRCh38
rs370160938 Chr.11: 78106708-78106708 on Build GRCh38
rs772173503 Chr.11: 78106782-78106782 on Build GRCh38
rs112211908 Chr.11: 78106951-78106951 on Build GRCh38
rs919124478 Chr.11: 78106979-78106979 on Build GRCh38
rs752322090 Chr.11: 78106985-78106986 on Build GRCh38
rs370752318 Chr.11: 78106777-78106777 on Build GRCh38
rs1015501031 Chr.11: 78106917-78106917 on Build GRCh38
rs753624972 Chr.11: 78106804-78106804 on Build GRCh38
rs775346045 Chr.11: 78106931-78106931 on Build GRCh38
rs770212501 Chr.11: 78106772-78106772 on Build GRCh38
rs780419546 Chr.11: 78106847-78106847 on Build GRCh38
rs981586005 Chr.11: 78106686-78106686 on Build GRCh38
rs767200342 Chr.11: 78106998-78106998 on Build GRCh38
rs766278533 Chr.11: 78106800-78106800 on Build GRCh38
rs990114515 Chr.11: 78107005-78107005 on Build GRCh38
rs958789272 Chr.11: 78106994-78106994 on Build GRCh38
rs376161880 Chr.11: 78106895-78106895 on Build GRCh38
rs963287770 Chr.11: 78106944-78106944 on Build GRCh38

View Full Product Details