Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 70416751-70417293 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTTN
Entrez Gene ID
2017
Gene Name
cortactin
Gene Aliases
EMS1
UniGene
Hs.596164
Gene Chromosome Location
Chr.11: 70398506-70436584 on Build GRCh38
Transcripts
NM_001184740.1 NM_005231.3 NM_138565.2 XM_006718447.2 XM_006718448.3 XM_017017312.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5729793 c.568G>A p.D190N Chr.11: 70417123-70417123 on Build GRCh38
5729791 c.568G>A p.D190N Chr.11: 70417123-70417123 on Build GRCh38
7133397 c.566G>A p.R189K Chr.11: 70417121-70417121 on Build GRCh38
1628198 c.518C>T p.A173V Chr.11: 70417073-70417073 on Build GRCh38
1356662 c.546G>A p.T182T Chr.11: 70417101-70417101 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs900966823 Chr.11: 70416824-70416824 on Build GRCh38
rs376180283 Chr.11: 70417131-70417131 on Build GRCh38
rs1056606989 Chr.11: 70416939-70416939 on Build GRCh38
rs7479958 Chr.11: 70417165-70417165 on Build GRCh38
rs986350223 Chr.11: 70417176-70417176 on Build GRCh38
rs534730873 Chr.11: 70417084-70417084 on Build GRCh38
rs999315288 Chr.11: 70416833-70416833 on Build GRCh38
rs930904190 Chr.11: 70416780-70416780 on Build GRCh38
rs373399027 Chr.11: 70416935-70416938 on Build GRCh38
rs886876075 Chr.11: 70416942-70416942 on Build GRCh38
rs921161151 Chr.11: 70416799-70416799 on Build GRCh38
rs540085825 Chr.11: 70417216-70417216 on Build GRCh38
rs986749655 Chr.11: 70416935-70416935 on Build GRCh38
rs578175808 Chr.11: 70417044-70417044 on Build GRCh38
rs759451424 Chr.11: 70417058-70417058 on Build GRCh38
rs368152967 Chr.11: 70417162-70417162 on Build GRCh38
rs996532524 Chr.11: 70417206-70417206 on Build GRCh38
rs116287469 Chr.11: 70417208-70417208 on Build GRCh38
rs1003724026 Chr.11: 70416932-70416932 on Build GRCh38
rs982140676 Chr.11: 70416879-70416881 on Build GRCh38
rs766029603 Chr.11: 70416898-70416898 on Build GRCh38
rs750486491 Chr.11: 70417153-70417153 on Build GRCh38
rs1052401900 Chr.11: 70416862-70416862 on Build GRCh38
rs780364528 Chr.11: 70417033-70417033 on Build GRCh38
rs768093218 Chr.11: 70416995-70416995 on Build GRCh38
rs767545353 Chr.11: 70417066-70417066 on Build GRCh38
rs372529558 Chr.11: 70417139-70417139 on Build GRCh38
rs955915199 Chr.11: 70417250-70417250 on Build GRCh38
rs375532004 Chr.11: 70417159-70417159 on Build GRCh38
rs772820741 Chr.11: 70417110-70417110 on Build GRCh38
rs1027744891 Chr.11: 70417135-70417135 on Build GRCh38
rs375084241 Chr.11: 70417111-70417111 on Build GRCh38
rs1041158727 Chr.11: 70416823-70416823 on Build GRCh38
rs935376273 Chr.11: 70416860-70416860 on Build GRCh38
rs905437134 Chr.11: 70416922-70416922 on Build GRCh38
rs775520998 Chr.11: 70417140-70417140 on Build GRCh38
rs780161925 Chr.11: 70417067-70417067 on Build GRCh38
rs946691089 Chr.11: 70416889-70416889 on Build GRCh38
rs777767011 Chr.11: 70417017-70417017 on Build GRCh38
rs1050713884 Chr.11: 70416790-70416790 on Build GRCh38
rs781592658 Chr.11: 70416982-70416982 on Build GRCh38
rs760556391 Chr.11: 70417004-70417004 on Build GRCh38
rs773797219 Chr.11: 70417041-70417041 on Build GRCh38
rs139313464 Chr.11: 70417053-70417053 on Build GRCh38
rs371660033 Chr.11: 70417174-70417174 on Build GRCh38
rs1031532888 Chr.11: 70417248-70417248 on Build GRCh38
rs142406325 Chr.11: 70417274-70417274 on Build GRCh38
rs780452438 Chr.11: 70417074-70417074 on Build GRCh38
rs575895164 Chr.11: 70417207-70417207 on Build GRCh38
rs750640661 Chr.11: 70417032-70417032 on Build GRCh38
rs758425906 Chr.11: 70416972-70416972 on Build GRCh38
rs542706268 Chr.11: 70417101-70417101 on Build GRCh38
rs1011209862 Chr.11: 70416852-70416852 on Build GRCh38
rs139067402 Chr.11: 70417019-70417019 on Build GRCh38
rs747232917 Chr.11: 70416975-70416975 on Build GRCh38
rs771160064 Chr.11: 70416981-70416981 on Build GRCh38
rs893712633 Chr.11: 70416843-70416843 on Build GRCh38
rs755388651 Chr.11: 70417164-70417164 on Build GRCh38
rs778686501 Chr.11: 70417066-70417067 on Build GRCh38
rs753725306 Chr.11: 70416851-70416851 on Build GRCh38
rs146240174 Chr.11: 70416984-70416984 on Build GRCh38
rs141902583 Chr.11: 70416882-70416882 on Build GRCh38
rs766589155 Chr.11: 70417154-70417154 on Build GRCh38
rs201218623 Chr.11: 70417130-70417130 on Build GRCh38
rs749807110 Chr.11: 70417034-70417034 on Build GRCh38
rs149012607 Chr.11: 70417092-70417092 on Build GRCh38
rs112826323 Chr.11: 70417005-70417005 on Build GRCh38
rs750025515 Chr.11: 70417003-70417003 on Build GRCh38
rs769190568 Chr.11: 70417038-70417038 on Build GRCh38
rs878947388 Chr.11: 70417072-70417072 on Build GRCh38
rs145795527 Chr.11: 70417071-70417071 on Build GRCh38
rs549183259 Chr.11: 70416998-70416998 on Build GRCh38
rs767631532 Chr.11: 70417150-70417150 on Build GRCh38
rs775999193 Chr.11: 70416990-70416990 on Build GRCh38
rs371255550 Chr.11: 70416971-70416971 on Build GRCh38
rs73518261 Chr.11: 70416868-70416868 on Build GRCh38
rs777865914 Chr.11: 70416974-70416974 on Build GRCh38
rs1198234 Chr.11: 70417245-70417245 on Build GRCh38
rs200493275 Chr.11: 70417023-70417023 on Build GRCh38
rs751344033 Chr.11: 70417054-70417054 on Build GRCh38
rs939560580 Chr.11: 70417143-70417143 on Build GRCh38
rs200865893 Chr.11: 70417045-70417045 on Build GRCh38
rs752511791 Chr.11: 70417060-70417060 on Build GRCh38
rs748549354 Chr.11: 70417100-70417100 on Build GRCh38
rs757488650 Chr.11: 70417020-70417020 on Build GRCh38
rs762844602 Chr.11: 70417148-70417148 on Build GRCh38
rs898230143 Chr.11: 70417199-70417199 on Build GRCh38
rs557402754 Chr.11: 70416902-70416902 on Build GRCh38
rs571450390 Chr.11: 70416827-70416827 on Build GRCh38
rs770696412 Chr.11: 70417116-70417116 on Build GRCh38
rs773891758 Chr.11: 70416996-70416996 on Build GRCh38
rs776466770 Chr.11: 70417057-70417057 on Build GRCh38
rs569843993 Chr.11: 70416913-70416913 on Build GRCh38
rs781398142 Chr.11: 70417021-70417021 on Build GRCh38
rs770077457 Chr.11: 70416986-70416986 on Build GRCh38
rs757114671 Chr.11: 70416917-70416917 on Build GRCh38
rs955271276 Chr.11: 70416822-70416822 on Build GRCh38
rs370498578 Chr.11: 70417048-70417048 on Build GRCh38
rs942383786 Chr.11: 70416807-70416807 on Build GRCh38
rs1038161136 Chr.11: 70417175-70417175 on Build GRCh38
rs975383674 Chr.11: 70416793-70416793 on Build GRCh38
rs745630985 Chr.11: 70417159-70417160 on Build GRCh38
rs540048066 Chr.11: 70417237-70417237 on Build GRCh38
rs1030816783 Chr.11: 70416837-70416837 on Build GRCh38
rs757372494 Chr.11: 70417064-70417064 on Build GRCh38
rs754915765 Chr.11: 70417010-70417010 on Build GRCh38
rs946773526 Chr.11: 70417121-70417121 on Build GRCh38
rs767197227 Chr.11: 70417001-70417001 on Build GRCh38
rs986586612 Chr.11: 70416839-70416839 on Build GRCh38
rs755013677 Chr.11: 70417252-70417252 on Build GRCh38

View Full Product Details